Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6956896-6957471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE4P
Entrez Gene ID
326342
Gene Name
adhesion G protein-coupled receptor E4, pseudogene
Gene Aliases
EMR4, EMR4P, FIRE, GPR127, PGR16
UniGene
Hs.688043
Gene Chromosome Location
Chr.19: 6952500-6990846 on Build GRCh38
Transcripts
NR_024075.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6017165 c.1241C>T p.S414F Chr.19: 6957177-6957177 on Build GRCh38
5583479 c.1374G>A p.Q458Q Chr.19: 6957044-6957044 on Build GRCh38
5887037 c.1278G>A p.G426G Chr.19: 6957140-6957140 on Build GRCh38
4998485 c.1369C>T p.R457C Chr.19: 6957049-6957049 on Build GRCh38
5926020 c.1284G>A p.T428T Chr.19: 6957134-6957134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760953272 Chr.19: 6956999-6956999 on Build GRCh38
rs775873675 Chr.19: 6956998-6956998 on Build GRCh38
rs1008708711 Chr.19: 6957198-6957198 on Build GRCh38
rs755760963 Chr.19: 6957248-6957248 on Build GRCh38
rs75501826 Chr.19: 6957379-6957379 on Build GRCh38
rs992981224 Chr.19: 6957134-6957134 on Build GRCh38
rs751483975 Chr.19: 6957001-6957001 on Build GRCh38
rs925624256 Chr.19: 6957387-6957387 on Build GRCh38
rs756653820 Chr.19: 6957261-6957261 on Build GRCh38
rs566977417 Chr.19: 6957253-6957253 on Build GRCh38
rs904152118 Chr.19: 6957193-6957197 on Build GRCh38
rs933734289 Chr.19: 6957240-6957240 on Build GRCh38
rs541567854 Chr.19: 6957048-6957048 on Build GRCh38
rs960258711 Chr.19: 6957349-6957349 on Build GRCh38
rs978512604 Chr.19: 6957381-6957381 on Build GRCh38
rs753800324 Chr.19: 6957423-6957423 on Build GRCh38
rs553537308 Chr.19: 6957435-6957435 on Build GRCh38
rs1008273659 Chr.19: 6957011-6957011 on Build GRCh38
rs546653927 Chr.19: 6957208-6957208 on Build GRCh38
rs552581925 Chr.19: 6957255-6957255 on Build GRCh38
rs561571784 Chr.19: 6957069-6957069 on Build GRCh38
rs983999553 Chr.19: 6957173-6957173 on Build GRCh38
rs563656095 Chr.19: 6957353-6957353 on Build GRCh38
rs530117066 Chr.19: 6957094-6957094 on Build GRCh38
rs913357697 Chr.19: 6957346-6957346 on Build GRCh38
rs891980276 Chr.19: 6956991-6956991 on Build GRCh38
rs1038851299 Chr.19: 6957426-6957426 on Build GRCh38
rs569446213 Chr.19: 6957297-6957297 on Build GRCh38
rs752388201 Chr.19: 6957194-6957194 on Build GRCh38
rs538609328 Chr.19: 6957254-6957254 on Build GRCh38
rs542266541 Chr.19: 6957220-6957220 on Build GRCh38
rs547976754 Chr.19: 6957049-6957049 on Build GRCh38
rs944618231 Chr.19: 6957415-6957415 on Build GRCh38
rs761516469 Chr.19: 6957145-6957147 on Build GRCh38
rs761233320 Chr.19: 6957165-6957165 on Build GRCh38
rs181051303 Chr.19: 6957034-6957034 on Build GRCh38
rs899960936 Chr.19: 6957215-6957215 on Build GRCh38
rs938801892 Chr.19: 6957104-6957104 on Build GRCh38
rs900293734 Chr.19: 6957428-6957428 on Build GRCh38
rs951201584 Chr.19: 6957154-6957154 on Build GRCh38
rs915684837 Chr.19: 6957135-6957135 on Build GRCh38
rs557971719 Chr.19: 6957412-6957412 on Build GRCh38
rs115225855 Chr.19: 6957399-6957399 on Build GRCh38
rs773986672 Chr.19: 6956955-6956955 on Build GRCh38
rs1016901247 Chr.19: 6957178-6957178 on Build GRCh38
rs773871152 Chr.19: 6957140-6957140 on Build GRCh38
rs946255810 Chr.19: 6957352-6957352 on Build GRCh38
rs946275945 Chr.19: 6957013-6957013 on Build GRCh38
rs1052939838 Chr.19: 6957196-6957196 on Build GRCh38
rs553201347 Chr.19: 6957053-6957053 on Build GRCh38
rs768893290 Chr.19: 6957025-6957025 on Build GRCh38
rs113583180 Chr.19: 6957386-6957386 on Build GRCh38
rs371399892 Chr.19: 6957345-6957345 on Build GRCh38
rs916049690 Chr.19: 6957362-6957362 on Build GRCh38
rs996906688 Chr.19: 6957235-6957235 on Build GRCh38
rs1014771737 Chr.19: 6957092-6957092 on Build GRCh38
rs952718135 Chr.19: 6957277-6957277 on Build GRCh38
rs975106629 Chr.19: 6957205-6957205 on Build GRCh38
rs759401878 Chr.19: 6957076-6957076 on Build GRCh38
rs779505873 Chr.19: 6957264-6957264 on Build GRCh38
rs1052701998 Chr.19: 6957406-6957406 on Build GRCh38
rs975858606 Chr.19: 6957052-6957052 on Build GRCh38
rs988311692 Chr.19: 6957291-6957291 on Build GRCh38
rs948493058 Chr.19: 6957157-6957158 on Build GRCh38
rs767277896 Chr.19: 6957099-6957099 on Build GRCh38
rs574842466 Chr.19: 6957101-6957101 on Build GRCh38
rs890246586 Chr.19: 6956986-6956986 on Build GRCh38
rs546115608 Chr.19: 6957436-6957436 on Build GRCh38
rs1051963516 Chr.19: 6956961-6956961 on Build GRCh38

View Full Product Details