Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10052221-10052770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3P1
Entrez Gene ID
388503
Gene Name
complement component 3 precursor pseudogene
Gene Aliases
CPLP
UniGene
Hs.655363
Gene Chromosome Location
Chr.19: 10041356-10074138 on Build GRCh38
Transcripts
NR_027300.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1750608 c.481G>A p.A161T Chr.19: 10052591-10052591 on Build GRCh38
7115515 c.478G>A p.E160K Chr.19: 10052588-10052588 on Build GRCh38
4445502 c.409G>A p.E137K Chr.19: 10052344-10052344 on Build GRCh38
1303904 c.409G>C p.E137Q Chr.19: 10052344-10052344 on Build GRCh38
7176178 c.476C>T p.T159M Chr.19: 10052586-10052586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771140974 Chr.19: 10052646-10052646 on Build GRCh38
rs897954843 Chr.19: 10052414-10052414 on Build GRCh38
rs757905003 Chr.19: 10052284-10052284 on Build GRCh38
rs764418911 Chr.19: 10052256-10052256 on Build GRCh38
rs73922311 Chr.19: 10052704-10052704 on Build GRCh38
rs957992842 Chr.19: 10052746-10052746 on Build GRCh38
rs781080957 Chr.19: 10052612-10052612 on Build GRCh38
rs913991018 Chr.19: 10052527-10052527 on Build GRCh38
rs550920830 Chr.19: 10052695-10052695 on Build GRCh38
rs768565262 Chr.19: 10052405-10052405 on Build GRCh38
rs747976632 Chr.19: 10052378-10052378 on Build GRCh38
rs754920739 Chr.19: 10052307-10052307 on Build GRCh38
rs765516450 Chr.19: 10052349-10052349 on Build GRCh38
rs756099143 Chr.19: 10052590-10052590 on Build GRCh38
rs556658502 Chr.19: 10052396-10052396 on Build GRCh38
rs759062592 Chr.19: 10052651-10052651 on Build GRCh38
rs971404023 Chr.19: 10052458-10052458 on Build GRCh38
rs993671410 Chr.19: 10052418-10052418 on Build GRCh38
rs770161612 Chr.19: 10052529-10052529 on Build GRCh38
rs370919452 Chr.19: 10052653-10052653 on Build GRCh38
rs73506927 Chr.19: 10052596-10052596 on Build GRCh38
rs773540534 Chr.19: 10052535-10052535 on Build GRCh38
rs561300402 Chr.19: 10052579-10052579 on Build GRCh38
rs747101479 Chr.19: 10052400-10052400 on Build GRCh38
rs750018001 Chr.19: 10052279-10052279 on Build GRCh38
rs756006026 Chr.19: 10052389-10052389 on Build GRCh38
rs372908491 Chr.19: 10052650-10052650 on Build GRCh38
rs140514229 Chr.19: 10052247-10052247 on Build GRCh38
rs879154255 Chr.19: 10052745-10052745 on Build GRCh38
rs781173901 Chr.19: 10052377-10052377 on Build GRCh38
rs531964381 Chr.19: 10052586-10052586 on Build GRCh38
rs751124650 Chr.19: 10052703-10052703 on Build GRCh38
rs138333377 Chr.19: 10052656-10052656 on Build GRCh38
rs750900323 Chr.19: 10052638-10052638 on Build GRCh38
rs747969592 Chr.19: 10052308-10052308 on Build GRCh38
rs752679164 Chr.19: 10052465-10052465 on Build GRCh38
rs764858802 Chr.19: 10052278-10052278 on Build GRCh38
rs768072297 Chr.19: 10052559-10052561 on Build GRCh38
rs750104668 Chr.19: 10052356-10052356 on Build GRCh38
rs777751860 Chr.19: 10052592-10052592 on Build GRCh38
rs545378288 Chr.19: 10052461-10052461 on Build GRCh38
rs753685248 Chr.19: 10052593-10052593 on Build GRCh38
rs766050003 Chr.19: 10052548-10052548 on Build GRCh38
rs945292805 Chr.19: 10052287-10052287 on Build GRCh38
rs754202909 Chr.19: 10052259-10052259 on Build GRCh38
rs369210538 Chr.19: 10052327-10052327 on Build GRCh38
rs377187806 Chr.19: 10052342-10052342 on Build GRCh38
rs777128028 Chr.19: 10052690-10052690 on Build GRCh38
rs906773551 Chr.19: 10052675-10052675 on Build GRCh38
rs956498868 Chr.19: 10052282-10052282 on Build GRCh38
rs555235738 Chr.19: 10052314-10052314 on Build GRCh38
rs770251225 Chr.19: 10052625-10052625 on Build GRCh38
rs190773945 Chr.19: 10052283-10052283 on Build GRCh38
rs765130846 Chr.19: 10052460-10052460 on Build GRCh38
rs377676586 Chr.19: 10052607-10052607 on Build GRCh38
rs372496665 Chr.19: 10052331-10052331 on Build GRCh38
rs762181185 Chr.19: 10052347-10052347 on Build GRCh38
rs371827491 Chr.19: 10052670-10052670 on Build GRCh38
rs767738739 Chr.19: 10052260-10052260 on Build GRCh38
rs375245456 Chr.19: 10052598-10052598 on Build GRCh38
rs544365576 Chr.19: 10052338-10052338 on Build GRCh38
rs182184425 Chr.19: 10052451-10052451 on Build GRCh38
rs761130924 Chr.19: 10052334-10052334 on Build GRCh38
rs370047759 Chr.19: 10052525-10052525 on Build GRCh38
rs1036528309 Chr.19: 10052407-10052407 on Build GRCh38
rs1022583315 Chr.19: 10052587-10052587 on Build GRCh38
rs757082463 Chr.19: 10052667-10052667 on Build GRCh38
rs754937908 Chr.19: 10052375-10052375 on Build GRCh38
rs760762866 Chr.19: 10052657-10052657 on Build GRCh38
rs766899070 Chr.19: 10052583-10052583 on Build GRCh38
rs765852914 Chr.19: 10052372-10052372 on Build GRCh38
rs762633778 Chr.19: 10052541-10052541 on Build GRCh38
rs560493388 Chr.19: 10052462-10052462 on Build GRCh38
rs775503538 Chr.19: 10052397-10052397 on Build GRCh38
rs754763789 Chr.19: 10052439-10052439 on Build GRCh38
rs751048847 Chr.19: 10052374-10052374 on Build GRCh38
rs1033640455 Chr.19: 10052705-10052705 on Build GRCh38
rs757992914 Chr.19: 10052367-10052367 on Build GRCh38
rs879206526 Chr.19: 10052457-10052457 on Build GRCh38
rs45586031 Chr.19: 10052325-10052325 on Build GRCh38
rs368555915 Chr.19: 10052661-10052661 on Build GRCh38
rs777663966 Chr.19: 10052390-10052390 on Build GRCh38
rs1047026469 Chr.19: 10052669-10052669 on Build GRCh38
rs746938367 Chr.19: 10052306-10052306 on Build GRCh38
rs777196458 Chr.19: 10052488-10052488 on Build GRCh38
rs763290061 Chr.19: 10052639-10052639 on Build GRCh38
rs543328575 Chr.19: 10052475-10052475 on Build GRCh38
rs779495000 Chr.19: 10052292-10052292 on Build GRCh38
rs1040928786 Chr.19: 10052288-10052288 on Build GRCh38
rs373881303 Chr.19: 10052472-10052472 on Build GRCh38
rs773632540 Chr.19: 10052638-10052638 on Build GRCh38
rs774720689 Chr.19: 10052323-10052323 on Build GRCh38
rs751086344 Chr.19: 10052575-10052575 on Build GRCh38
rs762265072 Chr.19: 10052490-10052490 on Build GRCh38
rs758470322 Chr.19: 10052674-10052674 on Build GRCh38
rs748698012 Chr.19: 10052618-10052618 on Build GRCh38

View Full Product Details