Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10058465-10059007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3P1
Entrez Gene ID
388503
Gene Name
complement component 3 precursor pseudogene
Gene Aliases
CPLP
UniGene
Hs.655363
Gene Chromosome Location
Chr.19: 10041356-10074138 on Build GRCh38
Transcripts
NR_027300.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7244195 c.768C>T p.S256S Chr.19: 10058585-10058585 on Build GRCh38
417783 c.853A>G p.S285G Chr.19: 10058822-10058822 on Build GRCh38
4932849 c.964A>G p.N322D Chr.19: 10058933-10058933 on Build GRCh38
6083904 c.989G>T p.S330I Chr.19: 10058958-10058958 on Build GRCh38
6083905 c.840+2T>A p.? Chr.19: 10058659-10058659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576225484 Chr.19: 10058761-10058761 on Build GRCh38
rs528033903 Chr.19: 10058901-10058901 on Build GRCh38
rs780052095 Chr.19: 10058694-10058694 on Build GRCh38
rs764403145 Chr.19: 10058651-10058651 on Build GRCh38
rs16998102 Chr.19: 10058878-10058878 on Build GRCh38
rs1002379682 Chr.19: 10058976-10058976 on Build GRCh38
rs368893265 Chr.19: 10058552-10058552 on Build GRCh38
rs188365235 Chr.19: 10058629-10058629 on Build GRCh38
rs567459535 Chr.19: 10058546-10058546 on Build GRCh38
rs764852533 Chr.19: 10058815-10058815 on Build GRCh38
rs527339453 Chr.19: 10058864-10058864 on Build GRCh38
rs985177774 Chr.19: 10058786-10058786 on Build GRCh38
rs531648670 Chr.19: 10058936-10058936 on Build GRCh38
rs747082616 Chr.19: 10058944-10058944 on Build GRCh38
rs759122161 Chr.19: 10058969-10058969 on Build GRCh38
rs553397199 Chr.19: 10058828-10058829 on Build GRCh38
rs771246248 Chr.19: 10058776-10058776 on Build GRCh38
rs760000989 Chr.19: 10058631-10058631 on Build GRCh38
rs751382738 Chr.19: 10058704-10058704 on Build GRCh38
rs780675171 Chr.19: 10058948-10058948 on Build GRCh38
rs746886165 Chr.19: 10058586-10058588 on Build GRCh38
rs751001884 Chr.19: 10058909-10058909 on Build GRCh38
rs756794896 Chr.19: 10058577-10058577 on Build GRCh38
rs372351443 Chr.19: 10058840-10058840 on Build GRCh38
rs775803753 Chr.19: 10058638-10058638 on Build GRCh38
rs373607404 Chr.19: 10058795-10058795 on Build GRCh38
rs756006099 Chr.19: 10058929-10058929 on Build GRCh38
rs369131259 Chr.19: 10058964-10058964 on Build GRCh38
rs751535533 Chr.19: 10058837-10058837 on Build GRCh38
rs139436763 Chr.19: 10058565-10058565 on Build GRCh38
rs376707855 Chr.19: 10058935-10058935 on Build GRCh38
rs370195069 Chr.19: 10058693-10058693 on Build GRCh38
rs199606350 Chr.19: 10058538-10058538 on Build GRCh38
rs773481691 Chr.19: 10058542-10058542 on Build GRCh38
rs536933915 Chr.19: 10058686-10058686 on Build GRCh38
rs373498985 Chr.19: 10058650-10058650 on Build GRCh38
rs767820770 Chr.19: 10058551-10058551 on Build GRCh38
rs11667703 Chr.19: 10058585-10058585 on Build GRCh38
rs748611850 Chr.19: 10058955-10058955 on Build GRCh38
rs922304188 Chr.19: 10058746-10058746 on Build GRCh38
rs966617541 Chr.19: 10058918-10058918 on Build GRCh38
rs779776684 Chr.19: 10058593-10058593 on Build GRCh38
rs761524604 Chr.19: 10058807-10058807 on Build GRCh38
rs775027210 Chr.19: 10058549-10058549 on Build GRCh38
rs377208220 Chr.19: 10058550-10058550 on Build GRCh38
rs776953596 Chr.19: 10058805-10058805 on Build GRCh38
rs779193583 Chr.19: 10058777-10058777 on Build GRCh38
rs763992112 Chr.19: 10058975-10058975 on Build GRCh38
rs754831806 Chr.19: 10058707-10058707 on Build GRCh38
rs1993466 Chr.19: 10058569-10058569 on Build GRCh38
rs372410117 Chr.19: 10058545-10058545 on Build GRCh38
rs375870801 Chr.19: 10058884-10058884 on Build GRCh38
rs376835782 Chr.19: 10058624-10058624 on Build GRCh38
rs4552116 Chr.19: 10058602-10058602 on Build GRCh38
rs749008077 Chr.19: 10058962-10058962 on Build GRCh38
rs753514919 Chr.19: 10058653-10058653 on Build GRCh38
rs746038977 Chr.19: 10058858-10058858 on Build GRCh38
rs181092055 Chr.19: 10058827-10058827 on Build GRCh38
rs765401168 Chr.19: 10058947-10058947 on Build GRCh38
rs773580266 Chr.19: 10058797-10058797 on Build GRCh38
rs376376727 Chr.19: 10058676-10058676 on Build GRCh38
rs749570555 Chr.19: 10058768-10058768 on Build GRCh38
rs370635027 Chr.19: 10058804-10058804 on Build GRCh38
rs753573482 Chr.19: 10058978-10058978 on Build GRCh38
rs372462550 Chr.19: 10058531-10058531 on Build GRCh38
rs767576627 Chr.19: 10058912-10058912 on Build GRCh38
rs779977360 Chr.19: 10058863-10058863 on Build GRCh38
rs745761038 Chr.19: 10058858-10058858 on Build GRCh38
rs764772161 Chr.19: 10058684-10058684 on Build GRCh38
rs201701007 Chr.19: 10058875-10058875 on Build GRCh38
rs576126685 Chr.19: 10058724-10058724 on Build GRCh38
rs758329435 Chr.19: 10058589-10058589 on Build GRCh38
rs748363021 Chr.19: 10058536-10058536 on Build GRCh38
rs1037128836 Chr.19: 10058932-10058932 on Build GRCh38
rs767096431 Chr.19: 10058970-10058970 on Build GRCh38
rs1046415380 Chr.19: 10058573-10058573 on Build GRCh38
rs763822549 Chr.19: 10058568-10058568 on Build GRCh38
rs768275389 Chr.19: 10058597-10058597 on Build GRCh38
rs765961531 Chr.19: 10058895-10058895 on Build GRCh38
rs925095587 Chr.19: 10058861-10058861 on Build GRCh38
rs11667515 Chr.19: 10058801-10058801 on Build GRCh38
rs750709221 Chr.19: 10058931-10058931 on Build GRCh38
rs144140729 Chr.19: 10058828-10058828 on Build GRCh38
rs371665216 Chr.19: 10058885-10058885 on Build GRCh38
rs1021274005 Chr.19: 10058632-10058632 on Build GRCh38
rs759613838 Chr.19: 10058910-10058910 on Build GRCh38
rs770163470 Chr.19: 10058960-10058960 on Build GRCh38
rs559915393 Chr.19: 10058506-10058506 on Build GRCh38
rs1034348457 Chr.19: 10058816-10058816 on Build GRCh38
rs776097590 Chr.19: 10058798-10058798 on Build GRCh38
rs770568546 Chr.19: 10058809-10058814 on Build GRCh38
rs184804114 Chr.19: 10058849-10058849 on Build GRCh38
rs17211059 Chr.19: 10058532-10058532 on Build GRCh38
rs778348088 Chr.19: 10058582-10058582 on Build GRCh38
rs749982045 Chr.19: 10058826-10058826 on Build GRCh38
rs781172690 Chr.19: 10058838-10058838 on Build GRCh38
rs769897471 Chr.19: 10058877-10058877 on Build GRCh38
rs367910891 Chr.19: 10058523-10058523 on Build GRCh38
rs777041700 Chr.19: 10058874-10058874 on Build GRCh38
rs976440846 Chr.19: 10058517-10058517 on Build GRCh38
rs770484770 Chr.19: 10058963-10058963 on Build GRCh38
rs781665341 Chr.19: 10058949-10058949 on Build GRCh38
rs369148845 Chr.19: 10058586-10058586 on Build GRCh38
rs979271945 Chr.19: 10058567-10058567 on Build GRCh38
rs746802496 Chr.19: 10058596-10058596 on Build GRCh38
rs371800310 Chr.19: 10058973-10058973 on Build GRCh38
rs752615728 Chr.19: 10058920-10058920 on Build GRCh38
rs773425061 Chr.19: 10058961-10058961 on Build GRCh38
rs768056075 Chr.19: 10058635-10058635 on Build GRCh38
rs760669127 Chr.19: 10058974-10058974 on Build GRCh38

View Full Product Details