Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40938621-40939177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2B7P
Entrez Gene ID
1556
Gene Name
cytochrome P450 family 2 subfamily B member 7, pseudogene
Gene Aliases
CYP2B, CYP2B7, CYP2B7P1
UniGene
Hs.529117
Gene Chromosome Location
Chr.19: 40924265-40950660 on Build GRCh38
Transcripts
NR_001278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3892617 c.564C>T p.F188F Chr.19: 40939048-40939048 on Build GRCh38
6151004 c.645+1G>T p.? Chr.19: 40939130-40939130 on Build GRCh38
3990048 c.524C>A p.T175N Chr.19: 40939008-40939008 on Build GRCh38
4837186 c.643C>G p.Q215E Chr.19: 40939127-40939127 on Build GRCh38
5907007 c.547A>G p.I183V Chr.19: 40939031-40939031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746854288 Chr.19: 40939045-40939045 on Build GRCh38
rs267605492 Chr.19: 40938991-40938991 on Build GRCh38
rs138116565 Chr.19: 40938837-40938837 on Build GRCh38
rs755376032 Chr.19: 40939009-40939009 on Build GRCh38
rs749972872 Chr.19: 40939157-40939157 on Build GRCh38
rs750653946 Chr.19: 40938925-40938925 on Build GRCh38
rs573438754 Chr.19: 40938977-40938977 on Build GRCh38
rs562995260 Chr.19: 40939077-40939077 on Build GRCh38
rs755575554 Chr.19: 40938937-40938937 on Build GRCh38
rs58418464 Chr.19: 40939028-40939028 on Build GRCh38
rs890796217 Chr.19: 40938756-40938756 on Build GRCh38
rs551332973 Chr.19: 40938822-40938822 on Build GRCh38
rs4803407 Chr.19: 40938833-40938833 on Build GRCh38
rs17609461 Chr.19: 40939006-40939006 on Build GRCh38
rs754331515 Chr.19: 40939072-40939073 on Build GRCh38
rs752818920 Chr.19: 40939094-40939094 on Build GRCh38
rs927266592 Chr.19: 40938838-40938838 on Build GRCh38
rs745517047 Chr.19: 40938951-40938951 on Build GRCh38
rs267605493 Chr.19: 40939081-40939081 on Build GRCh38
rs773374558 Chr.19: 40939078-40939078 on Build GRCh38
rs540015728 Chr.19: 40938950-40938950 on Build GRCh38
rs567290039 Chr.19: 40938904-40938904 on Build GRCh38
rs8112176 Chr.19: 40938970-40938970 on Build GRCh38
rs766765261 Chr.19: 40938790-40938792 on Build GRCh38
rs781775705 Chr.19: 40939010-40939010 on Build GRCh38
rs756772023 Chr.19: 40939027-40939027 on Build GRCh38
rs779631804 Chr.19: 40939044-40939044 on Build GRCh38
rs753948518 Chr.19: 40938921-40938921 on Build GRCh38
rs61730277 Chr.19: 40939015-40939015 on Build GRCh38
rs752949517 Chr.19: 40939129-40939129 on Build GRCh38
rs751796560 Chr.19: 40939109-40939109 on Build GRCh38
rs761864982 Chr.19: 40938974-40938974 on Build GRCh38
rs756652043 Chr.19: 40939131-40939131 on Build GRCh38
rs1024863627 Chr.19: 40938827-40938827 on Build GRCh38
rs765195936 Chr.19: 40938956-40938957 on Build GRCh38
rs781747657 Chr.19: 40938941-40938941 on Build GRCh38
rs765299602 Chr.19: 40939043-40939043 on Build GRCh38
rs760420775 Chr.19: 40938957-40938957 on Build GRCh38
rs769239845 Chr.19: 40938742-40938742 on Build GRCh38
rs763080912 Chr.19: 40939087-40939087 on Build GRCh38
rs751998227 Chr.19: 40939008-40939008 on Build GRCh38
rs767454777 Chr.19: 40938864-40938864 on Build GRCh38
rs201696770 Chr.19: 40938733-40938733 on Build GRCh38
rs540196036 Chr.19: 40938644-40938644 on Build GRCh38
rs548949944 Chr.19: 40938851-40938851 on Build GRCh38
rs1002114624 Chr.19: 40939140-40939140 on Build GRCh38
rs544315587 Chr.19: 40938766-40938766 on Build GRCh38
rs571304003 Chr.19: 40938931-40938931 on Build GRCh38
rs555307383 Chr.19: 40939000-40939000 on Build GRCh38
rs768603069 Chr.19: 40938959-40938959 on Build GRCh38
rs561751319 Chr.19: 40938671-40938671 on Build GRCh38
rs766760154 Chr.19: 40939102-40939102 on Build GRCh38
rs529321623 Chr.19: 40938716-40938716 on Build GRCh38
rs760039720 Chr.19: 40939118-40939118 on Build GRCh38
rs573683824 Chr.19: 40939031-40939031 on Build GRCh38
rs534244606 Chr.19: 40938979-40938979 on Build GRCh38
rs937389802 Chr.19: 40938659-40938659 on Build GRCh38
rs751908345 Chr.19: 40938936-40938936 on Build GRCh38
rs758182933 Chr.19: 40939015-40939017 on Build GRCh38
rs556275708 Chr.19: 40938928-40938928 on Build GRCh38
rs748802437 Chr.19: 40938942-40938942 on Build GRCh38
rs764470033 Chr.19: 40939138-40939138 on Build GRCh38
rs752632326 Chr.19: 40938972-40938972 on Build GRCh38
rs544227447 Chr.19: 40939051-40939051 on Build GRCh38
rs1017875553 Chr.19: 40938764-40938764 on Build GRCh38
rs753239897 Chr.19: 40939018-40939018 on Build GRCh38
rs771651646 Chr.19: 40938954-40938954 on Build GRCh38
rs927339982 Chr.19: 40938651-40938651 on Build GRCh38
rs898659561 Chr.19: 40938728-40938728 on Build GRCh38
rs759646863 Chr.19: 40938976-40938976 on Build GRCh38
rs776490959 Chr.19: 40939050-40939050 on Build GRCh38
rs770448472 Chr.19: 40938947-40938947 on Build GRCh38
rs777476299 Chr.19: 40939054-40939054 on Build GRCh38
rs112130173 Chr.19: 40938987-40938987 on Build GRCh38
rs533128048 Chr.19: 40938821-40938821 on Build GRCh38
rs974154419 Chr.19: 40938903-40938903 on Build GRCh38
rs537904629 Chr.19: 40938923-40938923 on Build GRCh38
rs768431663 Chr.19: 40938826-40938826 on Build GRCh38
rs1007868790 Chr.19: 40938757-40938757 on Build GRCh38
rs1000560648 Chr.19: 40938770-40938770 on Build GRCh38
rs112819506 Chr.19: 40938820-40938820 on Build GRCh38
rs767914961 Chr.19: 40939124-40939124 on Build GRCh38
rs1038578124 Chr.19: 40938696-40938696 on Build GRCh38
rs930262540 Chr.19: 40938732-40938732 on Build GRCh38
rs773268435 Chr.19: 40938690-40938690 on Build GRCh38

View Full Product Details