Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40943885-40944449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2B7P
Entrez Gene ID
1556
Gene Name
cytochrome P450 family 2 subfamily B member 7, pseudogene
Gene Aliases
CYP2B, CYP2B7, CYP2B7P1
UniGene
Hs.529117
Gene Chromosome Location
Chr.19: 40924265-40950660 on Build GRCh38
Transcripts
NR_001278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4803381 c.1051G>A p.A351T Chr.19: 40944417-40944417 on Build GRCh38
996876 c.1073G>T p.R358I Chr.19: 40944439-40944439 on Build GRCh38
7096975 c.1028G>A p.R343Q Chr.19: 40944394-40944394 on Build GRCh38
5713516 c.965-2_966delAGAG p.? Chr.19: 40944329-40944332 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs937647044 Chr.19: 40944269-40944269 on Build GRCh38
rs756694604 Chr.19: 40944321-40944321 on Build GRCh38
rs542957879 Chr.19: 40944313-40944313 on Build GRCh38
rs780005437 Chr.19: 40944119-40944119 on Build GRCh38
rs971443577 Chr.19: 40944255-40944255 on Build GRCh38
rs775870595 Chr.19: 40944418-40944418 on Build GRCh38
rs925909390 Chr.19: 40943985-40943986 on Build GRCh38
rs375806976 Chr.19: 40944387-40944387 on Build GRCh38
rs776400272 Chr.19: 40944343-40944343 on Build GRCh38
rs974721275 Chr.19: 40944276-40944276 on Build GRCh38
rs757989040 Chr.19: 40944327-40944327 on Build GRCh38
rs190966934 Chr.19: 40944383-40944383 on Build GRCh38
rs754869717 Chr.19: 40944394-40944394 on Build GRCh38
rs779830621 Chr.19: 40944328-40944328 on Build GRCh38
rs957519670 Chr.19: 40943987-40943987 on Build GRCh38
rs554011946 Chr.19: 40944274-40944274 on Build GRCh38
rs761176725 Chr.19: 40944426-40944426 on Build GRCh38
rs555817104 Chr.19: 40944203-40944203 on Build GRCh38
rs763467332 Chr.19: 40944284-40944284 on Build GRCh38
rs749869160 Chr.19: 40944374-40944374 on Build GRCh38
rs759070114 Chr.19: 40944305-40944306 on Build GRCh38
rs543447254 Chr.19: 40944370-40944370 on Build GRCh38
rs763268186 Chr.19: 40944366-40944366 on Build GRCh38
rs769979791 Chr.19: 40944350-40944350 on Build GRCh38
rs572342246 Chr.19: 40944306-40944306 on Build GRCh38
rs181108879 Chr.19: 40944216-40944216 on Build GRCh38
rs561265893 Chr.19: 40944358-40944358 on Build GRCh38
rs753143304 Chr.19: 40944372-40944372 on Build GRCh38
rs1018128449 Chr.19: 40943940-40943940 on Build GRCh38
rs148893750 Chr.19: 40944191-40944191 on Build GRCh38
rs950179608 Chr.19: 40944010-40944010 on Build GRCh38
rs748048377 Chr.19: 40944390-40944390 on Build GRCh38
rs182789621 Chr.19: 40944395-40944395 on Build GRCh38
rs185974238 Chr.19: 40944008-40944008 on Build GRCh38
rs1018492865 Chr.19: 40944142-40944142 on Build GRCh38
rs767130695 Chr.19: 40944018-40944018 on Build GRCh38
rs567535837 Chr.19: 40944174-40944174 on Build GRCh38
rs567948411 Chr.19: 40944373-40944373 on Build GRCh38
rs758078603 Chr.19: 40944381-40944381 on Build GRCh38
rs372217638 Chr.19: 40943936-40943936 on Build GRCh38
rs1046784332 Chr.19: 40944083-40944083 on Build GRCh38
rs541163241 Chr.19: 40944314-40944316 on Build GRCh38
rs3852875 Chr.19: 40944017-40944017 on Build GRCh38
rs533341261 Chr.19: 40943984-40943984 on Build GRCh38
rs937555585 Chr.19: 40944039-40944039 on Build GRCh38
rs769783909 Chr.19: 40944391-40944391 on Build GRCh38
rs774408378 Chr.19: 40944401-40944401 on Build GRCh38
rs898151679 Chr.19: 40944072-40944072 on Build GRCh38
rs778605746 Chr.19: 40944322-40944322 on Build GRCh38
rs919630511 Chr.19: 40944289-40944289 on Build GRCh38
rs538264288 Chr.19: 40944183-40944183 on Build GRCh38
rs760014633 Chr.19: 40944403-40944403 on Build GRCh38
rs964279623 Chr.19: 40943944-40943944 on Build GRCh38
rs988888370 Chr.19: 40943993-40943993 on Build GRCh38
rs181713892 Chr.19: 40943915-40943915 on Build GRCh38
rs561947664 Chr.19: 40944111-40944111 on Build GRCh38
rs531082032 Chr.19: 40944267-40944267 on Build GRCh38
rs566780502 Chr.19: 40944023-40944023 on Build GRCh38
rs753440492 Chr.19: 40944317-40944317 on Build GRCh38
rs927414306 Chr.19: 40944268-40944268 on Build GRCh38
rs186096524 Chr.19: 40944382-40944382 on Build GRCh38
rs766861126 Chr.19: 40944302-40944302 on Build GRCh38
rs747143933 Chr.19: 40944051-40944051 on Build GRCh38
rs745509717 Chr.19: 40944326-40944326 on Build GRCh38
rs555309317 Chr.19: 40944085-40944085 on Build GRCh38
rs913431700 Chr.19: 40944006-40944006 on Build GRCh38
rs746742050 Chr.19: 40944335-40944335 on Build GRCh38
rs974782321 Chr.19: 40943959-40943959 on Build GRCh38
rs768661283 Chr.19: 40944342-40944342 on Build GRCh38
rs114966387 Chr.19: 40944367-40944367 on Build GRCh38
rs778026724 Chr.19: 40944393-40944393 on Build GRCh38
rs748327172 Chr.19: 40944347-40944347 on Build GRCh38
rs569883348 Chr.19: 40944088-40944088 on Build GRCh38
rs1008031369 Chr.19: 40944106-40944106 on Build GRCh38
rs892365505 Chr.19: 40944340-40944340 on Build GRCh38
rs112735272 Chr.19: 40943919-40943919 on Build GRCh38
rs772314406 Chr.19: 40944408-40944408 on Build GRCh38
rs1001191080 Chr.19: 40944150-40944150 on Build GRCh38
rs188731302 Chr.19: 40944057-40944057 on Build GRCh38
rs778611688 Chr.19: 40944148-40944149 on Build GRCh38
rs993910716 Chr.19: 40944077-40944077 on Build GRCh38
rs777968954 Chr.19: 40944398-40944398 on Build GRCh38
rs571368494 Chr.19: 40943952-40943952 on Build GRCh38
rs751802103 Chr.19: 40944303-40944303 on Build GRCh38
rs1033050169 Chr.19: 40944186-40944186 on Build GRCh38
rs913338875 Chr.19: 40944209-40944209 on Build GRCh38

View Full Product Details