Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40944318-40944872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2B7P
Entrez Gene ID
1556
Gene Name
cytochrome P450 family 2 subfamily B member 7, pseudogene
Gene Aliases
CYP2B, CYP2B7, CYP2B7P1
UniGene
Hs.529117
Gene Chromosome Location
Chr.19: 40924265-40950660 on Build GRCh38
Transcripts
NR_001278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4803381 c.1051G>A p.A351T Chr.19: 40944417-40944417 on Build GRCh38
996876 c.1073G>T p.R358I Chr.19: 40944439-40944439 on Build GRCh38
6338738 c.1111G>A p.V371I Chr.19: 40944477-40944477 on Build GRCh38
5933792 c.1089C>T p.L363L Chr.19: 40944455-40944455 on Build GRCh38
7096975 c.1028G>A p.R343Q Chr.19: 40944394-40944394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375806976 Chr.19: 40944387-40944387 on Build GRCh38
rs776400272 Chr.19: 40944343-40944343 on Build GRCh38
rs913734127 Chr.19: 40944718-40944718 on Build GRCh38
rs964998396 Chr.19: 40944697-40944697 on Build GRCh38
rs761277149 Chr.19: 40944808-40944808 on Build GRCh38
rs190966934 Chr.19: 40944383-40944383 on Build GRCh38
rs754869717 Chr.19: 40944394-40944394 on Build GRCh38
rs749417066 Chr.19: 40944497-40944497 on Build GRCh38
rs143613710 Chr.19: 40944446-40944446 on Build GRCh38
rs571721690 Chr.19: 40944534-40944534 on Build GRCh38
rs3745220 Chr.19: 40944704-40944704 on Build GRCh38
rs769979791 Chr.19: 40944350-40944350 on Build GRCh38
rs754600882 Chr.19: 40944481-40944481 on Build GRCh38
rs753143304 Chr.19: 40944372-40944372 on Build GRCh38
rs912259852 Chr.19: 40944693-40944693 on Build GRCh38
rs748048377 Chr.19: 40944390-40944390 on Build GRCh38
rs182789621 Chr.19: 40944395-40944395 on Build GRCh38
rs34102690 Chr.19: 40944618-40944619 on Build GRCh38
rs776954450 Chr.19: 40944537-40944537 on Build GRCh38
rs192201716 Chr.19: 40944732-40944732 on Build GRCh38
rs1002987523 Chr.19: 40944561-40944561 on Build GRCh38
rs567948411 Chr.19: 40944373-40944373 on Build GRCh38
rs769202293 Chr.19: 40944531-40944531 on Build GRCh38
rs758078603 Chr.19: 40944381-40944381 on Build GRCh38
rs779255614 Chr.19: 40944501-40944501 on Build GRCh38
rs147163321 Chr.19: 40944788-40944788 on Build GRCh38
rs200776726 Chr.19: 40944810-40944810 on Build GRCh38
rs752497013 Chr.19: 40944491-40944491 on Build GRCh38
rs753816378 Chr.19: 40944550-40944550 on Build GRCh38
rs975085078 Chr.19: 40944701-40944701 on Build GRCh38
rs774408378 Chr.19: 40944401-40944401 on Build GRCh38
rs760014633 Chr.19: 40944403-40944403 on Build GRCh38
rs766111685 Chr.19: 40944483-40944483 on Build GRCh38
rs907192863 Chr.19: 40944558-40944558 on Build GRCh38
rs554559574 Chr.19: 40944696-40944696 on Build GRCh38
rs752206276 Chr.19: 40944518-40944518 on Build GRCh38
rs536498979 Chr.19: 40944681-40944681 on Build GRCh38
rs921027145 Chr.19: 40944713-40944713 on Build GRCh38
rs951632279 Chr.19: 40944631-40944631 on Build GRCh38
rs182964907 Chr.19: 40944794-40944794 on Build GRCh38
rs143636662 Chr.19: 40944776-40944776 on Build GRCh38
rs778026724 Chr.19: 40944393-40944393 on Build GRCh38
rs1027145064 Chr.19: 40944795-40944795 on Build GRCh38
rs148021275 Chr.19: 40944529-40944529 on Build GRCh38
rs770226036 Chr.19: 40944563-40944563 on Build GRCh38
rs958791841 Chr.19: 40944578-40944578 on Build GRCh38
rs982989479 Chr.19: 40944638-40944638 on Build GRCh38
rs777827609 Chr.19: 40944494-40944494 on Build GRCh38
rs777968954 Chr.19: 40944398-40944398 on Build GRCh38
rs767144134 Chr.19: 40944539-40944539 on Build GRCh38
rs746256306 Chr.19: 40944543-40944543 on Build GRCh38
rs766121242 Chr.19: 40944463-40944463 on Build GRCh38
rs1003301679 Chr.19: 40944777-40944777 on Build GRCh38
rs746096195 Chr.19: 40944509-40944509 on Build GRCh38
rs777092655 Chr.19: 40944532-40944532 on Build GRCh38
rs764492315 Chr.19: 40944734-40944734 on Build GRCh38
rs17411397 Chr.19: 40944748-40944748 on Build GRCh38
rs775870595 Chr.19: 40944418-40944418 on Build GRCh38
rs553974834 Chr.19: 40944650-40944650 on Build GRCh38
rs764197001 Chr.19: 40944545-40944545 on Build GRCh38
rs199647446 Chr.19: 40944792-40944792 on Build GRCh38
rs772273868 Chr.19: 40944527-40944527 on Build GRCh38
rs1054032134 Chr.19: 40944716-40944716 on Build GRCh38
rs576296027 Chr.19: 40944698-40944698 on Build GRCh38
rs761176725 Chr.19: 40944426-40944426 on Build GRCh38
rs763582319 Chr.19: 40944715-40944715 on Build GRCh38
rs572252204 Chr.19: 40944672-40944672 on Build GRCh38
rs749869160 Chr.19: 40944374-40944374 on Build GRCh38
rs780852202 Chr.19: 40944486-40944486 on Build GRCh38
rs763268186 Chr.19: 40944366-40944366 on Build GRCh38
rs543447254 Chr.19: 40944370-40944370 on Build GRCh38
rs561265893 Chr.19: 40944358-40944358 on Build GRCh38
rs1034828860 Chr.19: 40944574-40944574 on Build GRCh38
rs567993541 Chr.19: 40944474-40944474 on Build GRCh38
rs138769314 Chr.19: 40944826-40944826 on Build GRCh38
rs534746119 Chr.19: 40944427-40944427 on Build GRCh38
rs756138130 Chr.19: 40944493-40944493 on Build GRCh38
rs73557121 Chr.19: 40944498-40944498 on Build GRCh38
rs752586870 Chr.19: 40944542-40944542 on Build GRCh38
rs769783909 Chr.19: 40944391-40944391 on Build GRCh38
rs762506025 Chr.19: 40944533-40944533 on Build GRCh38
rs559048361 Chr.19: 40944738-40944738 on Build GRCh38
rs561378663 Chr.19: 40944780-40944780 on Build GRCh38
rs751013384 Chr.19: 40944536-40944536 on Build GRCh38
rs186096524 Chr.19: 40944382-40944382 on Build GRCh38
rs576497980 Chr.19: 40944729-40944729 on Build GRCh38
rs114966387 Chr.19: 40944367-40944367 on Build GRCh38
rs748327172 Chr.19: 40944347-40944347 on Build GRCh38
rs539157850 Chr.19: 40944649-40944649 on Build GRCh38
rs187353148 Chr.19: 40944728-40944728 on Build GRCh38
rs772314406 Chr.19: 40944408-40944408 on Build GRCh38
rs974657353 Chr.19: 40944587-40944587 on Build GRCh38
rs564921013 Chr.19: 40944705-40944705 on Build GRCh38
rs1020335878 Chr.19: 40944552-40944552 on Build GRCh38
rs549388317 Chr.19: 40944459-40944459 on Build GRCh38
rs112899021 Chr.19: 40944668-40944668 on Build GRCh38
rs1027075036 Chr.19: 40944627-40944628 on Build GRCh38

View Full Product Details