Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 113601109-113601662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX11L2
Entrez Gene ID
84771
Gene Name
DEAD/H-box helicase 11 like 2
Gene Aliases
-
UniGene
Hs.712940
Gene Chromosome Location
Chr.2: 113599028-113603717 on Build GRCh38
Transcripts
NR_024004.1 NR_024005.2

SNP Detail

SNP ID Chromosome Location
rs75916570 Chr.2: 113601249-113601249 on Build GRCh38
rs57442816 Chr.2: 113601518-113601518 on Build GRCh38
rs571228818 Chr.2: 113601297-113601297 on Build GRCh38
rs796336656 Chr.2: 113601390-113601390 on Build GRCh38
rs551360495 Chr.2: 113601231-113601231 on Build GRCh38
rs796822288 Chr.2: 113601318-113601318 on Build GRCh38
rs573531221 Chr.2: 113601463-113601463 on Build GRCh38
rs553526099 Chr.2: 113601433-113601433 on Build GRCh38
rs753170027 Chr.2: 113601188-113601188 on Build GRCh38
rs11897134 Chr.2: 113601195-113601195 on Build GRCh38
rs182113183 Chr.2: 113601171-113601171 on Build GRCh38
rs905206793 Chr.2: 113601133-113601133 on Build GRCh38
rs11510383 Chr.2: 113601571-113601571 on Build GRCh38
rs549745279 Chr.2: 113601161-113601161 on Build GRCh38
rs961978768 Chr.2: 113601168-113601168 on Build GRCh38
rs2475482 Chr.2: 113601174-113601174 on Build GRCh38
rs2462500 Chr.2: 113601476-113601476 on Build GRCh38
rs7357973 Chr.2: 113601469-113601469 on Build GRCh38
rs564528259 Chr.2: 113601631-113601631 on Build GRCh38
rs544248522 Chr.2: 113601593-113601593 on Build GRCh38
rs371893258 Chr.2: 113601423-113601423 on Build GRCh38
rs529589289 Chr.2: 113601139-113601139 on Build GRCh38
rs796593668 Chr.2: 113601290-113601290 on Build GRCh38
rs7340201 Chr.2: 113601211-113601211 on Build GRCh38
rs74400200 Chr.2: 113601270-113601270 on Build GRCh38
rs79461972 Chr.2: 113601143-113601143 on Build GRCh38
rs1035518209 Chr.2: 113601140-113601140 on Build GRCh38
rs76953915 Chr.2: 113601198-113601198 on Build GRCh38
rs759932052 Chr.2: 113601182-113601182 on Build GRCh38
rs796137017 Chr.2: 113601280-113601280 on Build GRCh38
rs555995626 Chr.2: 113601483-113601483 on Build GRCh38
rs750017533 Chr.2: 113601286-113601286 on Build GRCh38
rs7349310 Chr.2: 113601636-113601636 on Build GRCh38
rs4849250 Chr.2: 113601493-113601493 on Build GRCh38
rs76844454 Chr.2: 113601479-113601479 on Build GRCh38
rs762933768 Chr.2: 113601164-113601164 on Build GRCh38
rs62162365 Chr.2: 113601365-113601365 on Build GRCh38
rs7564627 Chr.2: 113601492-113601492 on Build GRCh38
rs536140383 Chr.2: 113601465-113601465 on Build GRCh38
rs7340200 Chr.2: 113601147-113601147 on Build GRCh38
rs911648675 Chr.2: 113601548-113601548 on Build GRCh38
rs1014846925 Chr.2: 113601163-113601163 on Build GRCh38
rs959827510 Chr.2: 113601144-113601144 on Build GRCh38
rs76129579 Chr.2: 113601238-113601238 on Build GRCh38
rs766642165 Chr.2: 113601167-113601167 on Build GRCh38
rs992694693 Chr.2: 113601160-113601160 on Build GRCh38
rs767830772 Chr.2: 113601183-113601183 on Build GRCh38

View Full Product Details