Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232386392-232386938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECEL1P2
Entrez Gene ID
347694
Gene Name
endothelin converting enzyme like 1 pseudogene 2
Gene Aliases
ECEL2
UniGene
Hs.452575
Gene Chromosome Location
Chr.2: 232385750-232387044 on Build GRCh38
Transcripts
NR_028501.1

SNP Detail

SNP ID Chromosome Location
rs988392590 Chr.2: 232386697-232386697 on Build GRCh38
rs979472736 Chr.2: 232386669-232386669 on Build GRCh38
rs560929476 Chr.2: 232386478-232386478 on Build GRCh38
rs569542416 Chr.2: 232386621-232386621 on Build GRCh38
rs547066569 Chr.2: 232386522-232386522 on Build GRCh38
rs967747762 Chr.2: 232386800-232386800 on Build GRCh38
rs917993447 Chr.2: 232386690-232386690 on Build GRCh38
rs988806330 Chr.2: 232386877-232386877 on Build GRCh38
rs540277681 Chr.2: 232386862-232386862 on Build GRCh38
rs1018031793 Chr.2: 232386658-232386658 on Build GRCh38
rs575186577 Chr.2: 232386434-232386434 on Build GRCh38
rs552817578 Chr.2: 232386604-232386604 on Build GRCh38
rs575132788 Chr.2: 232386827-232386827 on Build GRCh38
rs1008390602 Chr.2: 232386759-232386759 on Build GRCh38
rs116068897 Chr.2: 232386799-232386799 on Build GRCh38
rs544089902 Chr.2: 232386846-232386846 on Build GRCh38
rs538515449 Chr.2: 232386648-232386648 on Build GRCh38
rs923736665 Chr.2: 232386667-232386667 on Build GRCh38
rs992434900 Chr.2: 232386689-232386689 on Build GRCh38
rs918114551 Chr.2: 232386709-232386709 on Build GRCh38
rs530079531 Chr.2: 232386486-232386486 on Build GRCh38
rs1018644142 Chr.2: 232386588-232386588 on Build GRCh38
rs12328730 Chr.2: 232386656-232386656 on Build GRCh38
rs896611263 Chr.2: 232386512-232386512 on Build GRCh38
rs1045062546 Chr.2: 232386707-232386707 on Build GRCh38
rs377179904 Chr.2: 232386829-232386829 on Build GRCh38
rs898731795 Chr.2: 232386527-232386527 on Build GRCh38
rs1047542442 Chr.2: 232386879-232386879 on Build GRCh38
rs375232491 Chr.2: 232386858-232386858 on Build GRCh38
rs774201600 Chr.2: 232386819-232386819 on Build GRCh38
rs1005003364 Chr.2: 232386579-232386579 on Build GRCh38
rs1006176397 Chr.2: 232386536-232386536 on Build GRCh38
rs531441161 Chr.2: 232386897-232386897 on Build GRCh38
rs532696308 Chr.2: 232386578-232386578 on Build GRCh38
rs777626688 Chr.2: 232386644-232386644 on Build GRCh38
rs931520597 Chr.2: 232386731-232386731 on Build GRCh38
rs994030698 Chr.2: 232386570-232386570 on Build GRCh38
rs111285215 Chr.2: 232386849-232386849 on Build GRCh38
rs986579720 Chr.2: 232386657-232386657 on Build GRCh38
rs901584780 Chr.2: 232386774-232386774 on Build GRCh38
rs568908765 Chr.2: 232386789-232386789 on Build GRCh38
rs73001992 Chr.2: 232386824-232386824 on Build GRCh38
rs544129597 Chr.2: 232386453-232386453 on Build GRCh38
rs1047310407 Chr.2: 232386751-232386751 on Build GRCh38
rs113639360 Chr.2: 232386729-232386729 on Build GRCh38
rs926671694 Chr.2: 232386773-232386773 on Build GRCh38
rs948140667 Chr.2: 232386703-232386703 on Build GRCh38
rs979656391 Chr.2: 232386445-232386445 on Build GRCh38
rs1026205765 Chr.2: 232386540-232386540 on Build GRCh38
rs887398689 Chr.2: 232386754-232386754 on Build GRCh38
rs114158582 Chr.2: 232386664-232386664 on Build GRCh38
rs1030909661 Chr.2: 232386823-232386823 on Build GRCh38
rs995783129 Chr.2: 232386623-232386623 on Build GRCh38
rs1045166378 Chr.2: 232386761-232386761 on Build GRCh38
rs538568033 Chr.2: 232386701-232386701 on Build GRCh38
rs768180762 Chr.2: 232386816-232386816 on Build GRCh38
rs535875861 Chr.2: 232386675-232386675 on Build GRCh38
rs931325327 Chr.2: 232386878-232386878 on Build GRCh38
rs1028637680 Chr.2: 232386652-232386652 on Build GRCh38
rs530060030 Chr.2: 232386763-232386763 on Build GRCh38
rs769246994 Chr.2: 232386708-232386708 on Build GRCh38
rs560385239 Chr.2: 232386525-232386525 on Build GRCh38
rs1032446827 Chr.2: 232386850-232386850 on Build GRCh38
rs771919105 Chr.2: 232386826-232386826 on Build GRCh38
rs1045115044 Chr.2: 232386523-232386523 on Build GRCh38
rs1018043150 Chr.2: 232386835-232386836 on Build GRCh38
rs75195825 Chr.2: 232386831-232386831 on Build GRCh38
rs577110028 Chr.2: 232386795-232386795 on Build GRCh38
rs969431025 Chr.2: 232386857-232386857 on Build GRCh38
rs908782274 Chr.2: 232386915-232386915 on Build GRCh38
rs995793111 Chr.2: 232386539-232386539 on Build GRCh38

View Full Product Details