Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33193205-33193755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFA4P
Entrez Gene ID
317716
Gene Name
BPI fold containing family A member 4, pseudogene
Gene Aliases
BASE
UniGene
Hs.434194
Gene Chromosome Location
Chr.20: 33193605-33210462 on Build GRCh38
Transcripts
NR_026760.1

SNP Detail

SNP ID Chromosome Location
rs949843086 Chr.20: 33193688-33193688 on Build GRCh38
rs114911296 Chr.20: 33193706-33193706 on Build GRCh38
rs887413022 Chr.20: 33193574-33193574 on Build GRCh38
rs1046564386 Chr.20: 33193624-33193624 on Build GRCh38
rs141678644 Chr.20: 33193300-33193300 on Build GRCh38
rs920478269 Chr.20: 33193312-33193312 on Build GRCh38
rs931640366 Chr.20: 33193359-33193359 on Build GRCh38
rs573068394 Chr.20: 33193426-33193426 on Build GRCh38
rs980855370 Chr.20: 33193471-33193471 on Build GRCh38
rs562745256 Chr.20: 33193381-33193381 on Build GRCh38
rs913171168 Chr.20: 33193527-33193527 on Build GRCh38
rs983354792 Chr.20: 33193669-33193669 on Build GRCh38
rs975974890 Chr.20: 33193548-33193548 on Build GRCh38
rs892001736 Chr.20: 33193440-33193440 on Build GRCh38
rs575879912 Chr.20: 33193227-33193227 on Build GRCh38
rs745933154 Chr.20: 33193363-33193363 on Build GRCh38
rs941732286 Chr.20: 33193575-33193575 on Build GRCh38
rs925283302 Chr.20: 33193239-33193239 on Build GRCh38
rs1032321663 Chr.20: 33193472-33193472 on Build GRCh38
rs757805517 Chr.20: 33193277-33193277 on Build GRCh38
rs927339045 Chr.20: 33193723-33193723 on Build GRCh38
rs964712747 Chr.20: 33193296-33193296 on Build GRCh38
rs945930966 Chr.20: 33193303-33193303 on Build GRCh38
rs138882666 Chr.20: 33193588-33193588 on Build GRCh38
rs990459974 Chr.20: 33193513-33193513 on Build GRCh38
rs1000578756 Chr.20: 33193419-33193419 on Build GRCh38
rs1050149986 Chr.20: 33193573-33193573 on Build GRCh38
rs1010440117 Chr.20: 33193465-33193465 on Build GRCh38
rs1032457395 Chr.20: 33193712-33193712 on Build GRCh38
rs917196736 Chr.20: 33193681-33193681 on Build GRCh38
rs6057750 Chr.20: 33193431-33193431 on Build GRCh38
rs746858102 Chr.20: 33193658-33193658 on Build GRCh38
rs1014263595 Chr.20: 33193620-33193620 on Build GRCh38
rs987172470 Chr.20: 33193288-33193288 on Build GRCh38
rs772963833 Chr.20: 33193618-33193618 on Build GRCh38
rs1050015789 Chr.20: 33193375-33193375 on Build GRCh38
rs1002261859 Chr.20: 33193710-33193710 on Build GRCh38
rs1006859128 Chr.20: 33193405-33193405 on Build GRCh38
rs756154739 Chr.20: 33193374-33193374 on Build GRCh38
rs1025057151 Chr.20: 33193438-33193438 on Build GRCh38
rs377385196 Chr.20: 33193295-33193295 on Build GRCh38
rs558354125 Chr.20: 33193366-33193366 on Build GRCh38
rs552152581 Chr.20: 33193705-33193705 on Build GRCh38
rs945868055 Chr.20: 33193528-33193528 on Build GRCh38
rs950470217 Chr.20: 33193621-33193621 on Build GRCh38
rs866184728 Chr.20: 33193461-33193461 on Build GRCh38
rs529681598 Chr.20: 33193452-33193452 on Build GRCh38
rs1040508723 Chr.20: 33193335-33193335 on Build GRCh38
rs901640389 Chr.20: 33193346-33193346 on Build GRCh38
rs6057749 Chr.20: 33193313-33193313 on Build GRCh38
rs541792520 Chr.20: 33193454-33193454 on Build GRCh38
rs540729814 Chr.20: 33193427-33193427 on Build GRCh38
rs1044482717 Chr.20: 33193394-33193394 on Build GRCh38
rs150120882 Chr.20: 33193429-33193429 on Build GRCh38
rs563460342 Chr.20: 33193562-33193562 on Build GRCh38
rs958077654 Chr.20: 33193482-33193482 on Build GRCh38
rs1015186507 Chr.20: 33193420-33193420 on Build GRCh38
rs935865131 Chr.20: 33193270-33193270 on Build GRCh38
rs931692507 Chr.20: 33193567-33193567 on Build GRCh38
rs923115074 Chr.20: 33193550-33193550 on Build GRCh38

View Full Product Details