Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35559058-35559612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FER1L4
Entrez Gene ID
80307
Gene Name
fer-1 like family member 4, pseudogene
Gene Aliases
C20orf124
UniGene
Hs.72222
Gene Chromosome Location
Chr.20: 35558737-35607562 on Build GRCh38
Transcripts
NR_119376.1

SNP Detail

SNP ID Chromosome Location
rs966135288 Chr.20: 35559566-35559566 on Build GRCh38
rs773003418 Chr.20: 35559091-35559091 on Build GRCh38
rs930571052 Chr.20: 35559417-35559417 on Build GRCh38
rs551469371 Chr.20: 35559542-35559542 on Build GRCh38
rs766560287 Chr.20: 35559581-35559581 on Build GRCh38
rs573580346 Chr.20: 35559257-35559257 on Build GRCh38
rs556005228 Chr.20: 35559586-35559586 on Build GRCh38
rs750593084 Chr.20: 35559432-35559432 on Build GRCh38
rs527558371 Chr.20: 35559080-35559080 on Build GRCh38
rs766648281 Chr.20: 35559440-35559440 on Build GRCh38
rs951859878 Chr.20: 35559587-35559587 on Build GRCh38
rs535525255 Chr.20: 35559118-35559118 on Build GRCh38
rs376003194 Chr.20: 35559188-35559188 on Build GRCh38
rs575943265 Chr.20: 35559371-35559371 on Build GRCh38
rs1022520319 Chr.20: 35559518-35559518 on Build GRCh38
rs745439488 Chr.20: 35559499-35559499 on Build GRCh38
rs879491194 Chr.20: 35559578-35559578 on Build GRCh38
rs41293068 Chr.20: 35559222-35559222 on Build GRCh38
rs533207276 Chr.20: 35559541-35559541 on Build GRCh38
rs999558578 Chr.20: 35559536-35559536 on Build GRCh38
rs972902290 Chr.20: 35559124-35559124 on Build GRCh38
rs755379239 Chr.20: 35559453-35559453 on Build GRCh38
rs764182865 Chr.20: 35559190-35559190 on Build GRCh38
rs549303528 Chr.20: 35559104-35559104 on Build GRCh38
rs773154675 Chr.20: 35559567-35559567 on Build GRCh38
rs778565811 Chr.20: 35559331-35559331 on Build GRCh38
rs751901531 Chr.20: 35559451-35559451 on Build GRCh38
rs758099643 Chr.20: 35559501-35559501 on Build GRCh38
rs1028787840 Chr.20: 35559584-35559584 on Build GRCh38
rs1011164422 Chr.20: 35559508-35559508 on Build GRCh38
rs267605907 Chr.20: 35559402-35559402 on Build GRCh38
rs757100820 Chr.20: 35559341-35559341 on Build GRCh38
rs560343901 Chr.20: 35559394-35559394 on Build GRCh38
rs762684462 Chr.20: 35559138-35559138 on Build GRCh38
rs551029910 Chr.20: 35559142-35559142 on Build GRCh38
rs561052848 Chr.20: 35559438-35559438 on Build GRCh38
rs114591805 Chr.20: 35559204-35559204 on Build GRCh38
rs760593532 Chr.20: 35559367-35559367 on Build GRCh38
rs927589758 Chr.20: 35559419-35559419 on Build GRCh38
rs550663748 Chr.20: 35559488-35559488 on Build GRCh38
rs773462042 Chr.20: 35559575-35559591 on Build GRCh38
rs112633156 Chr.20: 35559418-35559418 on Build GRCh38
rs556208225 Chr.20: 35559269-35559269 on Build GRCh38
rs534180892 Chr.20: 35559212-35559212 on Build GRCh38
rs534171317 Chr.20: 35559578-35559578 on Build GRCh38
rs780403202 Chr.20: 35559227-35559227 on Build GRCh38
rs771914495 Chr.20: 35559349-35559349 on Build GRCh38
rs906076709 Chr.20: 35559186-35559186 on Build GRCh38
rs748803717 Chr.20: 35559281-35559281 on Build GRCh38
rs912665696 Chr.20: 35559117-35559117 on Build GRCh38
rs755466992 Chr.20: 35559248-35559248 on Build GRCh38
rs765246461 Chr.20: 35559421-35559421 on Build GRCh38
rs747784081 Chr.20: 35559419-35559421 on Build GRCh38
rs748040339 Chr.20: 35559552-35559552 on Build GRCh38
rs931491388 Chr.20: 35559393-35559393 on Build GRCh38
rs965351104 Chr.20: 35559577-35559577 on Build GRCh38
rs1042166785 Chr.20: 35559494-35559494 on Build GRCh38
rs966510065 Chr.20: 35559392-35559392 on Build GRCh38
rs770382748 Chr.20: 35559295-35559295 on Build GRCh38
rs866543799 Chr.20: 35559294-35559294 on Build GRCh38
rs753153812 Chr.20: 35559461-35559461 on Build GRCh38
rs571892676 Chr.20: 35559559-35559559 on Build GRCh38
rs751989264 Chr.20: 35559238-35559238 on Build GRCh38
rs952775664 Chr.20: 35559585-35559585 on Build GRCh38
rs775499594 Chr.20: 35559154-35559154 on Build GRCh38
rs1054828315 Chr.20: 35559443-35559443 on Build GRCh38
rs543895189 Chr.20: 35559267-35559267 on Build GRCh38
rs566712777 Chr.20: 35559553-35559553 on Build GRCh38
rs765459203 Chr.20: 35559205-35559205 on Build GRCh38
rs117685422 Chr.20: 35559519-35559519 on Build GRCh38
rs115259459 Chr.20: 35559115-35559115 on Build GRCh38
rs1054007658 Chr.20: 35559524-35559524 on Build GRCh38
rs918633254 Chr.20: 35559381-35559381 on Build GRCh38
rs966694449 Chr.20: 35559274-35559274 on Build GRCh38
rs985498788 Chr.20: 35559301-35559301 on Build GRCh38
rs906707725 Chr.20: 35559195-35559195 on Build GRCh38
rs767975486 Chr.20: 35559454-35559454 on Build GRCh38
rs779890516 Chr.20: 35559511-35559511 on Build GRCh38
rs778451339 Chr.20: 35559400-35559402 on Build GRCh38
rs750082290 Chr.20: 35559377-35559377 on Build GRCh38
rs768594439 Chr.20: 35559520-35559520 on Build GRCh38
rs937707491 Chr.20: 35559187-35559187 on Build GRCh38
rs78913502 Chr.20: 35559366-35559366 on Build GRCh38
rs778279170 Chr.20: 35559489-35559489 on Build GRCh38
rs78897011 Chr.20: 35559180-35559180 on Build GRCh38
rs181197622 Chr.20: 35559150-35559150 on Build GRCh38
rs186195106 Chr.20: 35559192-35559192 on Build GRCh38
rs529270135 Chr.20: 35559460-35559460 on Build GRCh38

View Full Product Details