Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35576269-35576829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FER1L4
Entrez Gene ID
80307
Gene Name
fer-1 like family member 4, pseudogene
Gene Aliases
C20orf124
UniGene
Hs.72222
Gene Chromosome Location
Chr.20: 35558737-35607562 on Build GRCh38
Transcripts
NR_119376.1

SNP Detail

SNP ID Chromosome Location
rs762767335 Chr.20: 35576498-35576498 on Build GRCh38
rs563718315 Chr.20: 35576328-35576328 on Build GRCh38
rs767625829 Chr.20: 35576533-35576533 on Build GRCh38
rs940870873 Chr.20: 35576803-35576803 on Build GRCh38
rs746045579 Chr.20: 35576437-35576437 on Build GRCh38
rs770633883 Chr.20: 35576499-35576499 on Build GRCh38
rs781296539 Chr.20: 35576397-35576397 on Build GRCh38
rs575228374 Chr.20: 35576674-35576674 on Build GRCh38
rs1027893575 Chr.20: 35576789-35576789 on Build GRCh38
rs889178860 Chr.20: 35576626-35576626 on Build GRCh38
rs113987168 Chr.20: 35576372-35576372 on Build GRCh38
rs746643572 Chr.20: 35576533-35576533 on Build GRCh38
rs570952152 Chr.20: 35576448-35576448 on Build GRCh38
rs919616011 Chr.20: 35576734-35576734 on Build GRCh38
rs201622622 Chr.20: 35576643-35576643 on Build GRCh38
rs1029072292 Chr.20: 35576296-35576296 on Build GRCh38
rs1557203 Chr.20: 35576541-35576541 on Build GRCh38
rs779134039 Chr.20: 35576748-35576748 on Build GRCh38
rs1039735452 Chr.20: 35576318-35576318 on Build GRCh38
rs993944585 Chr.20: 35576768-35576768 on Build GRCh38
rs762160010 Chr.20: 35576739-35576739 on Build GRCh38
rs962071618 Chr.20: 35576762-35576762 on Build GRCh38
rs942348800 Chr.20: 35576294-35576294 on Build GRCh38
rs769768588 Chr.20: 35576495-35576495 on Build GRCh38
rs747695819 Chr.20: 35576463-35576463 on Build GRCh38
rs556503349 Chr.20: 35576585-35576585 on Build GRCh38
rs962578303 Chr.20: 35576403-35576403 on Build GRCh38
rs865824608 Chr.20: 35576466-35576466 on Build GRCh38
rs535764619 Chr.20: 35576542-35576542 on Build GRCh38
rs769820571 Chr.20: 35576291-35576293 on Build GRCh38
rs757597324 Chr.20: 35576336-35576336 on Build GRCh38
rs972350737 Chr.20: 35576412-35576412 on Build GRCh38
rs539159050 Chr.20: 35576766-35576766 on Build GRCh38
rs368726782 Chr.20: 35576465-35576465 on Build GRCh38
rs772625519 Chr.20: 35576438-35576438 on Build GRCh38
rs930977474 Chr.20: 35576747-35576747 on Build GRCh38
rs771964218 Chr.20: 35576616-35576616 on Build GRCh38
rs1005006409 Chr.20: 35576426-35576426 on Build GRCh38
rs540344947 Chr.20: 35576327-35576327 on Build GRCh38
rs754015059 Chr.20: 35576583-35576583 on Build GRCh38
rs34371610 Chr.20: 35576712-35576713 on Build GRCh38
rs184125702 Chr.20: 35576516-35576516 on Build GRCh38
rs777547004 Chr.20: 35576420-35576420 on Build GRCh38
rs1557202 Chr.20: 35576500-35576500 on Build GRCh38
rs1000071362 Chr.20: 35576291-35576291 on Build GRCh38
rs757989724 Chr.20: 35576296-35576297 on Build GRCh38
rs757605961 Chr.20: 35576360-35576360 on Build GRCh38
rs904475296 Chr.20: 35576548-35576548 on Build GRCh38
rs11698021 Chr.20: 35576487-35576487 on Build GRCh38
rs757585586 Chr.20: 35576667-35576667 on Build GRCh38
rs995633663 Chr.20: 35576392-35576392 on Build GRCh38
rs1008997401 Chr.20: 35576672-35576672 on Build GRCh38
rs192734778 Chr.20: 35576442-35576442 on Build GRCh38
rs1000015631 Chr.20: 35576566-35576566 on Build GRCh38
rs1019327961 Chr.20: 35576710-35576710 on Build GRCh38
rs34733285 Chr.20: 35576620-35576620 on Build GRCh38
rs912475088 Chr.20: 35576292-35576292 on Build GRCh38
rs1001545929 Chr.20: 35576439-35576439 on Build GRCh38
rs1024549023 Chr.20: 35576427-35576427 on Build GRCh38
rs971587993 Chr.20: 35576436-35576436 on Build GRCh38
rs930824114 Chr.20: 35576449-35576449 on Build GRCh38
rs1039382487 Chr.20: 35576707-35576707 on Build GRCh38
rs77898589 Chr.20: 35576325-35576325 on Build GRCh38
rs368251371 Chr.20: 35576620-35576620 on Build GRCh38
rs942586785 Chr.20: 35576632-35576632 on Build GRCh38
rs764338748 Chr.20: 35576577-35576577 on Build GRCh38
rs866551951 Chr.20: 35576661-35576661 on Build GRCh38
rs895493126 Chr.20: 35576517-35576517 on Build GRCh38
rs779411199 Chr.20: 35576362-35576362 on Build GRCh38
rs530894723 Chr.20: 35576425-35576425 on Build GRCh38

View Full Product Details