Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35583828-35584371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FER1L4
Entrez Gene ID
80307
Gene Name
fer-1 like family member 4, pseudogene
Gene Aliases
C20orf124
UniGene
Hs.72222
Gene Chromosome Location
Chr.20: 35558737-35607562 on Build GRCh38
Transcripts
NR_119376.1

SNP Detail

SNP ID Chromosome Location
rs6087722 Chr.20: 35583907-35583907 on Build GRCh38
rs146360306 Chr.20: 35584088-35584088 on Build GRCh38
rs759013375 Chr.20: 35584000-35584000 on Build GRCh38
rs1013109902 Chr.20: 35584342-35584342 on Build GRCh38
rs546800411 Chr.20: 35583972-35583972 on Build GRCh38
rs1048300162 Chr.20: 35584043-35584043 on Build GRCh38
rs888058888 Chr.20: 35583886-35583886 on Build GRCh38
rs878874206 Chr.20: 35584123-35584123 on Build GRCh38
rs896650897 Chr.20: 35584156-35584156 on Build GRCh38
rs1002757313 Chr.20: 35583900-35583900 on Build GRCh38
rs148219804 Chr.20: 35583964-35583964 on Build GRCh38
rs748651895 Chr.20: 35583954-35583954 on Build GRCh38
rs1003279311 Chr.20: 35583936-35583936 on Build GRCh38
rs775052939 Chr.20: 35584171-35584171 on Build GRCh38
rs1017972381 Chr.20: 35584274-35584274 on Build GRCh38
rs1033324258 Chr.20: 35583980-35583980 on Build GRCh38
rs957328849 Chr.20: 35583997-35583997 on Build GRCh38
rs767146169 Chr.20: 35584013-35584013 on Build GRCh38
rs191658727 Chr.20: 35583924-35583924 on Build GRCh38
rs998499104 Chr.20: 35584194-35584194 on Build GRCh38
rs1007480281 Chr.20: 35584269-35584269 on Build GRCh38
rs564200185 Chr.20: 35584261-35584261 on Build GRCh38
rs187018230 Chr.20: 35583911-35583911 on Build GRCh38
rs1021898682 Chr.20: 35584162-35584162 on Build GRCh38
rs968756590 Chr.20: 35583971-35583971 on Build GRCh38
rs765138059 Chr.20: 35584114-35584114 on Build GRCh38
rs544213242 Chr.20: 35584126-35584126 on Build GRCh38
rs142187655 Chr.20: 35584087-35584087 on Build GRCh38
rs763695241 Chr.20: 35584324-35584324 on Build GRCh38
rs967029547 Chr.20: 35583978-35583978 on Build GRCh38
rs1013055969 Chr.20: 35584142-35584142 on Build GRCh38
rs1024102738 Chr.20: 35583974-35583974 on Build GRCh38
rs373021272 Chr.20: 35584042-35584042 on Build GRCh38
rs924737567 Chr.20: 35583992-35583992 on Build GRCh38
rs949568305 Chr.20: 35584160-35584160 on Build GRCh38
rs1026850463 Chr.20: 35583866-35583866 on Build GRCh38
rs1032692826 Chr.20: 35584195-35584195 on Build GRCh38
rs894180329 Chr.20: 35583941-35583941 on Build GRCh38
rs568689903 Chr.20: 35583989-35583989 on Build GRCh38
rs6087721 Chr.20: 35583884-35583884 on Build GRCh38
rs575594304 Chr.20: 35584182-35584182 on Build GRCh38
rs76645991 Chr.20: 35584164-35584164 on Build GRCh38
rs56183473 Chr.20: 35584323-35584323 on Build GRCh38
rs529580064 Chr.20: 35583993-35583993 on Build GRCh38
rs138494285 Chr.20: 35584109-35584109 on Build GRCh38
rs568789906 Chr.20: 35584049-35584049 on Build GRCh38
rs965836556 Chr.20: 35584286-35584286 on Build GRCh38
rs768972661 Chr.20: 35583943-35583943 on Build GRCh38
rs909700027 Chr.20: 35584231-35584231 on Build GRCh38
rs77300313 Chr.20: 35584311-35584311 on Build GRCh38
rs550515400 Chr.20: 35584041-35584041 on Build GRCh38
rs774677658 Chr.20: 35583979-35583979 on Build GRCh38
rs1017446188 Chr.20: 35584073-35584073 on Build GRCh38
rs970863232 Chr.20: 35583927-35583927 on Build GRCh38
rs573003278 Chr.20: 35584296-35584296 on Build GRCh38
rs894869149 Chr.20: 35584332-35584332 on Build GRCh38
rs965888457 Chr.20: 35584085-35584085 on Build GRCh38
rs79411969 Chr.20: 35584115-35584115 on Build GRCh38
rs939964067 Chr.20: 35584263-35584263 on Build GRCh38
rs984824674 Chr.20: 35584110-35584110 on Build GRCh38
rs758441863 Chr.20: 35584121-35584121 on Build GRCh38
rs561941229 Chr.20: 35583959-35583959 on Build GRCh38
rs545881112 Chr.20: 35584220-35584220 on Build GRCh38
rs914788322 Chr.20: 35583968-35583968 on Build GRCh38
rs1036945042 Chr.20: 35584280-35584280 on Build GRCh38
rs985160318 Chr.20: 35584329-35584329 on Build GRCh38
rs368190043 Chr.20: 35584297-35584297 on Build GRCh38
rs762008482 Chr.20: 35584007-35584007 on Build GRCh38
rs374698882 Chr.20: 35583939-35583939 on Build GRCh38
rs183816413 Chr.20: 35584124-35584124 on Build GRCh38
rs1048184414 Chr.20: 35584203-35584203 on Build GRCh38
rs777218411 Chr.20: 35583949-35583949 on Build GRCh38
rs906231689 Chr.20: 35584282-35584282 on Build GRCh38
rs965397689 Chr.20: 35584188-35584188 on Build GRCh38
rs975911543 Chr.20: 35584315-35584315 on Build GRCh38
rs902961268 Chr.20: 35584165-35584165 on Build GRCh38
rs958854019 Chr.20: 35583945-35583945 on Build GRCh38
rs771010917 Chr.20: 35583868-35583868 on Build GRCh38
rs748175722 Chr.20: 35583864-35583864 on Build GRCh38
rs770281098 Chr.20: 35583897-35583897 on Build GRCh38

View Full Product Details