Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35595379-35595922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FER1L4
Entrez Gene ID
80307
Gene Name
fer-1 like family member 4, pseudogene
Gene Aliases
C20orf124
UniGene
Hs.72222
Gene Chromosome Location
Chr.20: 35558737-35607562 on Build GRCh38
Transcripts
NR_119376.1

SNP Detail

SNP ID Chromosome Location
rs1032004763 Chr.20: 35595675-35595675 on Build GRCh38
rs1057354800 Chr.20: 35595740-35595740 on Build GRCh38
rs377326393 Chr.20: 35595444-35595444 on Build GRCh38
rs773656885 Chr.20: 35595673-35595673 on Build GRCh38
rs967397990 Chr.20: 35595890-35595890 on Build GRCh38
rs912379644 Chr.20: 35595537-35595537 on Build GRCh38
rs547954784 Chr.20: 35595809-35595809 on Build GRCh38
rs1024432625 Chr.20: 35595832-35595832 on Build GRCh38
rs1034029774 Chr.20: 35595854-35595854 on Build GRCh38
rs558185067 Chr.20: 35595465-35595465 on Build GRCh38
rs966327658 Chr.20: 35595445-35595445 on Build GRCh38
rs916964114 Chr.20: 35595478-35595478 on Build GRCh38
rs867571768 Chr.20: 35595775-35595775 on Build GRCh38
rs985382557 Chr.20: 35595681-35595681 on Build GRCh38
rs1001245427 Chr.20: 35595848-35595848 on Build GRCh38
rs1004982049 Chr.20: 35595663-35595663 on Build GRCh38
rs983235518 Chr.20: 35595456-35595457 on Build GRCh38
rs553581090 Chr.20: 35595518-35595518 on Build GRCh38
rs937602830 Chr.20: 35595560-35595560 on Build GRCh38
rs754912021 Chr.20: 35595641-35595641 on Build GRCh38
rs965183299 Chr.20: 35595693-35595693 on Build GRCh38
rs78498266 Chr.20: 35595735-35595735 on Build GRCh38
rs994650721 Chr.20: 35595803-35595803 on Build GRCh38
rs543839451 Chr.20: 35595692-35595692 on Build GRCh38
rs895973215 Chr.20: 35595568-35595568 on Build GRCh38
rs370908712 Chr.20: 35595556-35595556 on Build GRCh38
rs778276884 Chr.20: 35595545-35595545 on Build GRCh38
rs57545942 Chr.20: 35595557-35595557 on Build GRCh38
rs984649252 Chr.20: 35595724-35595724 on Build GRCh38
rs531695009 Chr.20: 35595678-35595678 on Build GRCh38
rs541188636 Chr.20: 35595502-35595502 on Build GRCh38
rs566170082 Chr.20: 35595821-35595821 on Build GRCh38
rs1038919126 Chr.20: 35595610-35595610 on Build GRCh38
rs1023962223 Chr.20: 35595699-35595699 on Build GRCh38
rs886386047 Chr.20: 35595819-35595819 on Build GRCh38
rs561254747 Chr.20: 35595595-35595595 on Build GRCh38
rs186049494 Chr.20: 35595494-35595494 on Build GRCh38
rs191325539 Chr.20: 35595564-35595564 on Build GRCh38
rs931377285 Chr.20: 35595717-35595717 on Build GRCh38
rs1019964055 Chr.20: 35595688-35595688 on Build GRCh38
rs765098727 Chr.20: 35595657-35595657 on Build GRCh38
rs972562152 Chr.20: 35595715-35595715 on Build GRCh38
rs772596641 Chr.20: 35595639-35595639 on Build GRCh38
rs985821198 Chr.20: 35595535-35595535 on Build GRCh38
rs890558703 Chr.20: 35595570-35595570 on Build GRCh38
rs944790716 Chr.20: 35595764-35595764 on Build GRCh38
rs943882479 Chr.20: 35595543-35595543 on Build GRCh38
rs1039104566 Chr.20: 35595766-35595766 on Build GRCh38
rs889432103 Chr.20: 35595839-35595839 on Build GRCh38
rs926787847 Chr.20: 35595529-35595529 on Build GRCh38
rs902282181 Chr.20: 35595625-35595625 on Build GRCh38
rs758118855 Chr.20: 35595802-35595804 on Build GRCh38
rs959548491 Chr.20: 35595856-35595856 on Build GRCh38
rs868501137 Chr.20: 35595546-35595546 on Build GRCh38
rs536658676 Chr.20: 35595450-35595450 on Build GRCh38
rs6060497 Chr.20: 35595742-35595742 on Build GRCh38
rs1041863674 Chr.20: 35595759-35595759 on Build GRCh38
rs994764788 Chr.20: 35595644-35595644 on Build GRCh38
rs1020623545 Chr.20: 35595620-35595620 on Build GRCh38
rs927295603 Chr.20: 35595732-35595732 on Build GRCh38
rs542374395 Chr.20: 35595751-35595754 on Build GRCh38
rs1042810697 Chr.20: 35595758-35595758 on Build GRCh38
rs1000603660 Chr.20: 35595811-35595811 on Build GRCh38
rs959496441 Chr.20: 35595736-35595736 on Build GRCh38
rs982718827 Chr.20: 35595524-35595524 on Build GRCh38
rs918640952 Chr.20: 35595716-35595716 on Build GRCh38
rs1027545446 Chr.20: 35595645-35595645 on Build GRCh38
rs931261332 Chr.20: 35595548-35595548 on Build GRCh38
rs949812954 Chr.20: 35595496-35595496 on Build GRCh38
rs917504017 Chr.20: 35595741-35595741 on Build GRCh38
rs1001059321 Chr.20: 35595660-35595660 on Build GRCh38
rs989630275 Chr.20: 35595883-35595883 on Build GRCh38

View Full Product Details