Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 13979522-13980073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD20A11P
Entrez Gene ID
391267
Gene Name
ankyrin repeat domain 20 family member A11, pseudogene
Gene Aliases
C21orf81
UniGene
Hs.364456
Gene Chromosome Location
Chr.21: 13943769-13980444 on Build GRCh38
Transcripts
NR_027270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5930059 c.142G>A p.E48K Chr.21: 13979763-13979763 on Build GRCh38
4295057 c.201C>A p.H67Q Chr.21: 13979704-13979704 on Build GRCh38
3972710 c.152G>A p.R51H Chr.21: 13979753-13979753 on Build GRCh38
3911952 c.141G>A p.A47A Chr.21: 13979764-13979764 on Build GRCh38
3549768 c.83G>A p.R28Q Chr.21: 13979822-13979822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912501640 Chr.21: 13979636-13979636 on Build GRCh38
rs551153935 Chr.21: 13979892-13979892 on Build GRCh38
rs573587940 Chr.21: 13979732-13979732 on Build GRCh38
rs914070159 Chr.21: 13979566-13979566 on Build GRCh38
rs961075964 Chr.21: 13979946-13979946 on Build GRCh38
rs533622795 Chr.21: 13979930-13979930 on Build GRCh38
rs533682138 Chr.21: 13979676-13979676 on Build GRCh38
rs919854623 Chr.21: 13979988-13979988 on Build GRCh38
rs542621884 Chr.21: 13979735-13979735 on Build GRCh38
rs541565208 Chr.21: 13979834-13979834 on Build GRCh38
rs561320907 Chr.21: 13979842-13979842 on Build GRCh38
rs912209951 Chr.21: 13979814-13979814 on Build GRCh38
rs200848501 Chr.21: 13979766-13979766 on Build GRCh38
rs1019749211 Chr.21: 13979945-13979945 on Build GRCh38
rs765902077 Chr.21: 13979782-13979782 on Build GRCh38
rs967796256 Chr.21: 13979574-13979574 on Build GRCh38
rs570521806 Chr.21: 13979625-13979625 on Build GRCh38
rs919816963 Chr.21: 13979836-13979836 on Build GRCh38
rs553350168 Chr.21: 13979691-13979691 on Build GRCh38
rs527781990 Chr.21: 13979830-13979830 on Build GRCh38
rs972369637 Chr.21: 13979980-13979980 on Build GRCh38
rs374263046 Chr.21: 13979929-13979929 on Build GRCh38
rs113190775 Chr.21: 13979692-13979692 on Build GRCh38
rs565516246 Chr.21: 13979785-13979785 on Build GRCh38
rs905309656 Chr.21: 13979850-13979850 on Build GRCh38
rs530011527 Chr.21: 13979844-13979844 on Build GRCh38
rs111492197 Chr.21: 13979662-13979662 on Build GRCh38
rs56270809 Chr.21: 13979932-13979932 on Build GRCh38
rs550172910 Chr.21: 13979856-13979856 on Build GRCh38
rs898318042 Chr.21: 13979679-13979679 on Build GRCh38
rs762224005 Chr.21: 13979704-13979704 on Build GRCh38
rs115967022 Chr.21: 13979876-13979876 on Build GRCh38
rs188144288 Chr.21: 13979767-13979767 on Build GRCh38
rs927146401 Chr.21: 13980024-13980024 on Build GRCh38
rs896521872 Chr.21: 13980038-13980038 on Build GRCh38
rs765599960 Chr.21: 13979728-13979728 on Build GRCh38
rs2297245 Chr.21: 13979992-13979992 on Build GRCh38
rs939948649 Chr.21: 13979651-13979651 on Build GRCh38
rs111551723 Chr.21: 13979789-13979789 on Build GRCh38
rs556134351 Chr.21: 13980029-13980029 on Build GRCh38
rs200320902 Chr.21: 13979760-13979760 on Build GRCh38
rs537618191 Chr.21: 13979602-13979602 on Build GRCh38
rs111862163 Chr.21: 13979923-13979923 on Build GRCh38
rs28449584 Chr.21: 13979668-13979668 on Build GRCh38
rs1002290313 Chr.21: 13979855-13979855 on Build GRCh38
rs201655089 Chr.21: 13980042-13980042 on Build GRCh38
rs947617684 Chr.21: 13979970-13979970 on Build GRCh38
rs1050746960 Chr.21: 13979630-13979630 on Build GRCh38
rs112029013 Chr.21: 13979896-13979896 on Build GRCh38
rs1035020009 Chr.21: 13979705-13979705 on Build GRCh38
rs368650000 Chr.21: 13979685-13979685 on Build GRCh38
rs979139451 Chr.21: 13979581-13979581 on Build GRCh38
rs968234022 Chr.21: 13979886-13979886 on Build GRCh38
rs567374884 Chr.21: 13979989-13979989 on Build GRCh38
rs961289923 Chr.21: 13979716-13979716 on Build GRCh38
rs988749047 Chr.21: 13979563-13979563 on Build GRCh38
rs1049191780 Chr.21: 13979684-13979684 on Build GRCh38
rs182625366 Chr.21: 13979725-13979725 on Build GRCh38
rs960954562 Chr.21: 13979555-13979555 on Build GRCh38
rs545574858 Chr.21: 13979768-13979768 on Build GRCh38
rs920985545 Chr.21: 13979619-13979619 on Build GRCh38
rs577279099 Chr.21: 13979670-13979670 on Build GRCh38
rs938127714 Chr.21: 13980028-13980028 on Build GRCh38
rs780060290 Chr.21: 13980009-13980009 on Build GRCh38
rs1009496631 Chr.21: 13979867-13979867 on Build GRCh38
rs569013673 Chr.21: 13979858-13979858 on Build GRCh38
rs986561216 Chr.21: 13979936-13979936 on Build GRCh38
rs986908560 Chr.21: 13979792-13979792 on Build GRCh38
rs995310036 Chr.21: 13979682-13979682 on Build GRCh38

View Full Product Details