Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 14273701-14274260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC13
Entrez Gene ID
150000
Gene Name
ATP binding cassette subfamily C member 13 (pseudogene)
Gene Aliases
ABCC13P, C21orf73, PRED6
UniGene
Hs.366575
Gene Chromosome Location
Chr.21: 14273799-14301386 on Build GRCh38
Transcripts
NR_003087.1 NR_003088.1

SNP Detail

SNP ID Chromosome Location
rs1009654655 Chr.21: 14274215-14274215 on Build GRCh38
rs1020575473 Chr.21: 14274004-14274004 on Build GRCh38
rs367824022 Chr.21: 14273940-14273940 on Build GRCh38
rs944957316 Chr.21: 14273934-14273934 on Build GRCh38
rs145572737 Chr.21: 14274226-14274226 on Build GRCh38
rs567491970 Chr.21: 14273810-14273810 on Build GRCh38
rs1002019871 Chr.21: 14273784-14273784 on Build GRCh38
rs986277869 Chr.21: 14273913-14273913 on Build GRCh38
rs765162907 Chr.21: 14274194-14274194 on Build GRCh38
rs978640401 Chr.21: 14274025-14274025 on Build GRCh38
rs8129984 Chr.21: 14274076-14274076 on Build GRCh38
rs896279760 Chr.21: 14274050-14274050 on Build GRCh38
rs762994706 Chr.21: 14274124-14274125 on Build GRCh38
rs758794604 Chr.21: 14274055-14274055 on Build GRCh38
rs930648773 Chr.21: 14273964-14273964 on Build GRCh38
rs963429104 Chr.21: 14273728-14273728 on Build GRCh38
rs1034858623 Chr.21: 14274036-14274036 on Build GRCh38
rs762736964 Chr.21: 14274115-14274115 on Build GRCh38
rs376111403 Chr.21: 14273876-14273877 on Build GRCh38
rs528472236 Chr.21: 14273742-14273742 on Build GRCh38
rs769140029 Chr.21: 14274098-14274098 on Build GRCh38
rs750950062 Chr.21: 14274200-14274200 on Build GRCh38
rs752720597 Chr.21: 14273796-14273796 on Build GRCh38
rs372111399 Chr.21: 14273787-14273787 on Build GRCh38
rs780220566 Chr.21: 14274065-14274065 on Build GRCh38
rs575059505 Chr.21: 14274214-14274214 on Build GRCh38
rs932173192 Chr.21: 14273897-14273897 on Build GRCh38
rs2822520 Chr.21: 14273739-14273739 on Build GRCh38
rs897855720 Chr.21: 14273955-14273955 on Build GRCh38
rs1001474277 Chr.21: 14274159-14274159 on Build GRCh38
rs768290080 Chr.21: 14274079-14274079 on Build GRCh38
rs770773609 Chr.21: 14274116-14274116 on Build GRCh38
rs751547087 Chr.21: 14274056-14274056 on Build GRCh38
rs915247578 Chr.21: 14273770-14273779 on Build GRCh38
rs899139515 Chr.21: 14273886-14273886 on Build GRCh38
rs1055744004 Chr.21: 14274179-14274179 on Build GRCh38
rs28413131 Chr.21: 14274232-14274232 on Build GRCh38
rs942350373 Chr.21: 14273806-14273806 on Build GRCh38
rs72559431 Chr.21: 14273926-14273926 on Build GRCh38
rs879034893 Chr.21: 14274196-14274196 on Build GRCh38
rs922301738 Chr.21: 14274237-14274237 on Build GRCh38
rs761045853 Chr.21: 14274143-14274143 on Build GRCh38
rs764038734 Chr.21: 14274021-14274021 on Build GRCh38
rs780718143 Chr.21: 14274081-14274081 on Build GRCh38
rs186679631 Chr.21: 14273778-14273778 on Build GRCh38
rs972756131 Chr.21: 14274225-14274225 on Build GRCh38
rs773236927 Chr.21: 14274105-14274105 on Build GRCh38
rs978448289 Chr.21: 14273792-14273792 on Build GRCh38
rs960426626 Chr.21: 14273942-14273942 on Build GRCh38
rs1039433845 Chr.21: 14273809-14273809 on Build GRCh38
rs912032475 Chr.21: 14273921-14273921 on Build GRCh38
rs886677991 Chr.21: 14273871-14273871 on Build GRCh38
rs960920902 Chr.21: 14273808-14273808 on Build GRCh38
rs899435767 Chr.21: 14274152-14274152 on Build GRCh38
rs759458857 Chr.21: 14274141-14274141 on Build GRCh38
rs993081289 Chr.21: 14273770-14273770 on Build GRCh38
rs2236001 Chr.21: 14274084-14274084 on Build GRCh38
rs569851490 Chr.21: 14273904-14273904 on Build GRCh38
rs975057714 Chr.21: 14274235-14274235 on Build GRCh38
rs988041265 Chr.21: 14273833-14273833 on Build GRCh38
rs762014341 Chr.21: 14274193-14274193 on Build GRCh38
rs534650579 Chr.21: 14274144-14274144 on Build GRCh38
rs940878476 Chr.21: 14274125-14274125 on Build GRCh38
rs372196047 Chr.21: 14274138-14274139 on Build GRCh38
rs1000049924 Chr.21: 14274229-14274229 on Build GRCh38
rs900942725 Chr.21: 14273823-14273823 on Build GRCh38
rs922333567 Chr.21: 14274089-14274089 on Build GRCh38
rs933761273 Chr.21: 14274092-14274092 on Build GRCh38
rs892436766 Chr.21: 14274204-14274204 on Build GRCh38
rs1032901066 Chr.21: 14274230-14274230 on Build GRCh38
rs992655609 Chr.21: 14273983-14273983 on Build GRCh38
rs549735841 Chr.21: 14273876-14273876 on Build GRCh38
rs986475718 Chr.21: 14274158-14274158 on Build GRCh38
rs1020374503 Chr.21: 14274222-14274222 on Build GRCh38
rs1009936929 Chr.21: 14274069-14274069 on Build GRCh38
rs1005588644 Chr.21: 14273873-14273873 on Build GRCh38
rs1049198203 Chr.21: 14273984-14273984 on Build GRCh38
rs913133252 Chr.21: 14273777-14273777 on Build GRCh38
rs61742419 Chr.21: 14274118-14274118 on Build GRCh38
rs554594060 Chr.21: 14274205-14274205 on Build GRCh38
rs370142949 Chr.21: 14274168-14274168 on Build GRCh38
rs1000467481 Chr.21: 14274117-14274117 on Build GRCh38
rs980191546 Chr.21: 14273734-14273734 on Build GRCh38
rs1001506883 Chr.21: 14273892-14273892 on Build GRCh38
rs933475863 Chr.21: 14273838-14273838 on Build GRCh38
rs57228841 Chr.21: 14273869-14273869 on Build GRCh38

View Full Product Details