Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 14299618-14300216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC13
Entrez Gene ID
150000
Gene Name
ATP binding cassette subfamily C member 13 (pseudogene)
Gene Aliases
ABCC13P, C21orf73, PRED6
UniGene
Hs.366575
Gene Chromosome Location
Chr.21: 14273799-14301386 on Build GRCh38
Transcripts
NR_003087.1

SNP Detail

SNP ID Chromosome Location
rs771142366 Chr.21: 14299691-14299691 on Build GRCh38
rs987020918 Chr.21: 14300011-14300011 on Build GRCh38
rs745556138 Chr.21: 14299934-14299935 on Build GRCh38
rs1043070086 Chr.21: 14299719-14299719 on Build GRCh38
rs925952580 Chr.21: 14300159-14300159 on Build GRCh38
rs570940829 Chr.21: 14299984-14299984 on Build GRCh38
rs375658662 Chr.21: 14299952-14299952 on Build GRCh38
rs912512650 Chr.21: 14300044-14300044 on Build GRCh38
rs887021272 Chr.21: 14299891-14299891 on Build GRCh38
rs1036687327 Chr.21: 14299747-14299747 on Build GRCh38
rs779008357 Chr.21: 14299711-14299711 on Build GRCh38
rs905367827 Chr.21: 14299840-14299840 on Build GRCh38
rs535281916 Chr.21: 14299842-14299842 on Build GRCh38
rs534926706 Chr.21: 14300086-14300086 on Build GRCh38
rs750430680 Chr.21: 14300000-14300000 on Build GRCh38
rs1044186604 Chr.21: 14300161-14300161 on Build GRCh38
rs528774348 Chr.21: 14299782-14299782 on Build GRCh38
rs181883300 Chr.21: 14300008-14300008 on Build GRCh38
rs960156950 Chr.21: 14300048-14300048 on Build GRCh38
rs898136496 Chr.21: 14299778-14299778 on Build GRCh38
rs1016584506 Chr.21: 14299974-14299974 on Build GRCh38
rs779828352 Chr.21: 14300069-14300069 on Build GRCh38
rs1020923088 Chr.21: 14299909-14299909 on Build GRCh38
rs554520962 Chr.21: 14300157-14300157 on Build GRCh38
rs967609551 Chr.21: 14300120-14300120 on Build GRCh38
rs979194274 Chr.21: 14300142-14300142 on Build GRCh38
rs866657799 Chr.21: 14299978-14299978 on Build GRCh38
rs189164003 Chr.21: 14299985-14299985 on Build GRCh38
rs939691945 Chr.21: 14299723-14299723 on Build GRCh38
rs754735011 Chr.21: 14299667-14299667 on Build GRCh38
rs1039961327 Chr.21: 14299796-14299796 on Build GRCh38
rs542116163 Chr.21: 14299837-14299837 on Build GRCh38
rs555070975 Chr.21: 14299970-14299970 on Build GRCh38
rs749531236 Chr.21: 14299684-14299684 on Build GRCh38
rs780983429 Chr.21: 14299678-14299678 on Build GRCh38
rs572672931 Chr.21: 14299692-14299692 on Build GRCh38
rs1153312 Chr.21: 14300055-14300055 on Build GRCh38
rs1046923187 Chr.21: 14299853-14299853 on Build GRCh38
rs1153313 Chr.21: 14300189-14300189 on Build GRCh38
rs1039756050 Chr.21: 14300169-14300169 on Build GRCh38
rs751712623 Chr.21: 14299731-14299731 on Build GRCh38
rs942780767 Chr.21: 14299791-14299791 on Build GRCh38
rs942791278 Chr.21: 14300165-14300165 on Build GRCh38
rs181637322 Chr.21: 14299852-14299852 on Build GRCh38
rs868812907 Chr.21: 14300043-14300043 on Build GRCh38
rs995284486 Chr.21: 14299797-14299797 on Build GRCh38
rs184704139 Chr.21: 14299890-14299890 on Build GRCh38

View Full Product Details