Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23376849-23377429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES5AP1
Entrez Gene ID
649264
Gene Name
carboxylesterase 5A pseudogene 1
Gene Aliases
-
UniGene
Hs.600549
Gene Chromosome Location
Chr.22: 23359606-23382126 on Build GRCh38
Transcripts
NR_037839.1

SNP Detail

SNP ID Chromosome Location
rs1026162182 Chr.22: 23377371-23377371 on Build GRCh38
rs963199927 Chr.22: 23377044-23377044 on Build GRCh38
rs113986486 Chr.22: 23377084-23377084 on Build GRCh38
rs778881803 Chr.22: 23377098-23377098 on Build GRCh38
rs41277293 Chr.22: 23376905-23376905 on Build GRCh38
rs200259220 Chr.22: 23377180-23377181 on Build GRCh38
rs201570886 Chr.22: 23377183-23377183 on Build GRCh38
rs958745602 Chr.22: 23377136-23377136 on Build GRCh38
rs150684337 Chr.22: 23377023-23377023 on Build GRCh38
rs754484168 Chr.22: 23377040-23377040 on Build GRCh38
rs546522227 Chr.22: 23377125-23377125 on Build GRCh38
rs914690489 Chr.22: 23377165-23377165 on Build GRCh38
rs1026632221 Chr.22: 23377074-23377074 on Build GRCh38
rs574183951 Chr.22: 23376960-23376960 on Build GRCh38
rs574917448 Chr.22: 23377223-23377223 on Build GRCh38
rs781499477 Chr.22: 23377309-23377309 on Build GRCh38
rs937456949 Chr.22: 23376909-23376909 on Build GRCh38
rs376742361 Chr.22: 23377301-23377301 on Build GRCh38
rs369642842 Chr.22: 23377348-23377348 on Build GRCh38
rs746769930 Chr.22: 23377270-23377270 on Build GRCh38
rs9624138 Chr.22: 23376954-23376954 on Build GRCh38
rs557244407 Chr.22: 23377293-23377293 on Build GRCh38
rs764334719 Chr.22: 23376881-23376881 on Build GRCh38
rs991539817 Chr.22: 23377147-23377147 on Build GRCh38
rs544955720 Chr.22: 23377378-23377378 on Build GRCh38
rs1055867647 Chr.22: 23376913-23376913 on Build GRCh38
rs924844197 Chr.22: 23377184-23377184 on Build GRCh38
rs550896617 Chr.22: 23377061-23377061 on Build GRCh38
rs771293414 Chr.22: 23377288-23377288 on Build GRCh38
rs543719002 Chr.22: 23376896-23376896 on Build GRCh38
rs143539157 Chr.22: 23377257-23377257 on Build GRCh38
rs951457730 Chr.22: 23376880-23376880 on Build GRCh38
rs993318022 Chr.22: 23377357-23377357 on Build GRCh38
rs144803871 Chr.22: 23376972-23376972 on Build GRCh38
rs574295940 Chr.22: 23377304-23377304 on Build GRCh38
rs13053718 Chr.22: 23377016-23377016 on Build GRCh38
rs868638163 Chr.22: 23377117-23377117 on Build GRCh38
rs8138608 Chr.22: 23377019-23377019 on Build GRCh38
rs536436977 Chr.22: 23377087-23377087 on Build GRCh38
rs928699696 Chr.22: 23376894-23376894 on Build GRCh38
rs5759749 Chr.22: 23377183-23377183 on Build GRCh38
rs1051467862 Chr.22: 23377000-23377000 on Build GRCh38
rs1051843248 Chr.22: 23377218-23377218 on Build GRCh38
rs566537461 Chr.22: 23377168-23377168 on Build GRCh38
rs148554157 Chr.22: 23376994-23376994 on Build GRCh38
rs993371680 Chr.22: 23377067-23377067 on Build GRCh38
rs375061251 Chr.22: 23377072-23377072 on Build GRCh38
rs543203316 Chr.22: 23377315-23377315 on Build GRCh38
rs75477984 Chr.22: 23377181-23377181 on Build GRCh38
rs755781444 Chr.22: 23376993-23376993 on Build GRCh38
rs528625274 Chr.22: 23377004-23377004 on Build GRCh38
rs928673741 Chr.22: 23377118-23377118 on Build GRCh38
rs914747674 Chr.22: 23376915-23376915 on Build GRCh38
rs933599145 Chr.22: 23377196-23377196 on Build GRCh38
rs777262837 Chr.22: 23377299-23377299 on Build GRCh38

View Full Product Details