Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 25447960-25448501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYBB2P1
Entrez Gene ID
1416
Gene Name
crystallin beta B2 pseudogene 1
Gene Aliases
CRYB2B
UniGene
Hs.571835
Gene Chromosome Location
Chr.22: 25448087-25461678 on Build GRCh38
Transcripts
NR_033733.1 NR_033734.1

SNP Detail

SNP ID Chromosome Location
rs762131080 Chr.22: 25448039-25448039 on Build GRCh38
rs942397801 Chr.22: 25448424-25448424 on Build GRCh38
rs960891037 Chr.22: 25448119-25448119 on Build GRCh38
rs145588736 Chr.22: 25447983-25447983 on Build GRCh38
rs961287271 Chr.22: 25448230-25448230 on Build GRCh38
rs992758015 Chr.22: 25448126-25448126 on Build GRCh38
rs191205078 Chr.22: 25448188-25448188 on Build GRCh38
rs913146519 Chr.22: 25448399-25448399 on Build GRCh38
rs1003946357 Chr.22: 25448238-25448238 on Build GRCh38
rs745792688 Chr.22: 25448352-25448352 on Build GRCh38
rs142581347 Chr.22: 25448452-25448452 on Build GRCh38
rs941559836 Chr.22: 25448143-25448143 on Build GRCh38
rs898174347 Chr.22: 25448204-25448204 on Build GRCh38
rs1012549659 Chr.22: 25448322-25448322 on Build GRCh38
rs925258369 Chr.22: 25448340-25448340 on Build GRCh38
rs748107428 Chr.22: 25448329-25448331 on Build GRCh38
rs58348884 Chr.22: 25448090-25448098 on Build GRCh38
rs965077338 Chr.22: 25448390-25448390 on Build GRCh38
rs575326909 Chr.22: 25448361-25448361 on Build GRCh38
rs534715951 Chr.22: 25448467-25448467 on Build GRCh38
rs770930750 Chr.22: 25448177-25448177 on Build GRCh38
rs981258457 Chr.22: 25448170-25448170 on Build GRCh38
rs28647320 Chr.22: 25448032-25448032 on Build GRCh38
rs990209573 Chr.22: 25448373-25448373 on Build GRCh38
rs1006553366 Chr.22: 25448068-25448068 on Build GRCh38
rs904443303 Chr.22: 25448016-25448016 on Build GRCh38
rs916679090 Chr.22: 25447980-25447980 on Build GRCh38
rs535413549 Chr.22: 25448105-25448105 on Build GRCh38
rs1043748678 Chr.22: 25447994-25447994 on Build GRCh38
rs956692356 Chr.22: 25448342-25448342 on Build GRCh38
rs956145645 Chr.22: 25448345-25448345 on Build GRCh38
rs955511896 Chr.22: 25448044-25448044 on Build GRCh38
rs979507071 Chr.22: 25448338-25448338 on Build GRCh38
rs1029402965 Chr.22: 25448058-25448058 on Build GRCh38
rs1003916593 Chr.22: 25448131-25448131 on Build GRCh38
rs183931236 Chr.22: 25448255-25448255 on Build GRCh38
rs981946383 Chr.22: 25448052-25448052 on Build GRCh38
rs780952753 Chr.22: 25448381-25448381 on Build GRCh38
rs993860334 Chr.22: 25448212-25448212 on Build GRCh38
rs555134654 Chr.22: 25448336-25448336 on Build GRCh38
rs969407830 Chr.22: 25448330-25448330 on Build GRCh38
rs1003151609 Chr.22: 25448333-25448333 on Build GRCh38
rs7289954 Chr.22: 25448149-25448149 on Build GRCh38
rs948171346 Chr.22: 25448129-25448129 on Build GRCh38
rs915712049 Chr.22: 25448049-25448049 on Build GRCh38
rs544695759 Chr.22: 25448227-25448227 on Build GRCh38
rs988126237 Chr.22: 25448351-25448351 on Build GRCh38
rs1016616290 Chr.22: 25448087-25448087 on Build GRCh38
rs889557565 Chr.22: 25448067-25448067 on Build GRCh38
rs777562267 Chr.22: 25448195-25448195 on Build GRCh38
rs898176058 Chr.22: 25448294-25448294 on Build GRCh38
rs1026202254 Chr.22: 25448113-25448113 on Build GRCh38
rs28535815 Chr.22: 25448463-25448463 on Build GRCh38
rs962916413 Chr.22: 25448088-25448088 on Build GRCh38
rs3198642 Chr.22: 25448180-25448180 on Build GRCh38
rs187696200 Chr.22: 25448419-25448419 on Build GRCh38
rs5752151 Chr.22: 25448060-25448060 on Build GRCh38
rs756269167 Chr.22: 25448139-25448139 on Build GRCh38
rs891737005 Chr.22: 25448151-25448151 on Build GRCh38
rs1038017930 Chr.22: 25448193-25448193 on Build GRCh38
rs914626829 Chr.22: 25448387-25448387 on Build GRCh38
rs553385743 Chr.22: 25448092-25448092 on Build GRCh38
rs72228903 Chr.22: 25448061-25448069 on Build GRCh38
rs908060921 Chr.22: 25448479-25448479 on Build GRCh38
rs755761077 Chr.22: 25448036-25448036 on Build GRCh38
rs1047949212 Chr.22: 25448474-25448474 on Build GRCh38
rs538971634 Chr.22: 25448135-25448135 on Build GRCh38
rs150921948 Chr.22: 25448457-25448457 on Build GRCh38
rs891645370 Chr.22: 25448051-25448051 on Build GRCh38
rs575480897 Chr.22: 25448130-25448130 on Build GRCh38
rs935898205 Chr.22: 25448148-25448148 on Build GRCh38
rs148844196 Chr.22: 25448425-25448425 on Build GRCh38
rs1038111082 Chr.22: 25448444-25448444 on Build GRCh38
rs369867215 Chr.22: 25448158-25448158 on Build GRCh38
rs572904410 Chr.22: 25448157-25448157 on Build GRCh38
rs546680895 Chr.22: 25448061-25448061 on Build GRCh38
rs924737151 Chr.22: 25448176-25448176 on Build GRCh38
rs902056820 Chr.22: 25448198-25448198 on Build GRCh38
rs1025875909 Chr.22: 25448229-25448229 on Build GRCh38
rs1013748555 Chr.22: 25448247-25448247 on Build GRCh38
rs573716555 Chr.22: 25448186-25448186 on Build GRCh38
rs1054381670 Chr.22: 25448114-25448114 on Build GRCh38
rs540338859 Chr.22: 25448167-25448167 on Build GRCh38
rs557928115 Chr.22: 25448152-25448152 on Build GRCh38
rs758144227 Chr.22: 25448189-25448189 on Build GRCh38
rs7288843 Chr.22: 25448128-25448128 on Build GRCh38
rs933822598 Chr.22: 25448156-25448156 on Build GRCh38
rs567822771 Chr.22: 25448383-25448387 on Build GRCh38
rs576585 Chr.22: 25448054-25448054 on Build GRCh38
rs781306945 Chr.22: 25448069-25448070 on Build GRCh38
rs933908763 Chr.22: 25448411-25448411 on Build GRCh38
rs1050908299 Chr.22: 25448418-25448418 on Build GRCh38

View Full Product Details