Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10017356-10017900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIDECP
Entrez Gene ID
152302
Gene Name
cell death-inducing DFFA-like effector c pseudogene
Gene Aliases
CICE
UniGene
Hs.635072
Gene Chromosome Location
Chr.3: 10017553-10026136 on Build GRCh38
Transcripts
NR_002786.1

SNP Detail

SNP ID Chromosome Location
rs1004195242 Chr.3: 10017846-10017846 on Build GRCh38
rs561352193 Chr.3: 10017654-10017654 on Build GRCh38
rs146053841 Chr.3: 10017710-10017710 on Build GRCh38
rs568865291 Chr.3: 10017791-10017791 on Build GRCh38
rs112881461 Chr.3: 10017855-10017855 on Build GRCh38
rs572727527 Chr.3: 10017552-10017552 on Build GRCh38
rs1053237 Chr.3: 10017857-10017857 on Build GRCh38
rs11713831 Chr.3: 10017400-10017400 on Build GRCh38
rs776474993 Chr.3: 10017595-10017596 on Build GRCh38
rs571948553 Chr.3: 10017808-10017808 on Build GRCh38
rs746058297 Chr.3: 10017533-10017533 on Build GRCh38
rs1053236 Chr.3: 10017864-10017864 on Build GRCh38
rs539306782 Chr.3: 10017451-10017451 on Build GRCh38
rs568715483 Chr.3: 10017528-10017528 on Build GRCh38
rs1012896100 Chr.3: 10017494-10017494 on Build GRCh38
rs550979542 Chr.3: 10017768-10017768 on Build GRCh38
rs1053235 Chr.3: 10017871-10017871 on Build GRCh38
rs769809910 Chr.3: 10017548-10017548 on Build GRCh38
rs543134789 Chr.3: 10017633-10017633 on Build GRCh38
rs1041575659 Chr.3: 10017607-10017607 on Build GRCh38
rs995595144 Chr.3: 10017729-10017729 on Build GRCh38
rs536384379 Chr.3: 10017540-10017540 on Build GRCh38
rs1015652463 Chr.3: 10017712-10017712 on Build GRCh38
rs183213639 Chr.3: 10017845-10017845 on Build GRCh38
rs1012928452 Chr.3: 10017687-10017687 on Build GRCh38
rs935819127 Chr.3: 10017632-10017632 on Build GRCh38
rs907311634 Chr.3: 10017695-10017695 on Build GRCh38
rs1000502311 Chr.3: 10017410-10017410 on Build GRCh38
rs1053268 Chr.3: 10017727-10017727 on Build GRCh38
rs963023617 Chr.3: 10017547-10017547 on Build GRCh38
rs963224794 Chr.3: 10017719-10017719 on Build GRCh38
rs915862974 Chr.3: 10017809-10017809 on Build GRCh38
rs894355911 Chr.3: 10017635-10017635 on Build GRCh38
rs550562521 Chr.3: 10017522-10017522 on Build GRCh38
rs1053270 Chr.3: 10017713-10017713 on Build GRCh38
rs894491704 Chr.3: 10017472-10017472 on Build GRCh38
rs535858056 Chr.3: 10017793-10017793 on Build GRCh38
rs554349297 Chr.3: 10017541-10017541 on Build GRCh38
rs1053244 Chr.3: 10017817-10017817 on Build GRCh38
rs139591167 Chr.3: 10017418-10017418 on Build GRCh38
rs117587886 Chr.3: 10017559-10017559 on Build GRCh38
rs1053276 Chr.3: 10017629-10017629 on Build GRCh38
rs911676601 Chr.3: 10017560-10017560 on Build GRCh38
rs1028445982 Chr.3: 10017870-10017870 on Build GRCh38
rs1053278 Chr.3: 10017624-10017624 on Build GRCh38
rs1053258 Chr.3: 10017767-10017767 on Build GRCh38
rs1053286 Chr.3: 10017568-10017568 on Build GRCh38
rs73122383 Chr.3: 10017802-10017802 on Build GRCh38
rs144223814 Chr.3: 10017621-10017621 on Build GRCh38
rs181270938 Chr.3: 10017435-10017435 on Build GRCh38
rs1053272 Chr.3: 10017691-10017691 on Build GRCh38
rs1053269 Chr.3: 10017714-10017714 on Build GRCh38
rs977586914 Chr.3: 10017800-10017800 on Build GRCh38
rs8411 Chr.3: 10017752-10017752 on Build GRCh38
rs866307541 Chr.3: 10017439-10017439 on Build GRCh38
rs971435424 Chr.3: 10017534-10017534 on Build GRCh38
rs2478 Chr.3: 10017614-10017614 on Build GRCh38
rs921494531 Chr.3: 10017557-10017557 on Build GRCh38
rs756939650 Chr.3: 10017833-10017833 on Build GRCh38
rs1053285 Chr.3: 10017582-10017582 on Build GRCh38

View Full Product Details