Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10025731-10026275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIDECP
Entrez Gene ID
152302
Gene Name
cell death-inducing DFFA-like effector c pseudogene
Gene Aliases
CICE
UniGene
Hs.635072
Gene Chromosome Location
Chr.3: 10017553-10026136 on Build GRCh38
Transcripts
NR_002786.1

SNP Detail

SNP ID Chromosome Location
rs867848652 Chr.3: 10025904-10025904 on Build GRCh38
rs9845756 Chr.3: 10025864-10025864 on Build GRCh38
rs773539910 Chr.3: 10025939-10025939 on Build GRCh38
rs145413280 Chr.3: 10025880-10025880 on Build GRCh38
rs892551579 Chr.3: 10026252-10026252 on Build GRCh38
rs35835698 Chr.3: 10025886-10025887 on Build GRCh38
rs202096667 Chr.3: 10026008-10026008 on Build GRCh38
rs909818633 Chr.3: 10026167-10026167 on Build GRCh38
rs559622868 Chr.3: 10025926-10025926 on Build GRCh38
rs33917318 Chr.3: 10026113-10026113 on Build GRCh38
rs772162757 Chr.3: 10025894-10025894 on Build GRCh38
rs190136953 Chr.3: 10026216-10026216 on Build GRCh38
rs1039618919 Chr.3: 10026194-10026194 on Build GRCh38
rs942720718 Chr.3: 10025911-10025911 on Build GRCh38
rs929832296 Chr.3: 10026123-10026123 on Build GRCh38
rs760054667 Chr.3: 10026147-10026147 on Build GRCh38
rs185373575 Chr.3: 10026090-10026090 on Build GRCh38
rs983967077 Chr.3: 10026140-10026140 on Build GRCh38
rs200273431 Chr.3: 10026025-10026025 on Build GRCh38
rs969646383 Chr.3: 10026029-10026029 on Build GRCh38
rs990142974 Chr.3: 10025830-10025830 on Build GRCh38
rs867157513 Chr.3: 10026075-10026075 on Build GRCh38
rs554044201 Chr.3: 10026064-10026064 on Build GRCh38
rs868522503 Chr.3: 10026019-10026019 on Build GRCh38
rs56070953 Chr.3: 10025961-10025961 on Build GRCh38
rs2075310 Chr.3: 10025967-10025967 on Build GRCh38
rs775602382 Chr.3: 10025807-10025807 on Build GRCh38
rs979788044 Chr.3: 10025849-10025849 on Build GRCh38
rs956836999 Chr.3: 10025775-10025775 on Build GRCh38
rs543532368 Chr.3: 10025764-10025764 on Build GRCh38
rs372885040 Chr.3: 10026041-10026041 on Build GRCh38
rs915464757 Chr.3: 10025833-10025833 on Build GRCh38
rs533032218 Chr.3: 10025938-10025938 on Build GRCh38
rs918437009 Chr.3: 10025877-10025877 on Build GRCh38
rs901146843 Chr.3: 10025933-10025933 on Build GRCh38
rs189758149 Chr.3: 10025975-10025975 on Build GRCh38
rs570749988 Chr.3: 10025962-10025962 on Build GRCh38
rs768012206 Chr.3: 10025774-10025775 on Build GRCh38
rs75478634 Chr.3: 10025889-10025890 on Build GRCh38
rs758768583 Chr.3: 10026003-10026003 on Build GRCh38
rs901177887 Chr.3: 10026207-10026207 on Build GRCh38
rs942646849 Chr.3: 10026191-10026191 on Build GRCh38
rs968826374 Chr.3: 10025835-10025835 on Build GRCh38
rs933947176 Chr.3: 10026211-10026211 on Build GRCh38
rs998289824 Chr.3: 10025945-10025945 on Build GRCh38
rs35485751 Chr.3: 10025987-10025987 on Build GRCh38
rs1052558577 Chr.3: 10026222-10026222 on Build GRCh38
rs959673997 Chr.3: 10026096-10026096 on Build GRCh38
rs35993975 Chr.3: 10026110-10026110 on Build GRCh38
rs755066005 Chr.3: 10026131-10026131 on Build GRCh38
rs927066916 Chr.3: 10025862-10025862 on Build GRCh38
rs529087095 Chr.3: 10025876-10025876 on Build GRCh38

View Full Product Details