Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101523080-101523625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM172BP
Entrez Gene ID
131909
Gene Name
family with sequence similarity 172, member B pseudogene
Gene Aliases
FAM172B
UniGene
Hs.518125
Gene Chromosome Location
Chr.3: 101518867-101523887 on Build GRCh38
Transcripts
NR_036433.2

SNP Detail

SNP ID Chromosome Location
rs918502686 Chr.3: 101523443-101523443 on Build GRCh38
rs950132947 Chr.3: 101523488-101523488 on Build GRCh38
rs1052758112 Chr.3: 101523205-101523205 on Build GRCh38
rs564496723 Chr.3: 101523527-101523527 on Build GRCh38
rs141635805 Chr.3: 101523552-101523552 on Build GRCh38
rs370365645 Chr.3: 101523126-101523126 on Build GRCh38
rs534584267 Chr.3: 101523194-101523194 on Build GRCh38
rs935619773 Chr.3: 101523513-101523513 on Build GRCh38
rs781075756 Chr.3: 101523208-101523208 on Build GRCh38
rs574334644 Chr.3: 101523414-101523414 on Build GRCh38
rs1045954402 Chr.3: 101523273-101523273 on Build GRCh38
rs937157235 Chr.3: 101523597-101523597 on Build GRCh38
rs949858392 Chr.3: 101523260-101523260 on Build GRCh38
rs62280746 Chr.3: 101523498-101523498 on Build GRCh38
rs986881338 Chr.3: 101523430-101523430 on Build GRCh38
rs905670310 Chr.3: 101523279-101523279 on Build GRCh38
rs371976656 Chr.3: 101523257-101523258 on Build GRCh38
rs912918676 Chr.3: 101523525-101523525 on Build GRCh38
rs376524044 Chr.3: 101523456-101523456 on Build GRCh38
rs909692020 Chr.3: 101523392-101523392 on Build GRCh38
rs979572497 Chr.3: 101523495-101523495 on Build GRCh38
rs1001551106 Chr.3: 101523573-101523573 on Build GRCh38
rs988869961 Chr.3: 101523515-101523515 on Build GRCh38
rs540557566 Chr.3: 101523560-101523560 on Build GRCh38
rs149777902 Chr.3: 101523418-101523418 on Build GRCh38
rs570629089 Chr.3: 101523184-101523184 on Build GRCh38
rs979626625 Chr.3: 101523131-101523132 on Build GRCh38
rs556631419 Chr.3: 101523446-101523446 on Build GRCh38
rs1016824502 Chr.3: 101523300-101523300 on Build GRCh38
rs575033811 Chr.3: 101523447-101523447 on Build GRCh38
rs994163173 Chr.3: 101523333-101523333 on Build GRCh38
rs759820781 Chr.3: 101523367-101523367 on Build GRCh38
rs898455840 Chr.3: 101523328-101523328 on Build GRCh38
rs949770244 Chr.3: 101523534-101523534 on Build GRCh38
rs1054555924 Chr.3: 101523588-101523588 on Build GRCh38
rs552044646 Chr.3: 101523170-101523170 on Build GRCh38
rs553247276 Chr.3: 101523360-101523360 on Build GRCh38
rs775885715 Chr.3: 101523528-101523528 on Build GRCh38
rs953809931 Chr.3: 101523347-101523347 on Build GRCh38
rs966814424 Chr.3: 101523255-101523255 on Build GRCh38
rs1001380596 Chr.3: 101523299-101523299 on Build GRCh38
rs922370215 Chr.3: 101523304-101523304 on Build GRCh38
rs1033112793 Chr.3: 101523128-101523128 on Build GRCh38
rs1038245639 Chr.3: 101523603-101523603 on Build GRCh38
rs1002132262 Chr.3: 101523119-101523119 on Build GRCh38
rs985301934 Chr.3: 101523383-101523383 on Build GRCh38
rs925547003 Chr.3: 101523151-101523151 on Build GRCh38
rs976444906 Chr.3: 101523259-101523259 on Build GRCh38
rs1025674835 Chr.3: 101523371-101523371 on Build GRCh38

View Full Product Details