Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130228076-130228274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A4P2
Entrez Gene ID
646300
Gene Name
collagen type VI alpha 4 pseudogene 2
Gene Aliases
COL6A4
UniGene
Hs.170475
Gene Chromosome Location
Chr.3: 130212820-130273806 on Build GRCh38
Transcripts
NR_027898.1

SNP Detail

SNP ID Chromosome Location
rs538674557 Chr.3: 130228191-130228191 on Build GRCh38
rs963453027 Chr.3: 130228212-130228212 on Build GRCh38
rs1040013643 Chr.3: 130228240-130228240 on Build GRCh38
rs35649687 Chr.3: 130228221-130228222 on Build GRCh38
rs1023050118 Chr.3: 130228204-130228204 on Build GRCh38
rs879280560 Chr.3: 130228241-130228241 on Build GRCh38
rs866305528 Chr.3: 130228117-130228117 on Build GRCh38
rs1015020539 Chr.3: 130228103-130228103 on Build GRCh38
rs1010985389 Chr.3: 130228123-130228123 on Build GRCh38
rs981642761 Chr.3: 130228107-130228107 on Build GRCh38
rs1035879886 Chr.3: 130228182-130228182 on Build GRCh38
rs927598288 Chr.3: 130228120-130228120 on Build GRCh38
rs570027985 Chr.3: 130228131-130228131 on Build GRCh38
rs1020578967 Chr.3: 130228124-130228124 on Build GRCh38
rs138176893 Chr.3: 130228237-130228240 on Build GRCh38
rs1041836737 Chr.3: 130228211-130228211 on Build GRCh38
rs756043447 Chr.3: 130228177-130228177 on Build GRCh38
rs75046107 Chr.3: 130228130-130228130 on Build GRCh38
rs775297311 Chr.3: 130228236-130228237 on Build GRCh38
rs770382279 Chr.3: 130228219-130228219 on Build GRCh38
rs74849217 Chr.3: 130228239-130228242 on Build GRCh38
rs912376622 Chr.3: 130228216-130228216 on Build GRCh38
rs943874410 Chr.3: 130228236-130228236 on Build GRCh38
rs796414409 Chr.3: 130228213-130228213 on Build GRCh38
rs747952475 Chr.3: 130228106-130228106 on Build GRCh38
rs150926820 Chr.3: 130228202-130228202 on Build GRCh38
rs990207035 Chr.3: 130228133-130228133 on Build GRCh38

View Full Product Details