Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130250707-130251259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A4P2
Entrez Gene ID
646300
Gene Name
collagen type VI alpha 4 pseudogene 2
Gene Aliases
COL6A4
UniGene
Hs.170475
Gene Chromosome Location
Chr.3: 130212820-130273806 on Build GRCh38
Transcripts
NR_027898.1

SNP Detail

SNP ID Chromosome Location
rs750676183 Chr.3: 130251152-130251152 on Build GRCh38
rs955161529 Chr.3: 130250875-130250875 on Build GRCh38
rs928702066 Chr.3: 130250973-130250973 on Build GRCh38
rs751809990 Chr.3: 130251213-130251213 on Build GRCh38
rs772499993 Chr.3: 130250796-130250796 on Build GRCh38
rs1020624493 Chr.3: 130251135-130251135 on Build GRCh38
rs1029528846 Chr.3: 130250910-130250910 on Build GRCh38
rs569793830 Chr.3: 130251018-130251018 on Build GRCh38
rs985413151 Chr.3: 130250940-130250940 on Build GRCh38
rs993896528 Chr.3: 130250795-130250795 on Build GRCh38
rs868259503 Chr.3: 130251222-130251222 on Build GRCh38
rs775789646 Chr.3: 130250899-130250899 on Build GRCh38
rs549470895 Chr.3: 130251013-130251013 on Build GRCh38
rs960437449 Chr.3: 130251210-130251210 on Build GRCh38
rs981509418 Chr.3: 130250938-130250938 on Build GRCh38
rs1016822090 Chr.3: 130251175-130251175 on Build GRCh38
rs891864868 Chr.3: 130251142-130251142 on Build GRCh38
rs574671555 Chr.3: 130251094-130251094 on Build GRCh38
rs535736719 Chr.3: 130251105-130251105 on Build GRCh38
rs1022915501 Chr.3: 130250779-130250779 on Build GRCh38
rs953819017 Chr.3: 130251149-130251149 on Build GRCh38
rs543938477 Chr.3: 130251128-130251128 on Build GRCh38
rs72992462 Chr.3: 130251047-130251047 on Build GRCh38
rs1044589539 Chr.3: 130251104-130251104 on Build GRCh38
rs998302813 Chr.3: 130251143-130251143 on Build GRCh38
rs11924774 Chr.3: 130250900-130250900 on Build GRCh38
rs182860387 Chr.3: 130251148-130251148 on Build GRCh38
rs972823195 Chr.3: 130251019-130251019 on Build GRCh38
rs569631181 Chr.3: 130250943-130250943 on Build GRCh38
rs142298313 Chr.3: 130250927-130250927 on Build GRCh38
rs937392889 Chr.3: 130251192-130251192 on Build GRCh38
rs916336121 Chr.3: 130251234-130251234 on Build GRCh38
rs6777871 Chr.3: 130250974-130250974 on Build GRCh38
rs976561478 Chr.3: 130250907-130250907 on Build GRCh38
rs962968499 Chr.3: 130251188-130251188 on Build GRCh38
rs758067300 Chr.3: 130251191-130251191 on Build GRCh38
rs370873655 Chr.3: 130251060-130251060 on Build GRCh38
rs942194149 Chr.3: 130251215-130251215 on Build GRCh38
rs957265473 Chr.3: 130250903-130250903 on Build GRCh38
rs961355260 Chr.3: 130250771-130250771 on Build GRCh38
rs1000609954 Chr.3: 130250834-130250834 on Build GRCh38
rs941002592 Chr.3: 130250982-130250982 on Build GRCh38
rs896778194 Chr.3: 130251048-130251048 on Build GRCh38
rs918277815 Chr.3: 130251203-130251203 on Build GRCh38
rs918319617 Chr.3: 130251042-130251042 on Build GRCh38
rs140231776 Chr.3: 130251109-130251109 on Build GRCh38
rs150819552 Chr.3: 130251046-130251046 on Build GRCh38
rs552417723 Chr.3: 130251076-130251076 on Build GRCh38
rs1006619205 Chr.3: 130251101-130251101 on Build GRCh38
rs779931481 Chr.3: 130251201-130251201 on Build GRCh38
rs551806884 Chr.3: 130251001-130251001 on Build GRCh38
rs553114359 Chr.3: 130251089-130251089 on Build GRCh38
rs576465855 Chr.3: 130251113-130251113 on Build GRCh38

View Full Product Details