Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130260454-130261024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A4P2
Entrez Gene ID
646300
Gene Name
collagen type VI alpha 4 pseudogene 2
Gene Aliases
COL6A4
UniGene
Hs.170475
Gene Chromosome Location
Chr.3: 130212820-130273806 on Build GRCh38
Transcripts
NR_027898.1

SNP Detail

SNP ID Chromosome Location
rs930406076 Chr.3: 130260943-130260943 on Build GRCh38
rs540276109 Chr.3: 130260687-130260687 on Build GRCh38
rs371035384 Chr.3: 130260959-130260959 on Build GRCh38
rs995449805 Chr.3: 130260617-130260617 on Build GRCh38
rs966065171 Chr.3: 130260635-130260635 on Build GRCh38
rs560219664 Chr.3: 130260657-130260657 on Build GRCh38
rs527559801 Chr.3: 130260660-130260660 on Build GRCh38
rs992430571 Chr.3: 130260824-130260824 on Build GRCh38
rs1044064457 Chr.3: 130260907-130260907 on Build GRCh38
rs1053563364 Chr.3: 130260499-130260499 on Build GRCh38
rs545301306 Chr.3: 130260650-130260650 on Build GRCh38
rs912759747 Chr.3: 130260931-130260931 on Build GRCh38
rs561956274 Chr.3: 130260566-130260566 on Build GRCh38
rs896821 Chr.3: 130261002-130261002 on Build GRCh38
rs868683029 Chr.3: 130260996-130260996 on Build GRCh38
rs747125848 Chr.3: 130260484-130260484 on Build GRCh38
rs766451118 Chr.3: 130260602-130260602 on Build GRCh38
rs950605327 Chr.3: 130260607-130260607 on Build GRCh38
rs369740857 Chr.3: 130260942-130260942 on Build GRCh38
rs949743985 Chr.3: 130260903-130260903 on Build GRCh38
rs989122398 Chr.3: 130260780-130260780 on Build GRCh38
rs886275503 Chr.3: 130260958-130260958 on Build GRCh38
rs975190060 Chr.3: 130260638-130260638 on Build GRCh38
rs550614092 Chr.3: 130260822-130260822 on Build GRCh38
rs905659424 Chr.3: 130260549-130260549 on Build GRCh38
rs1039927707 Chr.3: 130260584-130260584 on Build GRCh38
rs1004818625 Chr.3: 130260615-130260615 on Build GRCh38
rs1008854619 Chr.3: 130260558-130260558 on Build GRCh38
rs995482240 Chr.3: 130260964-130260964 on Build GRCh38
rs561736988 Chr.3: 130260774-130260774 on Build GRCh38
rs1042249118 Chr.3: 130260544-130260544 on Build GRCh38
rs913666990 Chr.3: 130260792-130260792 on Build GRCh38
rs1044142779 Chr.3: 130260508-130260508 on Build GRCh38
rs529355570 Chr.3: 130260786-130260786 on Build GRCh38
rs573829093 Chr.3: 130260486-130260486 on Build GRCh38
rs866551931 Chr.3: 130260842-130260842 on Build GRCh38
rs74683669 Chr.3: 130260645-130260645 on Build GRCh38
rs375766080 Chr.3: 130260978-130260978 on Build GRCh38
rs891383460 Chr.3: 130260557-130260557 on Build GRCh38
rs970585158 Chr.3: 130260680-130260680 on Build GRCh38
rs368439477 Chr.3: 130260624-130260624 on Build GRCh38
rs926515451 Chr.3: 130260735-130260735 on Build GRCh38
rs981132038 Chr.3: 130260713-130260713 on Build GRCh38
rs1004745198 Chr.3: 130260985-130260985 on Build GRCh38
rs913634564 Chr.3: 130260518-130260518 on Build GRCh38
rs945086076 Chr.3: 130260797-130260797 on Build GRCh38
rs534876528 Chr.3: 130260609-130260609 on Build GRCh38
rs550858329 Chr.3: 130260984-130260984 on Build GRCh38
rs1039960197 Chr.3: 130260952-130260952 on Build GRCh38
rs551936633 Chr.3: 130260986-130260986 on Build GRCh38
rs1025189956 Chr.3: 130260663-130260663 on Build GRCh38
rs996181374 Chr.3: 130260992-130260992 on Build GRCh38
rs115307318 Chr.3: 130261003-130261003 on Build GRCh38
rs758878965 Chr.3: 130260539-130260539 on Build GRCh38
rs1029011685 Chr.3: 130260998-130261007 on Build GRCh38
rs957714665 Chr.3: 130260779-130260779 on Build GRCh38

View Full Product Details