Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 168249828-168250374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFEM1P
Entrez Gene ID
93556
Gene Name
EGF like and EMI domain containing 1, pseudogene
Gene Aliases
C3orf50, NCRNA00259
UniGene
Hs.478158
Gene Chromosome Location
Chr.3: 168249522-168830599 on Build GRCh38
Transcripts
NR_021485.2

SNP Detail

SNP ID Chromosome Location
rs898090919 Chr.3: 168249916-168249916 on Build GRCh38
rs1005447092 Chr.3: 168250209-168250209 on Build GRCh38
rs1041128849 Chr.3: 168250152-168250152 on Build GRCh38
rs542511109 Chr.3: 168249899-168249899 on Build GRCh38
rs1041563103 Chr.3: 168249860-168249860 on Build GRCh38
rs888328421 Chr.3: 168250055-168250055 on Build GRCh38
rs901294888 Chr.3: 168250176-168250176 on Build GRCh38
rs1007676933 Chr.3: 168250008-168250008 on Build GRCh38
rs1049994421 Chr.3: 168250182-168250182 on Build GRCh38
rs772822374 Chr.3: 168250348-168250351 on Build GRCh38
rs1014370985 Chr.3: 168250262-168250262 on Build GRCh38
rs961048612 Chr.3: 168250078-168250078 on Build GRCh38
rs915664913 Chr.3: 168250133-168250133 on Build GRCh38
rs548080397 Chr.3: 168250280-168250280 on Build GRCh38
rs926910661 Chr.3: 168249980-168249980 on Build GRCh38
rs1015541962 Chr.3: 168250222-168250222 on Build GRCh38
rs993786600 Chr.3: 168250220-168250220 on Build GRCh38
rs888291820 Chr.3: 168250200-168250200 on Build GRCh38
rs908211691 Chr.3: 168249914-168249914 on Build GRCh38
rs1033785961 Chr.3: 168250321-168250321 on Build GRCh38
rs565105837 Chr.3: 168250263-168250263 on Build GRCh38
rs992571344 Chr.3: 168250084-168250084 on Build GRCh38
rs949605497 Chr.3: 168249934-168249934 on Build GRCh38
rs6778880 Chr.3: 168250260-168250260 on Build GRCh38
rs561247513 Chr.3: 168249909-168249909 on Build GRCh38
rs894193193 Chr.3: 168249897-168249897 on Build GRCh38
rs993736548 Chr.3: 168249923-168249923 on Build GRCh38
rs970911706 Chr.3: 168250118-168250118 on Build GRCh38
rs1052007990 Chr.3: 168250254-168250254 on Build GRCh38
rs914492408 Chr.3: 168249852-168249852 on Build GRCh38
rs181761643 Chr.3: 168249873-168249873 on Build GRCh38
rs897475033 Chr.3: 168250168-168250168 on Build GRCh38
rs1018113226 Chr.3: 168250010-168250010 on Build GRCh38
rs1017747694 Chr.3: 168250270-168250270 on Build GRCh38
rs1037398954 Chr.3: 168250151-168250151 on Build GRCh38
rs970172263 Chr.3: 168250276-168250276 on Build GRCh38
rs1015237898 Chr.3: 168250074-168250077 on Build GRCh38
rs1008142713 Chr.3: 168250264-168250264 on Build GRCh38
rs368877857 Chr.3: 168250232-168250232 on Build GRCh38
rs1020952197 Chr.3: 168250332-168250332 on Build GRCh38
rs9290335 Chr.3: 168249896-168249896 on Build GRCh38
rs1024502878 Chr.3: 168250093-168250093 on Build GRCh38
rs1055594872 Chr.3: 168250125-168250125 on Build GRCh38
rs913891910 Chr.3: 168250027-168250027 on Build GRCh38
rs928440686 Chr.3: 168249861-168249861 on Build GRCh38
rs926796029 Chr.3: 168250127-168250127 on Build GRCh38
rs938523851 Chr.3: 168250114-168250114 on Build GRCh38
rs773147249 Chr.3: 168250239-168250239 on Build GRCh38
rs936672946 Chr.3: 168249991-168249991 on Build GRCh38
rs1010927949 Chr.3: 168250316-168250316 on Build GRCh38
rs967292026 Chr.3: 168250043-168250043 on Build GRCh38
rs75717047 Chr.3: 168250140-168250140 on Build GRCh38
rs7614524 Chr.3: 168249887-168249887 on Build GRCh38
rs945942355 Chr.3: 168249859-168249859 on Build GRCh38
rs1045307156 Chr.3: 168249948-168249948 on Build GRCh38
rs1053674457 Chr.3: 168249998-168249999 on Build GRCh38
rs1037349603 Chr.3: 168249915-168249915 on Build GRCh38
rs1028240450 Chr.3: 168250051-168250051 on Build GRCh38
rs958107262 Chr.3: 168250016-168250016 on Build GRCh38
rs989457346 Chr.3: 168250020-168250020 on Build GRCh38
rs73026250 Chr.3: 168250279-168250279 on Build GRCh38
rs900993608 Chr.3: 168250030-168250030 on Build GRCh38
rs938484214 Chr.3: 168249877-168249877 on Build GRCh38
rs769509126 Chr.3: 168250193-168250193 on Build GRCh38
rs374925322 Chr.3: 168250335-168250335 on Build GRCh38
rs532580435 Chr.3: 168249911-168249911 on Build GRCh38
rs928522316 Chr.3: 168250062-168250062 on Build GRCh38
rs989710352 Chr.3: 168250138-168250138 on Build GRCh38
rs1030525882 Chr.3: 168250004-168250004 on Build GRCh38
rs186473017 Chr.3: 168250347-168250347 on Build GRCh38
rs945399744 Chr.3: 168250146-168250146 on Build GRCh38
rs1024055681 Chr.3: 168250272-168250272 on Build GRCh38
rs1028689088 Chr.3: 168249883-168249883 on Build GRCh38
rs977306638 Chr.3: 168250049-168250049 on Build GRCh38
rs932757220 Chr.3: 168250178-168250178 on Build GRCh38
rs955040689 Chr.3: 168250005-168250005 on Build GRCh38

View Full Product Details