Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 168519418-168519965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFEM1P
Entrez Gene ID
93556
Gene Name
EGF like and EMI domain containing 1, pseudogene
Gene Aliases
C3orf50, NCRNA00259
UniGene
Hs.478158
Gene Chromosome Location
Chr.3: 168249522-168830599 on Build GRCh38
Transcripts
NR_021485.2

SNP Detail

SNP ID Chromosome Location
rs527308961 Chr.3: 168519772-168519772 on Build GRCh38
rs567917728 Chr.3: 168519904-168519904 on Build GRCh38
rs753347940 Chr.3: 168519522-168519522 on Build GRCh38
rs1449349 Chr.3: 168519826-168519826 on Build GRCh38
rs1051803168 Chr.3: 168519695-168519695 on Build GRCh38
rs370792236 Chr.3: 168519654-168519654 on Build GRCh38
rs570447573 Chr.3: 168519790-168519790 on Build GRCh38
rs990387777 Chr.3: 168519902-168519902 on Build GRCh38
rs909004187 Chr.3: 168519769-168519769 on Build GRCh38
rs981065961 Chr.3: 168519628-168519628 on Build GRCh38
rs759513355 Chr.3: 168519459-168519459 on Build GRCh38
rs1017140706 Chr.3: 168519748-168519749 on Build GRCh38
rs1041348592 Chr.3: 168519618-168519618 on Build GRCh38
rs1029788766 Chr.3: 168519506-168519506 on Build GRCh38
rs781610871 Chr.3: 168519899-168519899 on Build GRCh38
rs552423804 Chr.3: 168519776-168519776 on Build GRCh38
rs1034689868 Chr.3: 168519893-168519893 on Build GRCh38
rs1025620114 Chr.3: 168519816-168519816 on Build GRCh38
rs1033574361 Chr.3: 168519587-168519587 on Build GRCh38
rs568566449 Chr.3: 168519905-168519905 on Build GRCh38
rs912900773 Chr.3: 168519617-168519617 on Build GRCh38
rs750941443 Chr.3: 168519924-168519924 on Build GRCh38
rs765930803 Chr.3: 168519505-168519505 on Build GRCh38
rs989435718 Chr.3: 168519612-168519612 on Build GRCh38
rs537639968 Chr.3: 168519801-168519801 on Build GRCh38
rs146858100 Chr.3: 168519457-168519457 on Build GRCh38
rs937068570 Chr.3: 168519667-168519667 on Build GRCh38
rs940619811 Chr.3: 168519770-168519770 on Build GRCh38
rs934216578 Chr.3: 168519690-168519690 on Build GRCh38
rs566123933 Chr.3: 168519911-168519911 on Build GRCh38
rs751901151 Chr.3: 168519750-168519750 on Build GRCh38
rs958684352 Chr.3: 168519894-168519894 on Build GRCh38
rs889925659 Chr.3: 168519714-168519714 on Build GRCh38
rs954095453 Chr.3: 168519548-168519548 on Build GRCh38
rs190831989 Chr.3: 168519588-168519588 on Build GRCh38
rs535761311 Chr.3: 168519768-168519768 on Build GRCh38
rs1007461132 Chr.3: 168519746-168519747 on Build GRCh38
rs1051078043 Chr.3: 168519858-168519858 on Build GRCh38
rs549971572 Chr.3: 168519570-168519570 on Build GRCh38
rs569817703 Chr.3: 168519713-168519713 on Build GRCh38
rs764839402 Chr.3: 168519627-168519627 on Build GRCh38
rs533008448 Chr.3: 168519482-168519482 on Build GRCh38
rs914430597 Chr.3: 168519941-168519941 on Build GRCh38
rs897110032 Chr.3: 168519489-168519489 on Build GRCh38
rs560058019 Chr.3: 168519757-168519757 on Build GRCh38
rs1033190661 Chr.3: 168519936-168519936 on Build GRCh38
rs889895134 Chr.3: 168519860-168519860 on Build GRCh38

View Full Product Details