Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 168807402-168807995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFEM1P
Entrez Gene ID
93556
Gene Name
EGF like and EMI domain containing 1, pseudogene
Gene Aliases
C3orf50, NCRNA00259
UniGene
Hs.478158
Gene Chromosome Location
Chr.3: 168249522-168830599 on Build GRCh38
Transcripts
NR_021485.2

SNP Detail

SNP ID Chromosome Location
rs535631759 Chr.3: 168807664-168807664 on Build GRCh38
rs368989295 Chr.3: 168807966-168807967 on Build GRCh38
rs1011741963 Chr.3: 168807492-168807492 on Build GRCh38
rs557573301 Chr.3: 168807798-168807798 on Build GRCh38
rs946362177 Chr.3: 168807962-168807962 on Build GRCh38
rs568583574 Chr.3: 168807936-168807936 on Build GRCh38
rs878990648 Chr.3: 168807622-168807622 on Build GRCh38
rs1001625887 Chr.3: 168807480-168807480 on Build GRCh38
rs1021803852 Chr.3: 168807708-168807708 on Build GRCh38
rs559486189 Chr.3: 168807956-168807956 on Build GRCh38
rs562226782 Chr.3: 168807887-168807887 on Build GRCh38
rs562040272 Chr.3: 168807682-168807682 on Build GRCh38
rs934989517 Chr.3: 168807561-168807561 on Build GRCh38
rs931149849 Chr.3: 168807712-168807712 on Build GRCh38
rs997720315 Chr.3: 168807816-168807816 on Build GRCh38
rs549805356 Chr.3: 168807559-168807559 on Build GRCh38
rs987924275 Chr.3: 168807923-168807923 on Build GRCh38
rs139491121 Chr.3: 168807521-168807521 on Build GRCh38
rs952048489 Chr.3: 168807498-168807498 on Build GRCh38
rs949037158 Chr.3: 168807950-168807950 on Build GRCh38
rs181142242 Chr.3: 168807634-168807634 on Build GRCh38
rs767896748 Chr.3: 168807491-168807491 on Build GRCh38
rs949276807 Chr.3: 168807640-168807640 on Build GRCh38
rs752945882 Chr.3: 168807799-168807799 on Build GRCh38
rs1035015466 Chr.3: 168807465-168807465 on Build GRCh38
rs184107236 Chr.3: 168807793-168807793 on Build GRCh38
rs552166107 Chr.3: 168807536-168807536 on Build GRCh38
rs894756927 Chr.3: 168807692-168807692 on Build GRCh38
rs981931484 Chr.3: 168807894-168807894 on Build GRCh38
rs887874557 Chr.3: 168807754-168807754 on Build GRCh38
rs1015104029 Chr.3: 168807510-168807510 on Build GRCh38
rs1029203480 Chr.3: 168807822-168807822 on Build GRCh38
rs905062773 Chr.3: 168807473-168807473 on Build GRCh38
rs1049113593 Chr.3: 168807727-168807727 on Build GRCh38
rs144184114 Chr.3: 168807840-168807840 on Build GRCh38
rs761128637 Chr.3: 168807570-168807570 on Build GRCh38
rs998994523 Chr.3: 168807756-168807756 on Build GRCh38
rs755108095 Chr.3: 168807774-168807774 on Build GRCh38
rs1005303879 Chr.3: 168807509-168807509 on Build GRCh38
rs917588380 Chr.3: 168807947-168807947 on Build GRCh38
rs1013534261 Chr.3: 168807444-168807444 on Build GRCh38
rs896422822 Chr.3: 168807436-168807436 on Build GRCh38
rs1044907712 Chr.3: 168807453-168807453 on Build GRCh38
rs1028159002 Chr.3: 168807493-168807493 on Build GRCh38
rs369342306 Chr.3: 168807694-168807694 on Build GRCh38
rs3072600 Chr.3: 168807961-168807962 on Build GRCh38
rs188401097 Chr.3: 168807910-168807910 on Build GRCh38
rs976333159 Chr.3: 168807531-168807531 on Build GRCh38
rs1011773226 Chr.3: 168807694-168807694 on Build GRCh38
rs986116182 Chr.3: 168807552-168807552 on Build GRCh38
rs778087722 Chr.3: 168807955-168807955 on Build GRCh38
rs80093086 Chr.3: 168807556-168807556 on Build GRCh38
rs554336570 Chr.3: 168807671-168807671 on Build GRCh38
rs34538048 Chr.3: 168807932-168807933 on Build GRCh38
rs906242109 Chr.3: 168807646-168807646 on Build GRCh38
rs901885765 Chr.3: 168807802-168807802 on Build GRCh38
rs558133141 Chr.3: 168807829-168807829 on Build GRCh38
rs1017598239 Chr.3: 168807810-168807810 on Build GRCh38
rs988195462 Chr.3: 168807566-168807566 on Build GRCh38
rs374783843 Chr.3: 168807607-168807607 on Build GRCh38
rs904989192 Chr.3: 168807693-168807693 on Build GRCh38
rs541246236 Chr.3: 168807953-168807953 on Build GRCh38
rs538146671 Chr.3: 168807541-168807541 on Build GRCh38
rs926541868 Chr.3: 168807967-168807967 on Build GRCh38
rs977923499 Chr.3: 168807523-168807523 on Build GRCh38
rs544134623 Chr.3: 168807880-168807880 on Build GRCh38
rs758588738 Chr.3: 168807846-168807846 on Build GRCh38
rs1030841854 Chr.3: 168807762-168807762 on Build GRCh38
rs760624778 Chr.3: 168807481-168807481 on Build GRCh38
rs570312363 Chr.3: 168807537-168807537 on Build GRCh38
rs903380823 Chr.3: 168807729-168807729 on Build GRCh38
rs931135853 Chr.3: 168807970-168807970 on Build GRCh38
rs1002313733 Chr.3: 168807674-168807674 on Build GRCh38

View Full Product Details