Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 425316-425874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA11P
Entrez Gene ID
79963
Gene Name
ATP binding cassette subfamily A member 11, pseudogene
Gene Aliases
ABCA11, EST1133530
UniGene
Hs.677493
Gene Chromosome Location
Chr.4: 425435-474209 on Build GRCh38
Transcripts
NR_002451.2

SNP Detail

SNP ID Chromosome Location
rs900276388 Chr.4: 425739-425739 on Build GRCh38
rs139010422 Chr.4: 425617-425617 on Build GRCh38
rs539555442 Chr.4: 425538-425538 on Build GRCh38
rs911680693 Chr.4: 425671-425671 on Build GRCh38
rs982151873 Chr.4: 425569-425569 on Build GRCh38
rs1009055907 Chr.4: 425439-425439 on Build GRCh38
rs547789342 Chr.4: 425527-425527 on Build GRCh38
rs191456965 Chr.4: 425479-425479 on Build GRCh38
rs782010324 Chr.4: 425582-425582 on Build GRCh38
rs564675035 Chr.4: 425371-425371 on Build GRCh38
rs782425903 Chr.4: 425354-425354 on Build GRCh38
rs146427247 Chr.4: 425598-425598 on Build GRCh38
rs569938937 Chr.4: 425339-425339 on Build GRCh38
rs894038249 Chr.4: 425499-425499 on Build GRCh38
rs983422248 Chr.4: 425653-425653 on Build GRCh38
rs929211289 Chr.4: 425620-425620 on Build GRCh38
rs4690259 Chr.4: 425786-425786 on Build GRCh38
rs552945101 Chr.4: 425777-425777 on Build GRCh38
rs550200068 Chr.4: 425474-425474 on Build GRCh38
rs929267828 Chr.4: 425417-425417 on Build GRCh38
rs570466270 Chr.4: 425744-425744 on Build GRCh38
rs545124485 Chr.4: 425803-425803 on Build GRCh38
rs1033874941 Chr.4: 425794-425794 on Build GRCh38
rs891501573 Chr.4: 425775-425775 on Build GRCh38
rs999837059 Chr.4: 425484-425484 on Build GRCh38
rs555353584 Chr.4: 425759-425759 on Build GRCh38
rs7666919 Chr.4: 425776-425776 on Build GRCh38
rs7681990 Chr.4: 425850-425850 on Build GRCh38
rs565799034 Chr.4: 425530-425530 on Build GRCh38
rs973388381 Chr.4: 425612-425612 on Build GRCh38
rs1009846828 Chr.4: 425523-425523 on Build GRCh38
rs782131233 Chr.4: 425603-425603 on Build GRCh38
rs372580519 Chr.4: 425804-425804 on Build GRCh38
rs782041520 Chr.4: 425343-425343 on Build GRCh38
rs959330202 Chr.4: 425584-425584 on Build GRCh38
rs782660736 Chr.4: 425795-425795 on Build GRCh38
rs937994761 Chr.4: 425347-425347 on Build GRCh38
rs140983277 Chr.4: 425433-425433 on Build GRCh38
rs58876623 Chr.4: 425495-425495 on Build GRCh38
rs782601841 Chr.4: 425761-425761 on Build GRCh38
rs367928878 Chr.4: 425674-425674 on Build GRCh38
rs368513313 Chr.4: 425541-425541 on Build GRCh38
rs1038768896 Chr.4: 425450-425450 on Build GRCh38
rs906669947 Chr.4: 425779-425779 on Build GRCh38
rs1048000508 Chr.4: 425419-425419 on Build GRCh38
rs950756436 Chr.4: 425832-425832 on Build GRCh38
rs370576758 Chr.4: 425676-425676 on Build GRCh38
rs143748212 Chr.4: 425760-425760 on Build GRCh38
rs1038737570 Chr.4: 425727-425727 on Build GRCh38

View Full Product Details