Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 3941796-3942361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM86EP
Entrez Gene ID
348926
Gene Name
family with sequence similarity 86, member A pseudogene
Gene Aliases
-
UniGene
Hs.591720
Gene Chromosome Location
Chr.4: 3941760-3955428 on Build GRCh38
Transcripts
NR_130740.1 NR_130741.1 NR_130742.1

SNP Detail

SNP ID Chromosome Location
rs923776376 Chr.4: 3941966-3941966 on Build GRCh38
rs530151449 Chr.4: 3941980-3941980 on Build GRCh38
rs764074605 Chr.4: 3942029-3942032 on Build GRCh38
rs7680741 Chr.4: 3942293-3942293 on Build GRCh38
rs915051139 Chr.4: 3942184-3942184 on Build GRCh38
rs552572898 Chr.4: 3942149-3942149 on Build GRCh38
rs532651229 Chr.4: 3942216-3942216 on Build GRCh38
rs558875265 Chr.4: 3942239-3942239 on Build GRCh38
rs953060037 Chr.4: 3941938-3941938 on Build GRCh38
rs575608700 Chr.4: 3941945-3941945 on Build GRCh38
rs374884550 Chr.4: 3942308-3942308 on Build GRCh38
rs188515450 Chr.4: 3941855-3941855 on Build GRCh38
rs953334368 Chr.4: 3942185-3942185 on Build GRCh38
rs7695071 Chr.4: 3942257-3942257 on Build GRCh38
rs560089540 Chr.4: 3942045-3942045 on Build GRCh38
rs140290081 Chr.4: 3942322-3942322 on Build GRCh38
rs144803779 Chr.4: 3942059-3942059 on Build GRCh38
rs948176516 Chr.4: 3942336-3942336 on Build GRCh38
rs573214738 Chr.4: 3941874-3941874 on Build GRCh38
rs377333504 Chr.4: 3942244-3942244 on Build GRCh38
rs918710834 Chr.4: 3942235-3942235 on Build GRCh38
rs897333755 Chr.4: 3942095-3942095 on Build GRCh38
rs192044204 Chr.4: 3942245-3942245 on Build GRCh38
rs34752642 Chr.4: 3942201-3942201 on Build GRCh38
rs544816541 Chr.4: 3942033-3942033 on Build GRCh38
rs1041428131 Chr.4: 3942229-3942229 on Build GRCh38
rs947386212 Chr.4: 3941824-3941824 on Build GRCh38
rs780758430 Chr.4: 3942328-3942328 on Build GRCh38
rs73076826 Chr.4: 3941844-3941844 on Build GRCh38
rs1011069310 Chr.4: 3942289-3942289 on Build GRCh38
rs373126178 Chr.4: 3942158-3942158 on Build GRCh38
rs535813545 Chr.4: 3942148-3942148 on Build GRCh38
rs536747987 Chr.4: 3942120-3942120 on Build GRCh38
rs546046432 Chr.4: 3941901-3941901 on Build GRCh38
rs550676532 Chr.4: 3942121-3942121 on Build GRCh38
rs149397289 Chr.4: 3941960-3941960 on Build GRCh38
rs371060373 Chr.4: 3942190-3942190 on Build GRCh38
rs559660322 Chr.4: 3942022-3942022 on Build GRCh38
rs990516500 Chr.4: 3941938-3941938 on Build GRCh38
rs147328893 Chr.4: 3942305-3942305 on Build GRCh38
rs545330501 Chr.4: 3942314-3942314 on Build GRCh38
rs180927694 Chr.4: 3941912-3941912 on Build GRCh38
rs1049698205 Chr.4: 3941840-3941840 on Build GRCh38
rs994373352 Chr.4: 3942111-3942111 on Build GRCh38
rs1027217748 Chr.4: 3941935-3941935 on Build GRCh38
rs1012509133 Chr.4: 3942250-3942250 on Build GRCh38
rs911235791 Chr.4: 3942192-3942192 on Build GRCh38
rs989358162 Chr.4: 3942164-3942164 on Build GRCh38
rs989822109 Chr.4: 3942032-3942032 on Build GRCh38
rs986029186 Chr.4: 3942298-3942298 on Build GRCh38
rs113500105 Chr.4: 3942151-3942151 on Build GRCh38
rs1030724611 Chr.4: 3942157-3942157 on Build GRCh38
rs148143860 Chr.4: 3942246-3942246 on Build GRCh38
rs571462224 Chr.4: 3942093-3942093 on Build GRCh38
rs62289005 Chr.4: 3942147-3942147 on Build GRCh38
rs1008257452 Chr.4: 3941846-3941846 on Build GRCh38
rs1019745247 Chr.4: 3941847-3941847 on Build GRCh38
rs1003233991 Chr.4: 3942258-3942258 on Build GRCh38
rs956516297 Chr.4: 3942285-3942285 on Build GRCh38
rs944135131 Chr.4: 3942079-3942079 on Build GRCh38
rs1031071815 Chr.4: 3942265-3942265 on Build GRCh38
rs981868750 Chr.4: 3942155-3942155 on Build GRCh38
rs565478015 Chr.4: 3941919-3941919 on Build GRCh38
rs910062782 Chr.4: 3942335-3942335 on Build GRCh38
rs185961313 Chr.4: 3941958-3941958 on Build GRCh38
rs547032779 Chr.4: 3941999-3941999 on Build GRCh38
rs1045310705 Chr.4: 3942253-3942253 on Build GRCh38
rs930044904 Chr.4: 3942241-3942241 on Build GRCh38
rs141975452 Chr.4: 3942292-3942292 on Build GRCh38
rs931877176 Chr.4: 3942043-3942043 on Build GRCh38
rs542623903 Chr.4: 3942064-3942064 on Build GRCh38
rs1048923333 Chr.4: 3942330-3942330 on Build GRCh38
rs561396641 Chr.4: 3942137-3942137 on Build GRCh38
rs935147821 Chr.4: 3941972-3941972 on Build GRCh38
rs7676066 Chr.4: 3942299-3942299 on Build GRCh38
rs564912512 Chr.4: 3942317-3942317 on Build GRCh38

View Full Product Details