Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 3951090-3951662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM86EP
Entrez Gene ID
348926
Gene Name
family with sequence similarity 86, member A pseudogene
Gene Aliases
-
UniGene
Hs.591720
Gene Chromosome Location
Chr.4: 3941760-3955428 on Build GRCh38
Transcripts
NR_130740.1 NR_130741.1 NR_130742.1

SNP Detail

SNP ID Chromosome Location
rs552066980 Chr.4: 3951333-3951333 on Build GRCh38
rs897104093 Chr.4: 3951469-3951470 on Build GRCh38
rs1026832101 Chr.4: 3951490-3951490 on Build GRCh38
rs560723488 Chr.4: 3951139-3951139 on Build GRCh38
rs570836029 Chr.4: 3951294-3951294 on Build GRCh38
rs889639899 Chr.4: 3951245-3951245 on Build GRCh38
rs539398606 Chr.4: 3951361-3951361 on Build GRCh38
rs892890197 Chr.4: 3951117-3951117 on Build GRCh38
rs1049919833 Chr.4: 3951239-3951239 on Build GRCh38
rs888363062 Chr.4: 3951622-3951622 on Build GRCh38
rs183551219 Chr.4: 3951320-3951320 on Build GRCh38
rs952120173 Chr.4: 3951169-3951169 on Build GRCh38
rs577650322 Chr.4: 3951161-3951161 on Build GRCh38
rs954122812 Chr.4: 3951124-3951124 on Build GRCh38
rs1006257224 Chr.4: 3951628-3951628 on Build GRCh38
rs61270399 Chr.4: 3951637-3951637 on Build GRCh38
rs141286629 Chr.4: 3951223-3951223 on Build GRCh38
rs4974749 Chr.4: 3951632-3951632 on Build GRCh38
rs575454938 Chr.4: 3951260-3951260 on Build GRCh38
rs190901908 Chr.4: 3951188-3951188 on Build GRCh38
rs911006288 Chr.4: 3951582-3951582 on Build GRCh38
rs138221351 Chr.4: 3951351-3951351 on Build GRCh38
rs994065968 Chr.4: 3951472-3951472 on Build GRCh38
rs149051909 Chr.4: 3951109-3951109 on Build GRCh38
rs981726790 Chr.4: 3951187-3951187 on Build GRCh38
rs115478970 Chr.4: 3951118-3951118 on Build GRCh38
rs368416026 Chr.4: 3951439-3951439 on Build GRCh38
rs4974748 Chr.4: 3951603-3951603 on Build GRCh38
rs918435331 Chr.4: 3951602-3951602 on Build GRCh38
rs569070355 Chr.4: 3951335-3951335 on Build GRCh38
rs934354832 Chr.4: 3951190-3951190 on Build GRCh38
rs956168924 Chr.4: 3951542-3951542 on Build GRCh38
rs767611701 Chr.4: 3951638-3951638 on Build GRCh38
rs188594449 Chr.4: 3951463-3951463 on Build GRCh38
rs527821903 Chr.4: 3951574-3951574 on Build GRCh38
rs547397328 Chr.4: 3951301-3951301 on Build GRCh38
rs1052763925 Chr.4: 3951222-3951222 on Build GRCh38
rs554623917 Chr.4: 3951358-3951358 on Build GRCh38
rs889492200 Chr.4: 3951423-3951423 on Build GRCh38
rs540438387 Chr.4: 3951568-3951568 on Build GRCh38
rs887788535 Chr.4: 3951494-3951494 on Build GRCh38
rs201588776 Chr.4: 3951394-3951394 on Build GRCh38
rs192784364 Chr.4: 3951541-3951541 on Build GRCh38
rs982015255 Chr.4: 3951349-3951349 on Build GRCh38
rs1008441945 Chr.4: 3951247-3951247 on Build GRCh38
rs531695797 Chr.4: 3951185-3951185 on Build GRCh38
rs1041266320 Chr.4: 3951440-3951440 on Build GRCh38
rs1008163402 Chr.4: 3951146-3951146 on Build GRCh38
rs557913637 Chr.4: 3951416-3951416 on Build GRCh38
rs143081251 Chr.4: 3951168-3951168 on Build GRCh38
rs1003071931 Chr.4: 3951537-3951537 on Build GRCh38
rs914258651 Chr.4: 3951371-3951371 on Build GRCh38
rs879369499 Chr.4: 3951240-3951240 on Build GRCh38
rs576550766 Chr.4: 3951521-3951521 on Build GRCh38
rs952026144 Chr.4: 3951308-3951308 on Build GRCh38
rs1026257569 Chr.4: 3951297-3951297 on Build GRCh38
rs897073008 Chr.4: 3951290-3951290 on Build GRCh38
rs988500113 Chr.4: 3951363-3951363 on Build GRCh38
rs564169766 Chr.4: 3951596-3951596 on Build GRCh38
rs1048303533 Chr.4: 3951614-3951614 on Build GRCh38
rs577589294 Chr.4: 3951473-3951473 on Build GRCh38
rs372117620 Chr.4: 3951184-3951184 on Build GRCh38
rs752895562 Chr.4: 3951354-3951354 on Build GRCh38
rs867981338 Chr.4: 3951531-3951531 on Build GRCh38
rs779011205 Chr.4: 3951318-3951318 on Build GRCh38
rs142982656 Chr.4: 3951379-3951379 on Build GRCh38
rs1019920034 Chr.4: 3951152-3951152 on Build GRCh38
rs200939625 Chr.4: 3951337-3951337 on Build GRCh38
rs947101664 Chr.4: 3951229-3951229 on Build GRCh38
rs1028143953 Chr.4: 3951119-3951119 on Build GRCh38
rs566421095 Chr.4: 3951319-3951319 on Build GRCh38
rs148236152 Chr.4: 3951184-3951184 on Build GRCh38
rs4974747 Chr.4: 3951583-3951583 on Build GRCh38
rs947006713 Chr.4: 3951375-3951375 on Build GRCh38
rs113933073 Chr.4: 3951505-3951505 on Build GRCh38
rs546510222 Chr.4: 3951501-3951501 on Build GRCh38
rs62289030 Chr.4: 3951503-3951503 on Build GRCh38
rs971997307 Chr.4: 3951167-3951167 on Build GRCh38
rs183263017 Chr.4: 3951569-3951569 on Build GRCh38
rs968414714 Chr.4: 3951570-3951570 on Build GRCh38

View Full Product Details