Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70067075-70067662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSN1S2AP
Entrez Gene ID
286828
Gene Name
casein alpha s2-like A, pseudogene
Gene Aliases
CSN1S2A
UniGene
Hs.631945
Gene Chromosome Location
Chr.4: 70067386-70085273 on Build GRCh38
Transcripts
NR_003720.1

SNP Detail

SNP ID Chromosome Location
rs919805962 Chr.4: 70067113-70067113 on Build GRCh38
rs1000561331 Chr.4: 70067260-70067260 on Build GRCh38
rs148955213 Chr.4: 70067440-70067440 on Build GRCh38
rs570616933 Chr.4: 70067485-70067485 on Build GRCh38
rs998911267 Chr.4: 70067344-70067344 on Build GRCh38
rs910746754 Chr.4: 70067464-70067464 on Build GRCh38
rs1010369595 Chr.4: 70067223-70067223 on Build GRCh38
rs375004172 Chr.4: 70067396-70067396 on Build GRCh38
rs1044787523 Chr.4: 70067134-70067134 on Build GRCh38
rs894994047 Chr.4: 70067613-70067613 on Build GRCh38
rs746567324 Chr.4: 70067341-70067341 on Build GRCh38
rs968734505 Chr.4: 70067330-70067330 on Build GRCh38
rs1055416070 Chr.4: 70067304-70067304 on Build GRCh38
rs906349701 Chr.4: 70067280-70067280 on Build GRCh38
rs891987209 Chr.4: 70067214-70067214 on Build GRCh38
rs570459540 Chr.4: 70067277-70067277 on Build GRCh38
rs904771368 Chr.4: 70067245-70067245 on Build GRCh38
rs1007952574 Chr.4: 70067311-70067311 on Build GRCh38
rs1028394697 Chr.4: 70067271-70067271 on Build GRCh38
rs552444305 Chr.4: 70067403-70067403 on Build GRCh38
rs572443532 Chr.4: 70067607-70067607 on Build GRCh38
rs958914569 Chr.4: 70067105-70067105 on Build GRCh38
rs62305499 Chr.4: 70067364-70067364 on Build GRCh38
rs964989545 Chr.4: 70067476-70067476 on Build GRCh38
rs1043600075 Chr.4: 70067488-70067488 on Build GRCh38
rs938640529 Chr.4: 70067415-70067415 on Build GRCh38
rs1021607391 Chr.4: 70067329-70067329 on Build GRCh38
rs867804379 Chr.4: 70067437-70067437 on Build GRCh38
rs920295561 Chr.4: 70067234-70067234 on Build GRCh38
rs927298346 Chr.4: 70067404-70067404 on Build GRCh38
rs780141320 Chr.4: 70067186-70067186 on Build GRCh38
rs910808266 Chr.4: 70067211-70067211 on Build GRCh38
rs920250044 Chr.4: 70067515-70067515 on Build GRCh38
rs931361515 Chr.4: 70067116-70067116 on Build GRCh38
rs968786714 Chr.4: 70067147-70067147 on Build GRCh38
rs558184381 Chr.4: 70067626-70067626 on Build GRCh38
rs777559796 Chr.4: 70067274-70067274 on Build GRCh38
rs946236039 Chr.4: 70067463-70067463 on Build GRCh38
rs539547061 Chr.4: 70067608-70067608 on Build GRCh38
rs954871172 Chr.4: 70067187-70067187 on Build GRCh38
rs938734687 Chr.4: 70067170-70067170 on Build GRCh38
rs895053504 Chr.4: 70067321-70067321 on Build GRCh38
rs1003331430 Chr.4: 70067297-70067297 on Build GRCh38
rs961205229 Chr.4: 70067371-70067371 on Build GRCh38
rs988105569 Chr.4: 70067189-70067189 on Build GRCh38
rs567984440 Chr.4: 70067434-70067434 on Build GRCh38
rs770372473 Chr.4: 70067499-70067499 on Build GRCh38
rs145542935 Chr.4: 70067111-70067111 on Build GRCh38
rs1037364036 Chr.4: 70067231-70067231 on Build GRCh38
rs929041158 Chr.4: 70067238-70067238 on Build GRCh38
rs185305699 Chr.4: 70067504-70067504 on Build GRCh38
rs928990217 Chr.4: 70067563-70067563 on Build GRCh38
rs974088753 Chr.4: 70067508-70067508 on Build GRCh38
rs1047500228 Chr.4: 70067278-70067278 on Build GRCh38
rs1043348555 Chr.4: 70067225-70067225 on Build GRCh38
rs73827709 Chr.4: 70067156-70067156 on Build GRCh38
rs1050677320 Chr.4: 70067588-70067588 on Build GRCh38
rs114512099 Chr.4: 70067583-70067583 on Build GRCh38
rs1026941304 Chr.4: 70067376-70067376 on Build GRCh38
rs904803340 Chr.4: 70067564-70067564 on Build GRCh38
rs987665267 Chr.4: 70067442-70067442 on Build GRCh38
rs116749398 Chr.4: 70067324-70067324 on Build GRCh38
rs972529370 Chr.4: 70067373-70067373 on Build GRCh38

View Full Product Details