Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70133492-70133954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSN1S2BP
Entrez Gene ID
100337616
Gene Name
casein alpha s2-like B, pseudogene
Gene Aliases
CSN1S2B
UniGene
Hs.729824
Gene Chromosome Location
Chr.4: 70133604-70146704 on Build GRCh38
Transcripts
NR_033311.1

SNP Detail

SNP ID Chromosome Location
rs144683873 Chr.4: 70133813-70133813 on Build GRCh38
rs991067817 Chr.4: 70133805-70133805 on Build GRCh38
rs369651632 Chr.4: 70133536-70133536 on Build GRCh38
rs902310419 Chr.4: 70133600-70133600 on Build GRCh38
rs147179792 Chr.4: 70133610-70133610 on Build GRCh38
rs1046360358 Chr.4: 70133593-70133593 on Build GRCh38
rs550016884 Chr.4: 70133855-70133855 on Build GRCh38
rs749567420 Chr.4: 70133864-70133864 on Build GRCh38
rs949265557 Chr.4: 70133592-70133592 on Build GRCh38
rs36016184 Chr.4: 70133658-70133659 on Build GRCh38
rs558807760 Chr.4: 70133588-70133589 on Build GRCh38
rs111825841 Chr.4: 70133902-70133902 on Build GRCh38
rs890948233 Chr.4: 70133854-70133854 on Build GRCh38
rs141739797 Chr.4: 70133527-70133527 on Build GRCh38
rs893876469 Chr.4: 70133659-70133659 on Build GRCh38
rs1053667358 Chr.4: 70133616-70133616 on Build GRCh38
rs960898672 Chr.4: 70133783-70133783 on Build GRCh38
rs768743125 Chr.4: 70133896-70133896 on Build GRCh38
rs190946492 Chr.4: 70133830-70133830 on Build GRCh38
rs182288237 Chr.4: 70133831-70133831 on Build GRCh38
rs548593103 Chr.4: 70133779-70133779 on Build GRCh38
rs1399252 Chr.4: 70133902-70133902 on Build GRCh38
rs998362332 Chr.4: 70133712-70133712 on Build GRCh38
rs983559865 Chr.4: 70133770-70133770 on Build GRCh38
rs928173292 Chr.4: 70133526-70133526 on Build GRCh38
rs990475858 Chr.4: 70133578-70133578 on Build GRCh38
rs916278611 Chr.4: 70133590-70133590 on Build GRCh38
rs781765796 Chr.4: 70133673-70133673 on Build GRCh38
rs951538814 Chr.4: 70133738-70133738 on Build GRCh38
rs540079458 Chr.4: 70133669-70133669 on Build GRCh38
rs1026323211 Chr.4: 70133716-70133716 on Build GRCh38
rs780279884 Chr.4: 70133764-70133764 on Build GRCh38
rs990591973 Chr.4: 70133760-70133760 on Build GRCh38
rs970390806 Chr.4: 70133769-70133769 on Build GRCh38
rs941797877 Chr.4: 70133554-70133554 on Build GRCh38
rs777395 Chr.4: 70133742-70133742 on Build GRCh38
rs1018214801 Chr.4: 70133676-70133676 on Build GRCh38
rs901145013 Chr.4: 70133878-70133878 on Build GRCh38
rs538160517 Chr.4: 70133852-70133852 on Build GRCh38
rs572888126 Chr.4: 70133645-70133645 on Build GRCh38
rs200926733 Chr.4: 70133848-70133848 on Build GRCh38
rs946351441 Chr.4: 70133821-70133821 on Build GRCh38
rs386675759 Chr.4: 70133740-70133742 on Build GRCh38
rs75687841 Chr.4: 70133740-70133740 on Build GRCh38
rs887348499 Chr.4: 70133906-70133906 on Build GRCh38
rs140837043 Chr.4: 70133768-70133768 on Build GRCh38
rs116030092 Chr.4: 70133748-70133748 on Build GRCh38
rs183646728 Chr.4: 70133776-70133776 on Build GRCh38
rs915102580 Chr.4: 70133847-70133847 on Build GRCh38
rs946404052 Chr.4: 70133587-70133587 on Build GRCh38
rs542319979 Chr.4: 70133625-70133625 on Build GRCh38
rs556556474 Chr.4: 70133551-70133551 on Build GRCh38
rs1039585664 Chr.4: 70133888-70133888 on Build GRCh38
rs554292605 Chr.4: 70133641-70133641 on Build GRCh38
rs187897982 Chr.4: 70133822-70133822 on Build GRCh38
rs901198641 Chr.4: 70133681-70133681 on Build GRCh38
rs1051346916 Chr.4: 70133628-70133628 on Build GRCh38
rs1027670287 Chr.4: 70133746-70133746 on Build GRCh38

View Full Product Details