Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70141275-70141832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSN1S2BP
Entrez Gene ID
100337616
Gene Name
casein alpha s2-like B, pseudogene
Gene Aliases
CSN1S2B
UniGene
Hs.729824
Gene Chromosome Location
Chr.4: 70133604-70146704 on Build GRCh38
Transcripts
NR_033311.1

SNP Detail

SNP ID Chromosome Location
rs905802157 Chr.4: 70141615-70141615 on Build GRCh38
rs188897645 Chr.4: 70141647-70141647 on Build GRCh38
rs939942580 Chr.4: 70141317-70141317 on Build GRCh38
rs891811650 Chr.4: 70141713-70141713 on Build GRCh38
rs879083358 Chr.4: 70141721-70141721 on Build GRCh38
rs79654820 Chr.4: 70141749-70141749 on Build GRCh38
rs904690749 Chr.4: 70141540-70141540 on Build GRCh38
rs921495554 Chr.4: 70141677-70141677 on Build GRCh38
rs755952359 Chr.4: 70141402-70141402 on Build GRCh38
rs972536251 Chr.4: 70141321-70141321 on Build GRCh38
rs1014570161 Chr.4: 70141596-70141596 on Build GRCh38
rs535748053 Chr.4: 70141680-70141680 on Build GRCh38
rs927390399 Chr.4: 70141698-70141698 on Build GRCh38
rs889515967 Chr.4: 70141785-70141785 on Build GRCh38
rs919784891 Chr.4: 70141324-70141324 on Build GRCh38
rs1028062142 Chr.4: 70141581-70141581 on Build GRCh38
rs201863918 Chr.4: 70141622-70141622 on Build GRCh38
rs904805603 Chr.4: 70141736-70141736 on Build GRCh38
rs566615185 Chr.4: 70141622-70141623 on Build GRCh38
rs1007992792 Chr.4: 70141807-70141807 on Build GRCh38
rs1043310630 Chr.4: 70141727-70141727 on Build GRCh38
rs938794499 Chr.4: 70141702-70141702 on Build GRCh38
rs185083551 Chr.4: 70141795-70141795 on Build GRCh38
rs148713475 Chr.4: 70141416-70141416 on Build GRCh38
rs909554895 Chr.4: 70141710-70141710 on Build GRCh38
rs142293629 Chr.4: 70141512-70141512 on Build GRCh38
rs946261351 Chr.4: 70141725-70141725 on Build GRCh38
rs961254648 Chr.4: 70141633-70141633 on Build GRCh38
rs986446929 Chr.4: 70141593-70141593 on Build GRCh38
rs1002138263 Chr.4: 70141623-70141623 on Build GRCh38
rs980811010 Chr.4: 70141368-70141368 on Build GRCh38
rs751937204 Chr.4: 70141400-70141400 on Build GRCh38
rs749028564 Chr.4: 70141576-70141576 on Build GRCh38
rs953804738 Chr.4: 70141589-70141589 on Build GRCh38
rs1021540192 Chr.4: 70141648-70141648 on Build GRCh38
rs539473121 Chr.4: 70141758-70141758 on Build GRCh38
rs34321013 Chr.4: 70141623-70141624 on Build GRCh38
rs370848859 Chr.4: 70141398-70141398 on Build GRCh38
rs888172590 Chr.4: 70141536-70141536 on Build GRCh38
rs776089103 Chr.4: 70141418-70141418 on Build GRCh38
rs996785548 Chr.4: 70141542-70141542 on Build GRCh38
rs189734310 Chr.4: 70141372-70141372 on Build GRCh38
rs544011755 Chr.4: 70141363-70141363 on Build GRCh38
rs10005191 Chr.4: 70141768-70141768 on Build GRCh38
rs192846485 Chr.4: 70141600-70141600 on Build GRCh38
rs2272040 Chr.4: 70141330-70141330 on Build GRCh38
rs10028117 Chr.4: 70141670-70141670 on Build GRCh38
rs764583552 Chr.4: 70141382-70141382 on Build GRCh38
rs185256775 Chr.4: 70141641-70141641 on Build GRCh38
rs965595850 Chr.4: 70141657-70141657 on Build GRCh38
rs562116441 Chr.4: 70141364-70141364 on Build GRCh38
rs1025393778 Chr.4: 70141334-70141334 on Build GRCh38
rs768164042 Chr.4: 70141510-70141510 on Build GRCh38
rs181957179 Chr.4: 70141707-70141707 on Build GRCh38
rs977068687 Chr.4: 70141450-70141450 on Build GRCh38
rs1048102706 Chr.4: 70141527-70141527 on Build GRCh38
rs375334684 Chr.4: 70141518-70141518 on Build GRCh38
rs1032309787 Chr.4: 70141630-70141630 on Build GRCh38
rs7683989 Chr.4: 70141538-70141538 on Build GRCh38
rs976495464 Chr.4: 70141636-70141636 on Build GRCh38

View Full Product Details