Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122732234-122732814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBS12
Entrez Gene ID
166379
Gene Name
Bardet-Biedl syndrome 12
Gene Aliases
C4orf24
UniGene
Hs.400698
Gene Chromosome Location
Chr.4: 122700437-122744943 on Build GRCh38
Transcripts
NM_001178007.1 NM_152618.2 XM_011531680.2 NR_024041.1

Gene Symbol
CETN4P
Entrez Gene ID
729338
Gene Name
centrin 4, pseudogene
Gene Aliases
-
UniGene
Hs.647968
Gene Chromosome Location
Chr.4: 122730189-122732458 on Build GRCh38
Transcripts
NM_001178007.1 NM_152618.2 XM_011531680.2 NR_024041.1

SNP Detail

SNP ID Chromosome Location
rs147382055 Chr.4: 122732558-122732558 on Build GRCh38
rs976603651 Chr.4: 122732613-122732613 on Build GRCh38
rs561069126 Chr.4: 122732755-122732755 on Build GRCh38
rs1005282053 Chr.4: 122732742-122732742 on Build GRCh38
rs1006545342 Chr.4: 122732740-122732740 on Build GRCh38
rs529405874 Chr.4: 122732502-122732502 on Build GRCh38
rs541180893 Chr.4: 122732731-122732731 on Build GRCh38
rs903311719 Chr.4: 122732719-122732719 on Build GRCh38
rs879661820 Chr.4: 122732643-122732643 on Build GRCh38
rs1001749002 Chr.4: 122732501-122732501 on Build GRCh38
rs1055994200 Chr.4: 122732682-122732682 on Build GRCh38
rs542937740 Chr.4: 122732662-122732662 on Build GRCh38
rs763530067 Chr.4: 122732787-122732787 on Build GRCh38
rs34605836 Chr.4: 122732767-122732768 on Build GRCh38
rs957938967 Chr.4: 122732331-122732331 on Build GRCh38
rs1056061149 Chr.4: 122732743-122732743 on Build GRCh38
rs965613189 Chr.4: 122732604-122732604 on Build GRCh38
rs900955589 Chr.4: 122732304-122732304 on Build GRCh38
rs309357 Chr.4: 122732421-122732421 on Build GRCh38
rs189642665 Chr.4: 122732790-122732790 on Build GRCh38
rs929650005 Chr.4: 122732728-122732731 on Build GRCh38
rs534444365 Chr.4: 122732639-122732639 on Build GRCh38
rs371366523 Chr.4: 122732511-122732511 on Build GRCh38
rs977515448 Chr.4: 122732455-122732455 on Build GRCh38
rs1048376813 Chr.4: 122732429-122732429 on Build GRCh38
rs1025321064 Chr.4: 122732394-122732395 on Build GRCh38
rs1014405867 Chr.4: 122732524-122732524 on Build GRCh38
rs556105506 Chr.4: 122732671-122732671 on Build GRCh38
rs951260378 Chr.4: 122732786-122732786 on Build GRCh38
rs187837108 Chr.4: 122732473-122732473 on Build GRCh38
rs886059055 Chr.4: 122732738-122732738 on Build GRCh38
rs886059056 Chr.4: 122732743-122732743 on Build GRCh38
rs993773089 Chr.4: 122732568-122732568 on Build GRCh38
rs530027112 Chr.4: 122732393-122732393 on Build GRCh38
rs952568394 Chr.4: 122732394-122732394 on Build GRCh38
rs894322113 Chr.4: 122732751-122732751 on Build GRCh38
rs192926352 Chr.4: 122732633-122732633 on Build GRCh38
rs944959496 Chr.4: 122732538-122732539 on Build GRCh38
rs1041887429 Chr.4: 122732710-122732710 on Build GRCh38
rs926061814 Chr.4: 122732672-122732672 on Build GRCh38
rs1040624952 Chr.4: 122732715-122732715 on Build GRCh38
rs985390248 Chr.4: 122732599-122732599 on Build GRCh38
rs149872106 Chr.4: 122732677-122732677 on Build GRCh38
rs999638132 Chr.4: 122732567-122732567 on Build GRCh38
rs977386330 Chr.4: 122732647-122732647 on Build GRCh38
rs984134459 Chr.4: 122732653-122732653 on Build GRCh38
rs1040716920 Chr.4: 122732538-122732538 on Build GRCh38
rs80287628 Chr.4: 122732702-122732702 on Build GRCh38
rs977032449 Chr.4: 122732763-122732763 on Build GRCh38
rs909899008 Chr.4: 122732670-122732670 on Build GRCh38
rs374620043 Chr.4: 122732645-122732645 on Build GRCh38
rs144275141 Chr.4: 122732324-122732324 on Build GRCh38
rs309358 Chr.4: 122732771-122732771 on Build GRCh38
rs28642232 Chr.4: 122732692-122732692 on Build GRCh38
rs985337852 Chr.4: 122732359-122732359 on Build GRCh38
rs1018236637 Chr.4: 122732757-122732757 on Build GRCh38
rs1025175175 Chr.4: 122732526-122732526 on Build GRCh38
rs777022148 Chr.4: 122732380-122732380 on Build GRCh38
rs999293239 Chr.4: 122732721-122732721 on Build GRCh38
rs184098819 Chr.4: 122732728-122732728 on Build GRCh38
rs1034686486 Chr.4: 122732415-122732415 on Build GRCh38
rs766777580 Chr.4: 122732616-122732616 on Build GRCh38
rs772762064 Chr.4: 122732673-122732673 on Build GRCh38
rs753223055 Chr.4: 122732603-122732603 on Build GRCh38
rs943859156 Chr.4: 122732389-122732389 on Build GRCh38
rs967279836 Chr.4: 122732773-122732773 on Build GRCh38
rs911120603 Chr.4: 122732602-122732602 on Build GRCh38
rs533547179 Chr.4: 122732413-122732413 on Build GRCh38
rs549255498 Chr.4: 122732539-122732539 on Build GRCh38
rs309358 Chr.4: 122732771-122732771 on Build GRCh38
rs569682788 Chr.4: 122732418-122732418 on Build GRCh38
rs768803225 Chr.4: 122732369-122732369 on Build GRCh38
rs957660733 Chr.4: 122732592-122732592 on Build GRCh38
rs886059057 Chr.4: 122732786-122732786 on Build GRCh38
rs1032013952 Chr.4: 122732583-122732583 on Build GRCh38
rs911068104 Chr.4: 122732386-122732386 on Build GRCh38
rs965319873 Chr.4: 122732761-122732761 on Build GRCh38
rs773949620 Chr.4: 122732698-122732705 on Build GRCh38
rs893496118 Chr.4: 122732738-122732738 on Build GRCh38
rs61739499 Chr.4: 122732408-122732408 on Build GRCh38
rs139110565 Chr.4: 122732652-122732652 on Build GRCh38
rs896005714 Chr.4: 122732514-122732514 on Build GRCh38
rs958639949 Chr.4: 122732416-122732416 on Build GRCh38

View Full Product Details