Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80350765-80351313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRSP8P
Entrez Gene ID
441089
Gene Name
mediator complex subunit 27 pseudogene
Gene Aliases
-
UniGene
Hs.568291
Gene Chromosome Location
Chr.5: 80350605-80351966 on Build GRCh38
Transcripts
NR_003665.1

SNP Detail

SNP ID Chromosome Location
rs34240292 Chr.5: 80351021-80351021 on Build GRCh38
rs887363614 Chr.5: 80351092-80351092 on Build GRCh38
rs13177210 Chr.5: 80350969-80350969 on Build GRCh38
rs951746610 Chr.5: 80350804-80350804 on Build GRCh38
rs998704655 Chr.5: 80350920-80350920 on Build GRCh38
rs114783982 Chr.5: 80351214-80351214 on Build GRCh38
rs906791489 Chr.5: 80350858-80350858 on Build GRCh38
rs895927097 Chr.5: 80351017-80351017 on Build GRCh38
rs574131513 Chr.5: 80351021-80351022 on Build GRCh38
rs754657845 Chr.5: 80350857-80350858 on Build GRCh38
rs764711111 Chr.5: 80350843-80350843 on Build GRCh38
rs954642473 Chr.5: 80350943-80350943 on Build GRCh38
rs377379489 Chr.5: 80350960-80350960 on Build GRCh38
rs963226020 Chr.5: 80351266-80351266 on Build GRCh38
rs777677343 Chr.5: 80351276-80351277 on Build GRCh38
rs1046563 Chr.5: 80351121-80351121 on Build GRCh38
rs116822866 Chr.5: 80351269-80351269 on Build GRCh38
rs62365916 Chr.5: 80351076-80351076 on Build GRCh38
rs893836477 Chr.5: 80350867-80350867 on Build GRCh38
rs987986133 Chr.5: 80351167-80351167 on Build GRCh38
rs56291906 Chr.5: 80351020-80351020 on Build GRCh38
rs902606235 Chr.5: 80351115-80351115 on Build GRCh38
rs565297117 Chr.5: 80350844-80350844 on Build GRCh38
rs756059402 Chr.5: 80351123-80351123 on Build GRCh38
rs551054883 Chr.5: 80350889-80350889 on Build GRCh38
rs1055413192 Chr.5: 80351074-80351074 on Build GRCh38
rs60187147 Chr.5: 80351021-80351021 on Build GRCh38
rs1030546041 Chr.5: 80350936-80350936 on Build GRCh38
rs938143780 Chr.5: 80351065-80351065 on Build GRCh38
rs774921098 Chr.5: 80350919-80350919 on Build GRCh38
rs922400739 Chr.5: 80350892-80350892 on Build GRCh38
rs967954325 Chr.5: 80351217-80351217 on Build GRCh38
rs1007453549 Chr.5: 80350949-80350949 on Build GRCh38
rs561273579 Chr.5: 80350895-80350895 on Build GRCh38
rs73767743 Chr.5: 80351273-80351273 on Build GRCh38
rs530125324 Chr.5: 80350925-80350925 on Build GRCh38
rs1055148573 Chr.5: 80350862-80350862 on Build GRCh38
rs975169158 Chr.5: 80350890-80350890 on Build GRCh38
rs552156723 Chr.5: 80350970-80350970 on Build GRCh38
rs13181295 Chr.5: 80350982-80350982 on Build GRCh38
rs867158133 Chr.5: 80351019-80351020 on Build GRCh38
rs767601416 Chr.5: 80351105-80351105 on Build GRCh38
rs377490573 Chr.5: 80351001-80351001 on Build GRCh38
rs963721235 Chr.5: 80351122-80351122 on Build GRCh38
rs115024002 Chr.5: 80351253-80351253 on Build GRCh38
rs1042511600 Chr.5: 80350874-80350874 on Build GRCh38
rs963750904 Chr.5: 80350860-80350860 on Build GRCh38
rs928093680 Chr.5: 80351028-80351028 on Build GRCh38
rs1043062101 Chr.5: 80350966-80350966 on Build GRCh38
rs554538308 Chr.5: 80351059-80351059 on Build GRCh38
rs955212007 Chr.5: 80350907-80350907 on Build GRCh38
rs752148208 Chr.5: 80351108-80351108 on Build GRCh38
rs537398745 Chr.5: 80351023-80351023 on Build GRCh38
rs973473389 Chr.5: 80350973-80350973 on Build GRCh38
rs150413213 Chr.5: 80350934-80350934 on Build GRCh38
rs35606070 Chr.5: 80351114-80351114 on Build GRCh38
rs925930194 Chr.5: 80351259-80351259 on Build GRCh38
rs946741300 Chr.5: 80351086-80351086 on Build GRCh38
rs398093513 Chr.5: 80351022-80351022 on Build GRCh38
rs573440245 Chr.5: 80351246-80351246 on Build GRCh38
rs530770228 Chr.5: 80350859-80350859 on Build GRCh38
rs545346243 Chr.5: 80350796-80350796 on Build GRCh38
rs757052826 Chr.5: 80351087-80351087 on Build GRCh38
rs1055822795 Chr.5: 80351008-80351008 on Build GRCh38
rs779140457 Chr.5: 80351160-80351160 on Build GRCh38
rs560039248 Chr.5: 80351022-80351023 on Build GRCh38
rs955328905 Chr.5: 80351166-80351166 on Build GRCh38
rs918992224 Chr.5: 80350827-80350827 on Build GRCh38
rs998650141 Chr.5: 80351133-80351133 on Build GRCh38
rs1014409345 Chr.5: 80351064-80351064 on Build GRCh38
rs1010898144 Chr.5: 80350871-80350871 on Build GRCh38

View Full Product Details