Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 168530517-168531064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLL1
Entrez Gene ID
345630
Gene Name
fibrillarin-like 1
Gene Aliases
-
UniGene
Hs.166262
Gene Chromosome Location
Chr.5: 168529577-168530634 on Build GRCh38
Transcripts
NR_024356.2

SNP Detail

SNP ID Chromosome Location
rs933983336 Chr.5: 168530812-168530812 on Build GRCh38
rs1026148466 Chr.5: 168530807-168530807 on Build GRCh38
rs1005638430 Chr.5: 168530950-168530950 on Build GRCh38
rs975648621 Chr.5: 168530944-168530944 on Build GRCh38
rs1039992259 Chr.5: 168530612-168530612 on Build GRCh38
rs1016783969 Chr.5: 168530598-168530598 on Build GRCh38
rs113353163 Chr.5: 168530798-168530798 on Build GRCh38
rs777629931 Chr.5: 168530670-168530670 on Build GRCh38
rs752187124 Chr.5: 168530627-168530627 on Build GRCh38
rs757190296 Chr.5: 168530787-168530787 on Build GRCh38
rs145984608 Chr.5: 168530809-168530809 on Build GRCh38
rs541517169 Chr.5: 168530841-168530841 on Build GRCh38
rs769802153 Chr.5: 168530555-168530555 on Build GRCh38
rs951458130 Chr.5: 168530834-168530834 on Build GRCh38
rs3733977 Chr.5: 168530549-168530549 on Build GRCh38
rs566927163 Chr.5: 168530630-168530630 on Build GRCh38
rs1054087033 Chr.5: 168530961-168530961 on Build GRCh38
rs1054604930 Chr.5: 168530667-168530667 on Build GRCh38
rs987008695 Chr.5: 168530535-168530535 on Build GRCh38
rs945769373 Chr.5: 168530978-168530978 on Build GRCh38
rs572419503 Chr.5: 168530816-168530816 on Build GRCh38
rs988233250 Chr.5: 168530683-168530683 on Build GRCh38
rs905015061 Chr.5: 168530977-168530977 on Build GRCh38
rs907055837 Chr.5: 168530759-168530759 on Build GRCh38
rs963023217 Chr.5: 168530802-168530802 on Build GRCh38
rs772557080 Chr.5: 168530733-168530733 on Build GRCh38
rs1045473095 Chr.5: 168530986-168530986 on Build GRCh38
rs553463302 Chr.5: 168530596-168530596 on Build GRCh38
rs56664166 Chr.5: 168530947-168530947 on Build GRCh38
rs748246452 Chr.5: 168531011-168531011 on Build GRCh38
rs912896130 Chr.5: 168530966-168530966 on Build GRCh38
rs244894 Chr.5: 168530842-168530842 on Build GRCh38
rs138161339 Chr.5: 168530661-168530661 on Build GRCh38
rs907024680 Chr.5: 168531040-168531040 on Build GRCh38
rs530155321 Chr.5: 168530930-168530930 on Build GRCh38
rs186276013 Chr.5: 168530655-168530655 on Build GRCh38
rs912706212 Chr.5: 168530878-168530878 on Build GRCh38
rs964225389 Chr.5: 168530932-168530932 on Build GRCh38
rs1009942452 Chr.5: 168530684-168530684 on Build GRCh38
rs978858572 Chr.5: 168530760-168530760 on Build GRCh38
rs745607280 Chr.5: 168530835-168530835 on Build GRCh38
rs73318542 Chr.5: 168530765-168530765 on Build GRCh38
rs913624407 Chr.5: 168530694-168530694 on Build GRCh38
rs532479348 Chr.5: 168530803-168530803 on Build GRCh38
rs969251093 Chr.5: 168530600-168530600 on Build GRCh38
rs115098361 Chr.5: 168530955-168530955 on Build GRCh38

View Full Product Details