Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 25093412-25093972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMAHP
Entrez Gene ID
8418
Gene Name
cytidine monophospho-N-acetylneuraminic acid hydroxylase, pseudogene
Gene Aliases
CMAH, CSAH
UniGene
Hs.484918
Gene Chromosome Location
Chr.6: 25081067-25138392 on Build GRCh38
Transcripts
NR_002174.2

SNP Detail

SNP ID Chromosome Location
rs116740791 Chr.6: 25093700-25093700 on Build GRCh38
rs186902618 Chr.6: 25093522-25093522 on Build GRCh38
rs931896453 Chr.6: 25093929-25093930 on Build GRCh38
rs757694047 Chr.6: 25093733-25093733 on Build GRCh38
rs376801198 Chr.6: 25093600-25093600 on Build GRCh38
rs575204953 Chr.6: 25093657-25093657 on Build GRCh38
rs975930831 Chr.6: 25093851-25093851 on Build GRCh38
rs373592155 Chr.6: 25093442-25093442 on Build GRCh38
rs540451365 Chr.6: 25093742-25093742 on Build GRCh38
rs575115951 Chr.6: 25093731-25093731 on Build GRCh38
rs776977796 Chr.6: 25093533-25093533 on Build GRCh38
rs757401790 Chr.6: 25093585-25093585 on Build GRCh38
rs763583154 Chr.6: 25093697-25093697 on Build GRCh38
rs901754295 Chr.6: 25093527-25093527 on Build GRCh38
rs1050479279 Chr.6: 25093935-25093935 on Build GRCh38
rs932029811 Chr.6: 25093891-25093891 on Build GRCh38
rs765438232 Chr.6: 25093681-25093681 on Build GRCh38
rs890457578 Chr.6: 25093448-25093448 on Build GRCh38
rs1010726026 Chr.6: 25093651-25093651 on Build GRCh38
rs1022166570 Chr.6: 25093684-25093684 on Build GRCh38
rs114581249 Chr.6: 25093834-25093834 on Build GRCh38
rs1050102675 Chr.6: 25093937-25093937 on Build GRCh38
rs753204453 Chr.6: 25093696-25093696 on Build GRCh38
rs538435745 Chr.6: 25093770-25093770 on Build GRCh38
rs996277022 Chr.6: 25093579-25093579 on Build GRCh38
rs765455550 Chr.6: 25093594-25093594 on Build GRCh38
rs371420372 Chr.6: 25093815-25093815 on Build GRCh38
rs759502442 Chr.6: 25093553-25093553 on Build GRCh38
rs991901801 Chr.6: 25093747-25093747 on Build GRCh38
rs1040716529 Chr.6: 25093479-25093479 on Build GRCh38
rs759684840 Chr.6: 25093607-25093607 on Build GRCh38
rs373737335 Chr.6: 25093936-25093937 on Build GRCh38
rs927611201 Chr.6: 25093743-25093743 on Build GRCh38
rs796577334 Chr.6: 25093814-25093814 on Build GRCh38
rs890579684 Chr.6: 25093937-25093937 on Build GRCh38
rs751793071 Chr.6: 25093557-25093557 on Build GRCh38
rs564977688 Chr.6: 25093938-25093938 on Build GRCh38
rs1037807456 Chr.6: 25093821-25093821 on Build GRCh38
rs982500338 Chr.6: 25093758-25093758 on Build GRCh38
rs1021557644 Chr.6: 25093759-25093759 on Build GRCh38
rs750551930 Chr.6: 25093746-25093746 on Build GRCh38
rs189910792 Chr.6: 25093923-25093923 on Build GRCh38
rs968786420 Chr.6: 25093764-25093765 on Build GRCh38
rs554921005 Chr.6: 25093725-25093725 on Build GRCh38
rs781280518 Chr.6: 25093626-25093626 on Build GRCh38
rs1014670892 Chr.6: 25093670-25093670 on Build GRCh38
rs375930614 Chr.6: 25093879-25093895 on Build GRCh38
rs760886001 Chr.6: 25093516-25093516 on Build GRCh38
rs977504535 Chr.6: 25093774-25093774 on Build GRCh38

View Full Product Details