Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 25108251-25108800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMAHP
Entrez Gene ID
8418
Gene Name
cytidine monophospho-N-acetylneuraminic acid hydroxylase, pseudogene
Gene Aliases
CMAH, CSAH
UniGene
Hs.484918
Gene Chromosome Location
Chr.6: 25081067-25138392 on Build GRCh38
Transcripts
NR_002174.2 NR_027626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4597088 c.542T>C p.I181T Chr.6: 25108695-25108695 on Build GRCh38
6221382 c.658A>G p.T220A Chr.6: 25108579-25108579 on Build GRCh38
4597087 c.542T>C p.I181T Chr.6: 25108695-25108695 on Build GRCh38
1580150 c.560C>A p.P187H Chr.6: 25108677-25108677 on Build GRCh38
5796977 c.524G>A p.G175D Chr.6: 25108713-25108713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000483284 Chr.6: 25108716-25108716 on Build GRCh38
rs3215685 Chr.6: 25108762-25108762 on Build GRCh38
rs750062006 Chr.6: 25108314-25108314 on Build GRCh38
rs369592727 Chr.6: 25108696-25108696 on Build GRCh38
rs895533313 Chr.6: 25108654-25108654 on Build GRCh38
rs761480650 Chr.6: 25108302-25108302 on Build GRCh38
rs531735835 Chr.6: 25108588-25108588 on Build GRCh38
rs749946069 Chr.6: 25108766-25108766 on Build GRCh38
rs192260641 Chr.6: 25108618-25108618 on Build GRCh38
rs746011514 Chr.6: 25108698-25108699 on Build GRCh38
rs540022992 Chr.6: 25108468-25108468 on Build GRCh38
rs762818152 Chr.6: 25108429-25108429 on Build GRCh38
rs901726642 Chr.6: 25108657-25108657 on Build GRCh38
rs757221191 Chr.6: 25108648-25108648 on Build GRCh38
rs1047930644 Chr.6: 25108305-25108305 on Build GRCh38
rs917449256 Chr.6: 25108487-25108487 on Build GRCh38
rs970269995 Chr.6: 25108399-25108399 on Build GRCh38
rs302989 Chr.6: 25108313-25108313 on Build GRCh38
rs1036649652 Chr.6: 25108522-25108522 on Build GRCh38
rs1037581228 Chr.6: 25108574-25108574 on Build GRCh38
rs889216923 Chr.6: 25108554-25108554 on Build GRCh38
rs187030574 Chr.6: 25108540-25108540 on Build GRCh38
rs79916468 Chr.6: 25108763-25108763 on Build GRCh38
rs956227195 Chr.6: 25108436-25108437 on Build GRCh38
rs1009481164 Chr.6: 25108664-25108664 on Build GRCh38
rs898818139 Chr.6: 25108291-25108291 on Build GRCh38
rs147615482 Chr.6: 25108578-25108578 on Build GRCh38
rs755632090 Chr.6: 25108359-25108359 on Build GRCh38
rs756036593 Chr.6: 25108677-25108677 on Build GRCh38
rs991855923 Chr.6: 25108382-25108382 on Build GRCh38
rs1128826 Chr.6: 25108695-25108695 on Build GRCh38
rs1033227844 Chr.6: 25108735-25108735 on Build GRCh38
rs745641832 Chr.6: 25108671-25108671 on Build GRCh38
rs1000430716 Chr.6: 25108419-25108419 on Build GRCh38
rs16890087 Chr.6: 25108525-25108525 on Build GRCh38
rs751131335 Chr.6: 25108541-25108541 on Build GRCh38
rs397746701 Chr.6: 25108764-25108764 on Build GRCh38
rs764856711 Chr.6: 25108551-25108551 on Build GRCh38
rs780788839 Chr.6: 25108651-25108651 on Build GRCh38
rs781051964 Chr.6: 25108587-25108587 on Build GRCh38
rs774701745 Chr.6: 25108477-25108477 on Build GRCh38
rs1037360036 Chr.6: 25108284-25108284 on Build GRCh38
rs79378120 Chr.6: 25108767-25108767 on Build GRCh38
rs574416787 Chr.6: 25108422-25108422 on Build GRCh38
rs746599707 Chr.6: 25108321-25108321 on Build GRCh38
rs752386364 Chr.6: 25108567-25108567 on Build GRCh38
rs554437457 Chr.6: 25108369-25108369 on Build GRCh38
rs758886729 Chr.6: 25108669-25108669 on Build GRCh38
rs762407572 Chr.6: 25108479-25108479 on Build GRCh38
rs928930801 Chr.6: 25108603-25108603 on Build GRCh38
rs867701863 Chr.6: 25108334-25108334 on Build GRCh38
rs1014799299 Chr.6: 25108338-25108338 on Build GRCh38
rs757880627 Chr.6: 25108577-25108577 on Build GRCh38
rs950337013 Chr.6: 25108516-25108516 on Build GRCh38
rs2295738 Chr.6: 25108762-25108762 on Build GRCh38
rs545929395 Chr.6: 25108543-25108543 on Build GRCh38
rs375189656 Chr.6: 25108762-25108763 on Build GRCh38
rs765806433 Chr.6: 25108423-25108423 on Build GRCh38
rs937661958 Chr.6: 25108381-25108381 on Build GRCh38

View Full Product Details