Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 25114876-25115404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMAHP
Entrez Gene ID
8418
Gene Name
cytidine monophospho-N-acetylneuraminic acid hydroxylase, pseudogene
Gene Aliases
CMAH, CSAH
UniGene
Hs.484918
Gene Chromosome Location
Chr.6: 25081067-25138392 on Build GRCh38
Transcripts
NR_002174.2 NR_027626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4597061 c.20T>G p.L7R Chr.6: 25115073-25115073 on Build GRCh38
4353448 c.20T>G p.L7R Chr.6: 25115073-25115073 on Build GRCh38
4592346 c.25C>A p.L9I Chr.6: 25115068-25115068 on Build GRCh38
4592345 c.25C>A p.L9I Chr.6: 25115068-25115068 on Build GRCh38
5966410 c.55G>C p.D19H Chr.6: 25115038-25115038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2166683 Chr.6: 25115299-25115299 on Build GRCh38
rs375702525 Chr.6: 25114991-25114991 on Build GRCh38
rs302982 Chr.6: 25115169-25115169 on Build GRCh38
rs943287227 Chr.6: 25115136-25115136 on Build GRCh38
rs983204897 Chr.6: 25115130-25115130 on Build GRCh38
rs532052684 Chr.6: 25115073-25115073 on Build GRCh38
rs756102131 Chr.6: 25115255-25115255 on Build GRCh38
rs561245379 Chr.6: 25115049-25115049 on Build GRCh38
rs889149921 Chr.6: 25115106-25115106 on Build GRCh38
rs200729469 Chr.6: 25115101-25115101 on Build GRCh38
rs112196716 Chr.6: 25115373-25115373 on Build GRCh38
rs537486570 Chr.6: 25115180-25115180 on Build GRCh38
rs1000504766 Chr.6: 25115228-25115228 on Build GRCh38
rs903604243 Chr.6: 25115223-25115223 on Build GRCh38
rs113969967 Chr.6: 25115349-25115349 on Build GRCh38
rs7755584 Chr.6: 25115248-25115248 on Build GRCh38
rs1021152914 Chr.6: 25114905-25114905 on Build GRCh38
rs943402059 Chr.6: 25115382-25115382 on Build GRCh38
rs767882661 Chr.6: 25115132-25115132 on Build GRCh38
rs1020680487 Chr.6: 25115222-25115222 on Build GRCh38
rs201725492 Chr.6: 25115104-25115104 on Build GRCh38
rs368384670 Chr.6: 25115287-25115287 on Build GRCh38
rs553898381 Chr.6: 25115308-25115308 on Build GRCh38
rs972144581 Chr.6: 25115077-25115077 on Build GRCh38
rs302981 Chr.6: 25115358-25115358 on Build GRCh38
rs936231521 Chr.6: 25114912-25114912 on Build GRCh38
rs367942613 Chr.6: 25115078-25115078 on Build GRCh38
rs57247838 Chr.6: 25114949-25114954 on Build GRCh38
rs984932405 Chr.6: 25115359-25115359 on Build GRCh38
rs1033757824 Chr.6: 25115229-25115229 on Build GRCh38
rs971530500 Chr.6: 25115329-25115329 on Build GRCh38
rs370293600 Chr.6: 25114949-25114949 on Build GRCh38
rs942761885 Chr.6: 25115297-25115297 on Build GRCh38
rs777300293 Chr.6: 25115191-25115191 on Build GRCh38
rs900327233 Chr.6: 25115322-25115322 on Build GRCh38
rs927450305 Chr.6: 25115180-25115181 on Build GRCh38
rs979528692 Chr.6: 25114943-25114943 on Build GRCh38
rs551855926 Chr.6: 25115102-25115102 on Build GRCh38
rs972643477 Chr.6: 25115330-25115330 on Build GRCh38
rs1009829492 Chr.6: 25115202-25115202 on Build GRCh38
rs2273671 Chr.6: 25115068-25115068 on Build GRCh38
rs1024029842 Chr.6: 25115156-25115156 on Build GRCh38
rs980317123 Chr.6: 25115178-25115178 on Build GRCh38
rs143425637 Chr.6: 25115295-25115295 on Build GRCh38
rs891788961 Chr.6: 25114948-25114953 on Build GRCh38
rs910535031 Chr.6: 25115379-25115379 on Build GRCh38
rs1009253029 Chr.6: 25114904-25114904 on Build GRCh38
rs375000195 Chr.6: 25115069-25115069 on Build GRCh38
rs990327655 Chr.6: 25115239-25115239 on Build GRCh38
rs542521708 Chr.6: 25114916-25114916 on Build GRCh38
rs749722221 Chr.6: 25115309-25115309 on Build GRCh38
rs2273670 Chr.6: 25114994-25114994 on Build GRCh38
rs991776759 Chr.6: 25115004-25115004 on Build GRCh38
rs754789884 Chr.6: 25115050-25115050 on Build GRCh38
rs796567503 Chr.6: 25114955-25114960 on Build GRCh38
rs753723951 Chr.6: 25114997-25114997 on Build GRCh38

View Full Product Details