Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32006911-32007466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP21A1P
Entrez Gene ID
1590
Gene Name
cytochrome P450 family 21 subfamily A member 1, pseudogene
Gene Aliases
CYP21A, CYP21P, P450c21A
UniGene
Hs.654479
Gene Chromosome Location
Chr.6: 32005636-32008909 on Build GRCh38
Transcripts
NR_040090.1

SNP Detail

SNP ID Chromosome Location
rs777707809 Chr.6: 32007278-32007278 on Build GRCh38
rs924365137 Chr.6: 32007174-32007174 on Build GRCh38
rs569265740 Chr.6: 32007386-32007386 on Build GRCh38
rs751599637 Chr.6: 32007356-32007356 on Build GRCh38
rs767556005 Chr.6: 32007443-32007443 on Build GRCh38
rs754645746 Chr.6: 32006948-32006948 on Build GRCh38
rs368778838 Chr.6: 32007277-32007277 on Build GRCh38
rs41315832 Chr.6: 32007333-32007333 on Build GRCh38
rs769732238 Chr.6: 32007298-32007298 on Build GRCh38
rs541124841 Chr.6: 32007118-32007118 on Build GRCh38
rs745759338 Chr.6: 32007233-32007233 on Build GRCh38
rs757773718 Chr.6: 32007067-32007067 on Build GRCh38
rs528320222 Chr.6: 32007327-32007327 on Build GRCh38
rs756659543 Chr.6: 32007126-32007126 on Build GRCh38
rs550675891 Chr.6: 32007373-32007373 on Build GRCh38
rs62402680 Chr.6: 32007336-32007336 on Build GRCh38
rs771312641 Chr.6: 32007407-32007421 on Build GRCh38
rs574483600 Chr.6: 32007152-32007152 on Build GRCh38
rs112398415 Chr.6: 32007080-32007080 on Build GRCh38
rs752254890 Chr.6: 32006956-32006956 on Build GRCh38
rs374137317 Chr.6: 32007258-32007258 on Build GRCh38
rs757655488 Chr.6: 32006980-32006980 on Build GRCh38
rs762682770 Chr.6: 32007240-32007240 on Build GRCh38
rs776391776 Chr.6: 32007100-32007100 on Build GRCh38
rs552745567 Chr.6: 32007119-32007119 on Build GRCh38
rs751760639 Chr.6: 32007432-32007432 on Build GRCh38
rs749500941 Chr.6: 32007074-32007074 on Build GRCh38
rs41315836 Chr.6: 32007374-32007374 on Build GRCh38
rs761410667 Chr.6: 32007250-32007250 on Build GRCh38
rs563932443 Chr.6: 32007313-32007313 on Build GRCh38
rs779209947 Chr.6: 32007153-32007153 on Build GRCh38
rs990324724 Chr.6: 32007190-32007190 on Build GRCh38
rs760570773 Chr.6: 32006943-32006943 on Build GRCh38
rs768103044 Chr.6: 32007129-32007129 on Build GRCh38
rs781612255 Chr.6: 32006988-32006988 on Build GRCh38
rs200950724 Chr.6: 32007270-32007270 on Build GRCh38
rs771995112 Chr.6: 32007229-32007229 on Build GRCh38
rs373414892 Chr.6: 32007369-32007369 on Build GRCh38
rs191345746 Chr.6: 32007273-32007273 on Build GRCh38
rs28691121 Chr.6: 32006941-32006941 on Build GRCh38
rs935787976 Chr.6: 32007184-32007184 on Build GRCh38
rs372243441 Chr.6: 32007008-32007008 on Build GRCh38
rs773332680 Chr.6: 32007023-32007023 on Build GRCh38
rs745872528 Chr.6: 32007288-32007288 on Build GRCh38
rs774460064 Chr.6: 32007112-32007112 on Build GRCh38
rs772880263 Chr.6: 32007404-32007404 on Build GRCh38
rs757949937 Chr.6: 32007056-32007056 on Build GRCh38
rs540127758 Chr.6: 32007403-32007403 on Build GRCh38
rs182375882 Chr.6: 32007407-32007407 on Build GRCh38
rs760113014 Chr.6: 32007409-32007409 on Build GRCh38
rs28361034 Chr.6: 32006946-32006946 on Build GRCh38
rs761341646 Chr.6: 32007317-32007317 on Build GRCh38
rs573516769 Chr.6: 32007111-32007111 on Build GRCh38
rs759578581 Chr.6: 32007137-32007137 on Build GRCh38
rs768809260 Chr.6: 32007021-32007021 on Build GRCh38
rs750980619 Chr.6: 32007255-32007255 on Build GRCh38
rs776497918 Chr.6: 32007043-32007043 on Build GRCh38
rs781497080 Chr.6: 32007070-32007070 on Build GRCh38
rs752838965 Chr.6: 32007274-32007274 on Build GRCh38
rs541427879 Chr.6: 32007216-32007216 on Build GRCh38
rs781237156 Chr.6: 32007230-32007230 on Build GRCh38
rs386699253 Chr.6: 32007066-32007112 on Build GRCh38
rs200027963 Chr.6: 32007073-32007075 on Build GRCh38
rs754346829 Chr.6: 32007334-32007334 on Build GRCh38
rs754259087 Chr.6: 32007261-32007261 on Build GRCh38
rs763072610 Chr.6: 32006938-32006938 on Build GRCh38
rs760658843 Chr.6: 32007034-32007034 on Build GRCh38
rs563318824 Chr.6: 32007251-32007251 on Build GRCh38
rs28539678 Chr.6: 32007352-32007352 on Build GRCh38
rs759395672 Chr.6: 32007109-32007109 on Build GRCh38
rs748288708 Chr.6: 32007222-32007222 on Build GRCh38
rs747893394 Chr.6: 32007395-32007395 on Build GRCh38
rs527666626 Chr.6: 32007046-32007046 on Build GRCh38
rs111610287 Chr.6: 32007066-32007066 on Build GRCh38
rs767568777 Chr.6: 32006939-32006939 on Build GRCh38
rs778371085 Chr.6: 32006952-32006952 on Build GRCh38
rs764257334 Chr.6: 32007428-32007428 on Build GRCh38
rs777316437 Chr.6: 32007408-32007408 on Build GRCh38
rs773955594 Chr.6: 32007249-32007249 on Build GRCh38
rs778077285 Chr.6: 32007053-32007053 on Build GRCh38
rs748181528 Chr.6: 32007087-32007087 on Build GRCh38
rs568441258 Chr.6: 32007017-32007017 on Build GRCh38
rs9501393 Chr.6: 32007072-32007072 on Build GRCh38
rs770072258 Chr.6: 32007009-32007009 on Build GRCh38
rs775726282 Chr.6: 32007418-32007418 on Build GRCh38
rs765752014 Chr.6: 32007416-32007416 on Build GRCh38
rs757379200 Chr.6: 32007442-32007442 on Build GRCh38
rs140037782 Chr.6: 32007248-32007248 on Build GRCh38
rs763560671 Chr.6: 32007060-32007060 on Build GRCh38
rs766908438 Chr.6: 32007260-32007260 on Build GRCh38
rs757291783 Chr.6: 32007359-32007359 on Build GRCh38
rs780536487 Chr.6: 32007146-32007146 on Build GRCh38
rs754809704 Chr.6: 32007377-32007377 on Build GRCh38
rs770904488 Chr.6: 32007093-32007093 on Build GRCh38
rs771680841 Chr.6: 32007321-32007321 on Build GRCh38
rs763306478 Chr.6: 32007424-32007424 on Build GRCh38
rs779904135 Chr.6: 32007303-32007303 on Build GRCh38
rs780347608 Chr.6: 32007073-32007073 on Build GRCh38
rs778272467 Chr.6: 32006972-32006972 on Build GRCh38
rs377253481 Chr.6: 32007050-32007050 on Build GRCh38
rs545727458 Chr.6: 32007311-32007311 on Build GRCh38
rs750848616 Chr.6: 32007135-32007135 on Build GRCh38
rs760163886 Chr.6: 32007328-32007328 on Build GRCh38
rs768897311 Chr.6: 32007075-32007075 on Build GRCh38
rs769693341 Chr.6: 32007239-32007239 on Build GRCh38
rs765663905 Chr.6: 32007340-32007340 on Build GRCh38
rs535437184 Chr.6: 32007018-32007018 on Build GRCh38
rs747489692 Chr.6: 32006973-32006973 on Build GRCh38
rs557337677 Chr.6: 32007035-32007035 on Build GRCh38
rs753027395 Chr.6: 32007351-32007351 on Build GRCh38

View Full Product Details