Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 109305835-109306389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC162P
Entrez Gene ID
221262
Gene Name
coiled-coil domain containing 162, pseudogene
Gene Aliases
C6orf184, C6orf185, CCDC162, bA425D10.3, bA425D10.7
UniGene
Hs.373914
Gene Chromosome Location
Chr.6: 109294303-109308220 on Build GRCh38
Transcripts
NR_028595.1

SNP Detail

SNP ID Chromosome Location
rs776946093 Chr.6: 109306153-109306153 on Build GRCh38
rs545625756 Chr.6: 109306276-109306276 on Build GRCh38
rs573446818 Chr.6: 109306166-109306166 on Build GRCh38
rs750115676 Chr.6: 109306107-109306107 on Build GRCh38
rs922204860 Chr.6: 109306201-109306201 on Build GRCh38
rs559970595 Chr.6: 109306101-109306101 on Build GRCh38
rs1029782461 Chr.6: 109306280-109306280 on Build GRCh38
rs556976630 Chr.6: 109306037-109306037 on Build GRCh38
rs759626975 Chr.6: 109305946-109305946 on Build GRCh38
rs944952055 Chr.6: 109306134-109306134 on Build GRCh38
rs1049210371 Chr.6: 109306274-109306274 on Build GRCh38
rs1011165504 Chr.6: 109306210-109306210 on Build GRCh38
rs1015601736 Chr.6: 109306302-109306302 on Build GRCh38
rs892745245 Chr.6: 109306161-109306161 on Build GRCh38
rs530672894 Chr.6: 109306333-109306333 on Build GRCh38
rs779291047 Chr.6: 109305939-109305940 on Build GRCh38
rs756799671 Chr.6: 109306021-109306021 on Build GRCh38
rs764725209 Chr.6: 109306299-109306299 on Build GRCh38
rs913441956 Chr.6: 109306118-109306118 on Build GRCh38
rs940473330 Chr.6: 109306282-109306282 on Build GRCh38
rs76576241 Chr.6: 109306348-109306348 on Build GRCh38
rs779650755 Chr.6: 109306147-109306147 on Build GRCh38
rs139751785 Chr.6: 109306069-109306069 on Build GRCh38
rs755457073 Chr.6: 109305921-109305921 on Build GRCh38
rs760907808 Chr.6: 109306015-109306015 on Build GRCh38
rs1036186901 Chr.6: 109306283-109306283 on Build GRCh38
rs187215166 Chr.6: 109306253-109306253 on Build GRCh38
rs1013493385 Chr.6: 109306344-109306344 on Build GRCh38
rs973361694 Chr.6: 109306335-109306335 on Build GRCh38
rs749399850 Chr.6: 109305940-109305940 on Build GRCh38
rs947867221 Chr.6: 109306073-109306073 on Build GRCh38
rs182173935 Chr.6: 109306111-109306111 on Build GRCh38
rs770848709 Chr.6: 109305941-109305941 on Build GRCh38
rs754244476 Chr.6: 109306010-109306010 on Build GRCh38
rs976328317 Chr.6: 109306191-109306191 on Build GRCh38
rs577717346 Chr.6: 109305947-109305947 on Build GRCh38
rs529768851 Chr.6: 109305884-109305884 on Build GRCh38
rs1014767310 Chr.6: 109305858-109305858 on Build GRCh38
rs1037986396 Chr.6: 109306052-109306052 on Build GRCh38
rs776035166 Chr.6: 109305983-109305983 on Build GRCh38
rs896398573 Chr.6: 109305909-109305909 on Build GRCh38
rs931637636 Chr.6: 109306223-109306223 on Build GRCh38
rs748578622 Chr.6: 109305940-109305940 on Build GRCh38
rs1052105204 Chr.6: 109306157-109306157 on Build GRCh38
rs565433336 Chr.6: 109306320-109306320 on Build GRCh38
rs761112998 Chr.6: 109305991-109305991 on Build GRCh38
rs543330540 Chr.6: 109306086-109306086 on Build GRCh38
rs887494107 Chr.6: 109306278-109306278 on Build GRCh38
rs772275624 Chr.6: 109305990-109305991 on Build GRCh38
rs537143303 Chr.6: 109305948-109305948 on Build GRCh38
rs764751022 Chr.6: 109305992-109305992 on Build GRCh38
rs905123603 Chr.6: 109306364-109306364 on Build GRCh38
rs879236369 Chr.6: 109306102-109306102 on Build GRCh38
rs970109285 Chr.6: 109305944-109305944 on Build GRCh38

View Full Product Details