Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30371988-30372544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZP586I1420
Entrez Gene ID
222161
Gene Name
uncharacterized protein DKFZp586I1420
Gene Aliases
-
UniGene
Hs.112423
Gene Chromosome Location
Chr.7: 30370050-30372809 on Build GRCh38
Transcripts
NR_002186.1

SNP Detail

SNP ID Chromosome Location
rs1002524255 Chr.7: 30372155-30372155 on Build GRCh38
rs143022868 Chr.7: 30372182-30372182 on Build GRCh38
rs769026880 Chr.7: 30372374-30372374 on Build GRCh38
rs185346458 Chr.7: 30372404-30372404 on Build GRCh38
rs897191311 Chr.7: 30372393-30372393 on Build GRCh38
rs553757677 Chr.7: 30372405-30372405 on Build GRCh38
rs879117320 Chr.7: 30372187-30372187 on Build GRCh38
rs1007412879 Chr.7: 30372415-30372415 on Build GRCh38
rs537600594 Chr.7: 30372340-30372340 on Build GRCh38
rs769926028 Chr.7: 30372397-30372397 on Build GRCh38
rs899683876 Chr.7: 30372415-30372430 on Build GRCh38
rs529280578 Chr.7: 30372205-30372205 on Build GRCh38
rs545338456 Chr.7: 30372336-30372336 on Build GRCh38
rs79618343 Chr.7: 30372513-30372513 on Build GRCh38
rs908611320 Chr.7: 30372075-30372075 on Build GRCh38
rs955583258 Chr.7: 30372220-30372220 on Build GRCh38
rs553398887 Chr.7: 30372244-30372245 on Build GRCh38
rs181784015 Chr.7: 30372116-30372116 on Build GRCh38
rs985771023 Chr.7: 30372199-30372199 on Build GRCh38
rs148379273 Chr.7: 30372244-30372244 on Build GRCh38
rs555992472 Chr.7: 30372363-30372363 on Build GRCh38
rs1051212595 Chr.7: 30372070-30372070 on Build GRCh38
rs111286379 Chr.7: 30372488-30372488 on Build GRCh38
rs898748905 Chr.7: 30372096-30372096 on Build GRCh38
rs574439915 Chr.7: 30372395-30372395 on Build GRCh38
rs145820714 Chr.7: 30372301-30372301 on Build GRCh38
rs955955160 Chr.7: 30372453-30372453 on Build GRCh38
rs552039479 Chr.7: 30372286-30372286 on Build GRCh38
rs1038565159 Chr.7: 30372335-30372335 on Build GRCh38
rs940212616 Chr.7: 30372082-30372082 on Build GRCh38
rs1054998554 Chr.7: 30372307-30372307 on Build GRCh38
rs1012928895 Chr.7: 30372069-30372069 on Build GRCh38
rs907305473 Chr.7: 30372122-30372122 on Build GRCh38
rs777599605 Chr.7: 30372037-30372037 on Build GRCh38
rs775148761 Chr.7: 30372312-30372312 on Build GRCh38
rs924394339 Chr.7: 30372450-30372450 on Build GRCh38
rs749204076 Chr.7: 30372083-30372083 on Build GRCh38
rs564768732 Chr.7: 30372464-30372464 on Build GRCh38
rs368927365 Chr.7: 30372188-30372188 on Build GRCh38
rs948900429 Chr.7: 30372352-30372352 on Build GRCh38
rs374209310 Chr.7: 30372040-30372040 on Build GRCh38
rs934934770 Chr.7: 30372019-30372019 on Build GRCh38
rs777045929 Chr.7: 30372394-30372394 on Build GRCh38
rs1030704765 Chr.7: 30372431-30372431 on Build GRCh38
rs762285568 Chr.7: 30372434-30372434 on Build GRCh38
rs1002942783 Chr.7: 30372129-30372129 on Build GRCh38
rs910193048 Chr.7: 30372203-30372203 on Build GRCh38
rs745582671 Chr.7: 30372234-30372234 on Build GRCh38
rs893340092 Chr.7: 30372044-30372044 on Build GRCh38
rs1002993773 Chr.7: 30372378-30372378 on Build GRCh38
rs1017252925 Chr.7: 30372416-30372416 on Build GRCh38
rs572055049 Chr.7: 30372426-30372426 on Build GRCh38
rs934842214 Chr.7: 30372277-30372277 on Build GRCh38
rs190245164 Chr.7: 30372512-30372512 on Build GRCh38
rs1037118470 Chr.7: 30372388-30372388 on Build GRCh38
rs546560689 Chr.7: 30372457-30372457 on Build GRCh38
rs531933945 Chr.7: 30372166-30372166 on Build GRCh38
rs961897853 Chr.7: 30372174-30372174 on Build GRCh38
rs914523671 Chr.7: 30372012-30372012 on Build GRCh38
rs906889421 Chr.7: 30372137-30372137 on Build GRCh38
rs993862775 Chr.7: 30372177-30372177 on Build GRCh38
rs564703382 Chr.7: 30372151-30372151 on Build GRCh38
rs576713196 Chr.7: 30372467-30372467 on Build GRCh38
rs952012595 Chr.7: 30372411-30372411 on Build GRCh38
rs952020264 Chr.7: 30372184-30372184 on Build GRCh38
rs921241450 Chr.7: 30372065-30372065 on Build GRCh38
rs1031421657 Chr.7: 30372160-30372160 on Build GRCh38
rs1044662638 Chr.7: 30372490-30372490 on Build GRCh38
rs140579671 Chr.7: 30372264-30372264 on Build GRCh38
rs975442618 Chr.7: 30372045-30372045 on Build GRCh38
rs550054780 Chr.7: 30372172-30372172 on Build GRCh38
rs920057797 Chr.7: 30372348-30372349 on Build GRCh38
rs948927036 Chr.7: 30372360-30372360 on Build GRCh38
rs975842897 Chr.7: 30372439-30372439 on Build GRCh38
rs1054703954 Chr.7: 30372026-30372026 on Build GRCh38
rs547738558 Chr.7: 30372235-30372235 on Build GRCh38

View Full Product Details