Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 32655785-32656334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L1P1
Entrez Gene ID
100129460
Gene Name
DPY19L1 pseudogene 1
Gene Aliases
-
UniGene
Hs.633705
Gene Chromosome Location
Chr.7: 32580939-32719168 on Build GRCh38
Transcripts
NR_036680.1

SNP Detail

SNP ID Chromosome Location
rs1002779889 Chr.7: 32655839-32655839 on Build GRCh38
rs1028364226 Chr.7: 32655979-32655979 on Build GRCh38
rs968113676 Chr.7: 32655954-32655954 on Build GRCh38
rs571317068 Chr.7: 32656222-32656222 on Build GRCh38
rs191305560 Chr.7: 32656276-32656276 on Build GRCh38
rs555634295 Chr.7: 32656263-32656263 on Build GRCh38
rs559379606 Chr.7: 32656029-32656029 on Build GRCh38
rs1057004456 Chr.7: 32656137-32656137 on Build GRCh38
rs765053947 Chr.7: 32656127-32656127 on Build GRCh38
rs1966829 Chr.7: 32656166-32656166 on Build GRCh38
rs573574548 Chr.7: 32655994-32655994 on Build GRCh38
rs750175986 Chr.7: 32656144-32656144 on Build GRCh38
rs1000805834 Chr.7: 32656196-32656196 on Build GRCh38
rs1028393732 Chr.7: 32656237-32656237 on Build GRCh38
rs1009582453 Chr.7: 32655923-32655928 on Build GRCh38
rs561979057 Chr.7: 32656123-32656123 on Build GRCh38
rs954100728 Chr.7: 32656275-32656275 on Build GRCh38
rs534598855 Chr.7: 32656281-32656281 on Build GRCh38
rs1049264745 Chr.7: 32655813-32655813 on Build GRCh38
rs903905592 Chr.7: 32656192-32656192 on Build GRCh38
rs539261521 Chr.7: 32656162-32656162 on Build GRCh38
rs531997733 Chr.7: 32656109-32656109 on Build GRCh38
rs918891268 Chr.7: 32656078-32656078 on Build GRCh38
rs572714100 Chr.7: 32656032-32656032 on Build GRCh38
rs547602103 Chr.7: 32656134-32656134 on Build GRCh38
rs761502408 Chr.7: 32656022-32656022 on Build GRCh38
rs930287776 Chr.7: 32656112-32656112 on Build GRCh38
rs182983980 Chr.7: 32655983-32655983 on Build GRCh38
rs142808065 Chr.7: 32656114-32656114 on Build GRCh38
rs1021337141 Chr.7: 32655928-32655928 on Build GRCh38
rs188076061 Chr.7: 32656037-32656037 on Build GRCh38
rs540817282 Chr.7: 32656012-32656012 on Build GRCh38
rs1000941278 Chr.7: 32655968-32655968 on Build GRCh38
rs896716891 Chr.7: 32655916-32655916 on Build GRCh38
rs966332881 Chr.7: 32656057-32656057 on Build GRCh38
rs557663621 Chr.7: 32655828-32655828 on Build GRCh38
rs564791537 Chr.7: 32656105-32656105 on Build GRCh38
rs536904939 Chr.7: 32656234-32656234 on Build GRCh38
rs543033852 Chr.7: 32655969-32655969 on Build GRCh38
rs529283228 Chr.7: 32656126-32656126 on Build GRCh38
rs1813562 Chr.7: 32655833-32655833 on Build GRCh38
rs1470717 Chr.7: 32656147-32656147 on Build GRCh38
rs569615438 Chr.7: 32656189-32656189 on Build GRCh38
rs1056739465 Chr.7: 32655886-32655886 on Build GRCh38
rs971758618 Chr.7: 32656058-32656058 on Build GRCh38

View Full Product Details