Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 35090321-35090918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L2P1
Entrez Gene ID
554236
Gene Name
DPY19L2 pseudogene 1
Gene Aliases
-
UniGene
Hs.594815
Gene Chromosome Location
Chr.7: 35081287-35186162 on Build GRCh38
Transcripts
NR_002833.2

SNP Detail

SNP ID Chromosome Location
rs2107637 Chr.7: 35090887-35090887 on Build GRCh38
rs962510146 Chr.7: 35090873-35090873 on Build GRCh38
rs536745212 Chr.7: 35090375-35090375 on Build GRCh38
rs941220440 Chr.7: 35090561-35090561 on Build GRCh38
rs200640988 Chr.7: 35090746-35090747 on Build GRCh38
rs772272270 Chr.7: 35090393-35090393 on Build GRCh38
rs528917411 Chr.7: 35090827-35090827 on Build GRCh38
rs71537666 Chr.7: 35090679-35090679 on Build GRCh38
rs748437355 Chr.7: 35090392-35090392 on Build GRCh38
rs766765306 Chr.7: 35090439-35090439 on Build GRCh38
rs6973304 Chr.7: 35090729-35090729 on Build GRCh38
rs866924415 Chr.7: 35090666-35090666 on Build GRCh38
rs893200044 Chr.7: 35090693-35090693 on Build GRCh38
rs763540581 Chr.7: 35090422-35090422 on Build GRCh38
rs746883943 Chr.7: 35090397-35090397 on Build GRCh38
rs13229724 Chr.7: 35090852-35090852 on Build GRCh38
rs553370936 Chr.7: 35090470-35090470 on Build GRCh38
rs1050959407 Chr.7: 35090610-35090610 on Build GRCh38
rs933903010 Chr.7: 35090590-35090590 on Build GRCh38
rs755285685 Chr.7: 35090441-35090441 on Build GRCh38
rs181028762 Chr.7: 35090883-35090883 on Build GRCh38
rs193223271 Chr.7: 35090562-35090562 on Build GRCh38
rs559023961 Chr.7: 35090644-35090644 on Build GRCh38
rs770791629 Chr.7: 35090400-35090400 on Build GRCh38
rs990488108 Chr.7: 35090862-35090862 on Build GRCh38
rs555354370 Chr.7: 35090440-35090440 on Build GRCh38
rs749303815 Chr.7: 35090480-35090481 on Build GRCh38
rs780968252 Chr.7: 35090493-35090493 on Build GRCh38
rs771078304 Chr.7: 35090488-35090488 on Build GRCh38
rs13230044 Chr.7: 35090870-35090870 on Build GRCh38
rs762601084 Chr.7: 35090416-35090416 on Build GRCh38
rs747112920 Chr.7: 35090487-35090487 on Build GRCh38
rs1034564627 Chr.7: 35090837-35090840 on Build GRCh38
rs777950140 Chr.7: 35090483-35090483 on Build GRCh38
rs575785446 Chr.7: 35090675-35090675 on Build GRCh38
rs755413204 Chr.7: 35090384-35090384 on Build GRCh38
rs759218245 Chr.7: 35090406-35090406 on Build GRCh38
rs778982206 Chr.7: 35090389-35090389 on Build GRCh38
rs573298091 Chr.7: 35090476-35090476 on Build GRCh38
rs780142723 Chr.7: 35090465-35090465 on Build GRCh38
rs918426236 Chr.7: 35090884-35090884 on Build GRCh38
rs773428940 Chr.7: 35090395-35090395 on Build GRCh38
rs776442229 Chr.7: 35090402-35090402 on Build GRCh38
rs779400528 Chr.7: 35090454-35090454 on Build GRCh38
rs189385693 Chr.7: 35090835-35090835 on Build GRCh38
rs780417359 Chr.7: 35090380-35090380 on Build GRCh38
rs761131927 Chr.7: 35090435-35090435 on Build GRCh38
rs749646305 Chr.7: 35090382-35090382 on Build GRCh38
rs6973067 Chr.7: 35090842-35090842 on Build GRCh38
rs769676786 Chr.7: 35090410-35090410 on Build GRCh38
rs750866245 Chr.7: 35090357-35090357 on Build GRCh38
rs751095489 Chr.7: 35090431-35090431 on Build GRCh38
rs377501982 Chr.7: 35090840-35090840 on Build GRCh38
rs200972612 Chr.7: 35090411-35090411 on Build GRCh38
rs971506350 Chr.7: 35090750-35090750 on Build GRCh38
rs896967059 Chr.7: 35090573-35090573 on Build GRCh38
rs909648281 Chr.7: 35090872-35090872 on Build GRCh38
rs933808527 Chr.7: 35090894-35090894 on Build GRCh38
rs756564906 Chr.7: 35090374-35090375 on Build GRCh38

View Full Product Details