Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 35102471-35103020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L2P1
Entrez Gene ID
554236
Gene Name
DPY19L2 pseudogene 1
Gene Aliases
-
UniGene
Hs.594815
Gene Chromosome Location
Chr.7: 35081287-35186162 on Build GRCh38
Transcripts
NR_002833.2

SNP Detail

SNP ID Chromosome Location
rs916349659 Chr.7: 35102876-35102876 on Build GRCh38
rs2908078 Chr.7: 35102823-35102823 on Build GRCh38
rs778629959 Chr.7: 35102977-35102977 on Build GRCh38
rs570127896 Chr.7: 35102955-35102955 on Build GRCh38
rs187922727 Chr.7: 35102768-35102768 on Build GRCh38
rs770557518 Chr.7: 35102988-35102988 on Build GRCh38
rs990336421 Chr.7: 35102761-35102761 on Build GRCh38
rs1038585115 Chr.7: 35102503-35102503 on Build GRCh38
rs527381323 Chr.7: 35102841-35102841 on Build GRCh38
rs770555307 Chr.7: 35102830-35102830 on Build GRCh38
rs541021823 Chr.7: 35102725-35102725 on Build GRCh38
rs967656727 Chr.7: 35102792-35102792 on Build GRCh38
rs149679635 Chr.7: 35102746-35102746 on Build GRCh38
rs866194441 Chr.7: 35102530-35102530 on Build GRCh38
rs184994683 Chr.7: 35102820-35102820 on Build GRCh38
rs1012092598 Chr.7: 35102502-35102502 on Build GRCh38
rs561413632 Chr.7: 35102855-35102855 on Build GRCh38
rs994039707 Chr.7: 35102531-35102531 on Build GRCh38
rs527654439 Chr.7: 35102799-35102799 on Build GRCh38
rs753710706 Chr.7: 35102967-35102967 on Build GRCh38
rs754921115 Chr.7: 35102974-35102974 on Build GRCh38
rs2392341 Chr.7: 35102805-35102805 on Build GRCh38
rs1038242618 Chr.7: 35102908-35102908 on Build GRCh38
rs746695070 Chr.7: 35102986-35102986 on Build GRCh38
rs955357867 Chr.7: 35102663-35102663 on Build GRCh38
rs2860550 Chr.7: 35102928-35102928 on Build GRCh38
rs4000003 Chr.7: 35102822-35102823 on Build GRCh38
rs748006323 Chr.7: 35102980-35102980 on Build GRCh38
rs1025641182 Chr.7: 35102609-35102609 on Build GRCh38
rs557786580 Chr.7: 35102553-35102553 on Build GRCh38
rs1008296726 Chr.7: 35102690-35102690 on Build GRCh38
rs560245061 Chr.7: 35102787-35102787 on Build GRCh38
rs2908076 Chr.7: 35102653-35102653 on Build GRCh38
rs766513423 Chr.7: 35102959-35102959 on Build GRCh38
rs749765960 Chr.7: 35102997-35102997 on Build GRCh38
rs764497655 Chr.7: 35102916-35102916 on Build GRCh38
rs767752967 Chr.7: 35102937-35102937 on Build GRCh38
rs780851511 Chr.7: 35102990-35102990 on Build GRCh38
rs373831272 Chr.7: 35102509-35102509 on Build GRCh38
rs544151722 Chr.7: 35102760-35102760 on Build GRCh38
rs770668947 Chr.7: 35102897-35102898 on Build GRCh38
rs397842356 Chr.7: 35102826-35102827 on Build GRCh38
rs898422212 Chr.7: 35102520-35102520 on Build GRCh38
rs1056504224 Chr.7: 35102822-35102822 on Build GRCh38
rs774805090 Chr.7: 35102930-35102930 on Build GRCh38
rs761905500 Chr.7: 35102936-35102936 on Build GRCh38
rs763619876 Chr.7: 35102909-35102909 on Build GRCh38
rs758167681 Chr.7: 35102982-35102982 on Build GRCh38
rs2908077 Chr.7: 35102680-35102680 on Build GRCh38
rs192608851 Chr.7: 35102804-35102804 on Build GRCh38
rs532039610 Chr.7: 35102875-35102876 on Build GRCh38
rs545111921 Chr.7: 35102696-35102696 on Build GRCh38
rs371154581 Chr.7: 35102757-35102757 on Build GRCh38
rs1056207281 Chr.7: 35102491-35102491 on Build GRCh38
rs770610664 Chr.7: 35102902-35102902 on Build GRCh38
rs994070877 Chr.7: 35102983-35102983 on Build GRCh38
rs547037113 Chr.7: 35102887-35102887 on Build GRCh38
rs750660583 Chr.7: 35102939-35102939 on Build GRCh38
rs115267440 Chr.7: 35102813-35102813 on Build GRCh38

View Full Product Details