Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 35178700-35179294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L2P1
Entrez Gene ID
554236
Gene Name
DPY19L2 pseudogene 1
Gene Aliases
-
UniGene
Hs.594815
Gene Chromosome Location
Chr.7: 35081287-35186162 on Build GRCh38
Transcripts
NR_002833.2

SNP Detail

SNP ID Chromosome Location
rs901497253 Chr.7: 35179038-35179038 on Build GRCh38
rs5019626 Chr.7: 35178821-35178821 on Build GRCh38
rs534908854 Chr.7: 35179163-35179163 on Build GRCh38
rs5019623 Chr.7: 35178911-35178911 on Build GRCh38
rs1032178372 Chr.7: 35179084-35179084 on Build GRCh38
rs764466825 Chr.7: 35179250-35179250 on Build GRCh38
rs796687234 Chr.7: 35179161-35179162 on Build GRCh38
rs551169454 Chr.7: 35178907-35178907 on Build GRCh38
rs1015122751 Chr.7: 35179000-35179000 on Build GRCh38
rs114773564 Chr.7: 35179129-35179129 on Build GRCh38
rs527500381 Chr.7: 35178875-35178875 on Build GRCh38
rs1028886520 Chr.7: 35179042-35179042 on Build GRCh38
rs576084914 Chr.7: 35178818-35178818 on Build GRCh38
rs561929014 Chr.7: 35178849-35178849 on Build GRCh38
rs542166792 Chr.7: 35178834-35178834 on Build GRCh38
rs925328871 Chr.7: 35178727-35178727 on Build GRCh38
rs917920101 Chr.7: 35178790-35178790 on Build GRCh38
rs111922351 Chr.7: 35179161-35179161 on Build GRCh38
rs545577922 Chr.7: 35178767-35178767 on Build GRCh38
rs1000780402 Chr.7: 35179245-35179245 on Build GRCh38
rs529223961 Chr.7: 35179075-35179075 on Build GRCh38
rs5019625 Chr.7: 35178880-35178880 on Build GRCh38
rs931569854 Chr.7: 35179156-35179157 on Build GRCh38
rs918022024 Chr.7: 35179110-35179110 on Build GRCh38
rs569060777 Chr.7: 35179030-35179030 on Build GRCh38
rs539578386 Chr.7: 35179041-35179041 on Build GRCh38
rs569746345 Chr.7: 35179031-35179031 on Build GRCh38
rs763670188 Chr.7: 35178786-35178786 on Build GRCh38
rs956563798 Chr.7: 35179095-35179095 on Build GRCh38
rs979326276 Chr.7: 35178726-35178726 on Build GRCh38
rs977819647 Chr.7: 35178752-35178752 on Build GRCh38
rs34304105 Chr.7: 35179162-35179162 on Build GRCh38
rs549849650 Chr.7: 35179020-35179020 on Build GRCh38
rs185728680 Chr.7: 35179227-35179227 on Build GRCh38
rs549459646 Chr.7: 35179157-35179158 on Build GRCh38
rs1004587447 Chr.7: 35178924-35178924 on Build GRCh38
rs949392835 Chr.7: 35179122-35179122 on Build GRCh38
rs866641452 Chr.7: 35178840-35178840 on Build GRCh38
rs1052394959 Chr.7: 35178758-35178759 on Build GRCh38
rs953287838 Chr.7: 35179045-35179045 on Build GRCh38
rs988213992 Chr.7: 35179103-35179103 on Build GRCh38
rs796277573 Chr.7: 35179157-35179158 on Build GRCh38
rs1045305489 Chr.7: 35178830-35178830 on Build GRCh38
rs201158021 Chr.7: 35179161-35179162 on Build GRCh38
rs926862483 Chr.7: 35179147-35179147 on Build GRCh38
rs997162122 Chr.7: 35179235-35179235 on Build GRCh38
rs901607424 Chr.7: 35179169-35179169 on Build GRCh38
rs73691631 Chr.7: 35179162-35179162 on Build GRCh38
rs780873706 Chr.7: 35179187-35179187 on Build GRCh38
rs1032292466 Chr.7: 35179270-35179270 on Build GRCh38
rs888995109 Chr.7: 35179244-35179245 on Build GRCh38
rs533506875 Chr.7: 35178923-35178923 on Build GRCh38
rs1029083272 Chr.7: 35179243-35179244 on Build GRCh38
rs143718247 Chr.7: 35179157-35179157 on Build GRCh38
rs1048668084 Chr.7: 35179160-35179160 on Build GRCh38
rs556236683 Chr.7: 35178804-35178804 on Build GRCh38

View Full Product Details