Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 38685400-38685952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM183BP
Entrez Gene ID
340286
Gene Name
acyloxyacyl hydrolase (neutrophil)
Gene Aliases
FAM183B, THEG6
UniGene
Hs.144075
Gene Chromosome Location
Chr.7: 38685346-38687089 on Build GRCh38
Transcripts
NR_028347.1

SNP Detail

SNP ID Chromosome Location
rs759540476 Chr.7: 38685653-38685653 on Build GRCh38
rs753121473 Chr.7: 38685777-38685777 on Build GRCh38
rs145599176 Chr.7: 38685693-38685693 on Build GRCh38
rs764414668 Chr.7: 38685885-38685885 on Build GRCh38
rs768399766 Chr.7: 38685822-38685822 on Build GRCh38
rs932378250 Chr.7: 38685661-38685661 on Build GRCh38
rs780173876 Chr.7: 38685919-38685919 on Build GRCh38
rs747242304 Chr.7: 38685706-38685706 on Build GRCh38
rs375214802 Chr.7: 38685731-38685731 on Build GRCh38
rs751083448 Chr.7: 38685680-38685680 on Build GRCh38
rs772717151 Chr.7: 38685855-38685855 on Build GRCh38
rs777017874 Chr.7: 38685670-38685671 on Build GRCh38
rs777942219 Chr.7: 38685624-38685624 on Build GRCh38
rs767943629 Chr.7: 38685452-38685452 on Build GRCh38
rs1024396311 Chr.7: 38685440-38685440 on Build GRCh38
rs370914399 Chr.7: 38685817-38685817 on Build GRCh38
rs562305778 Chr.7: 38685679-38685679 on Build GRCh38
rs1033964721 Chr.7: 38685534-38685534 on Build GRCh38
rs754386340 Chr.7: 38685685-38685685 on Build GRCh38
rs984104891 Chr.7: 38685814-38685814 on Build GRCh38
rs774177005 Chr.7: 38685835-38685835 on Build GRCh38
rs767067349 Chr.7: 38685761-38685761 on Build GRCh38
rs760070934 Chr.7: 38685766-38685766 on Build GRCh38
rs906691527 Chr.7: 38685775-38685775 on Build GRCh38
rs368968799 Chr.7: 38685915-38685915 on Build GRCh38
rs746995709 Chr.7: 38685644-38685644 on Build GRCh38
rs757582617 Chr.7: 38685891-38685891 on Build GRCh38
rs777015558 Chr.7: 38685431-38685431 on Build GRCh38
rs769751140 Chr.7: 38685657-38685657 on Build GRCh38
rs879133977 Chr.7: 38685509-38685516 on Build GRCh38
rs201780133 Chr.7: 38685808-38685808 on Build GRCh38
rs957966829 Chr.7: 38685506-38685506 on Build GRCh38
rs773928162 Chr.7: 38685738-38685738 on Build GRCh38
rs868043861 Chr.7: 38685746-38685746 on Build GRCh38
rs771203136 Chr.7: 38685708-38685708 on Build GRCh38
rs752967629 Chr.7: 38685695-38685695 on Build GRCh38
rs757955321 Chr.7: 38685581-38685581 on Build GRCh38
rs556587223 Chr.7: 38685596-38685596 on Build GRCh38
rs1021140881 Chr.7: 38685511-38685511 on Build GRCh38
rs970514062 Chr.7: 38685442-38685442 on Build GRCh38
rs61745354 Chr.7: 38685771-38685771 on Build GRCh38
rs759792874 Chr.7: 38685928-38685928 on Build GRCh38
rs17416951 Chr.7: 38685557-38685557 on Build GRCh38
rs766543365 Chr.7: 38685618-38685620 on Build GRCh38
rs375635929 Chr.7: 38685918-38685918 on Build GRCh38
rs762687629 Chr.7: 38685675-38685675 on Build GRCh38
rs755405890 Chr.7: 38685597-38685597 on Build GRCh38
rs754899319 Chr.7: 38685925-38685925 on Build GRCh38
rs760707452 Chr.7: 38685603-38685604 on Build GRCh38
rs888776328 Chr.7: 38685429-38685429 on Build GRCh38
rs6977659 Chr.7: 38685518-38685518 on Build GRCh38
rs747824586 Chr.7: 38685851-38685851 on Build GRCh38
rs748454146 Chr.7: 38685609-38685609 on Build GRCh38
rs1001602790 Chr.7: 38685866-38685866 on Build GRCh38
rs749889426 Chr.7: 38685763-38685763 on Build GRCh38
rs763708717 Chr.7: 38685590-38685590 on Build GRCh38
rs547012619 Chr.7: 38685878-38685878 on Build GRCh38
rs758748141 Chr.7: 38685780-38685780 on Build GRCh38
rs749231427 Chr.7: 38685821-38685821 on Build GRCh38
rs373489323 Chr.7: 38685852-38685852 on Build GRCh38
rs775401583 Chr.7: 38685668-38685668 on Build GRCh38
rs761165910 Chr.7: 38685681-38685681 on Build GRCh38
rs554279446 Chr.7: 38685435-38685435 on Build GRCh38
rs769803997 Chr.7: 38685719-38685719 on Build GRCh38
rs574329233 Chr.7: 38685496-38685496 on Build GRCh38
rs770914779 Chr.7: 38685648-38685648 on Build GRCh38
rs779795402 Chr.7: 38685820-38685820 on Build GRCh38
rs775928436 Chr.7: 38685879-38685879 on Build GRCh38
rs980112659 Chr.7: 38685479-38685479 on Build GRCh38
rs767780877 Chr.7: 38685912-38685912 on Build GRCh38
rs1009731639 Chr.7: 38685760-38685760 on Build GRCh38
rs781389017 Chr.7: 38685714-38685714 on Build GRCh38
rs749219684 Chr.7: 38685921-38685921 on Build GRCh38
rs760122112 Chr.7: 38685873-38685873 on Build GRCh38
rs757531246 Chr.7: 38685796-38685796 on Build GRCh38
rs373121392 Chr.7: 38685794-38685794 on Build GRCh38
rs756674304 Chr.7: 38685594-38685594 on Build GRCh38
rs765862801 Chr.7: 38685877-38685877 on Build GRCh38
rs1037283429 Chr.7: 38685739-38685739 on Build GRCh38
rs561171468 Chr.7: 38685757-38685757 on Build GRCh38
rs529719469 Chr.7: 38685537-38685537 on Build GRCh38
rs755459118 Chr.7: 38685692-38685692 on Build GRCh38
rs764516256 Chr.7: 38685787-38685787 on Build GRCh38
rs775384446 Chr.7: 38685727-38685727 on Build GRCh38
rs976955890 Chr.7: 38685543-38685543 on Build GRCh38
rs745901723 Chr.7: 38685813-38685813 on Build GRCh38
rs779221398 Chr.7: 38685608-38685608 on Build GRCh38
rs762585621 Chr.7: 38685730-38685730 on Build GRCh38
rs750983478 Chr.7: 38685593-38685593 on Build GRCh38
rs763486308 Chr.7: 38685881-38685881 on Build GRCh38
rs772138490 Chr.7: 38685617-38685617 on Build GRCh38
rs184847620 Chr.7: 38685683-38685683 on Build GRCh38
rs745848716 Chr.7: 38685717-38685717 on Build GRCh38
rs533890733 Chr.7: 38685512-38685512 on Build GRCh38
rs373142679 Chr.7: 38685696-38685696 on Build GRCh38
rs777801963 Chr.7: 38685705-38685705 on Build GRCh38
rs375311276 Chr.7: 38685669-38685669 on Build GRCh38

View Full Product Details