Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55682613-55683158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP9P1
Entrez Gene ID
360132
Gene Name
FK506 binding protein 9 pseudogene 1
Gene Aliases
FKBP9L
UniGene
Hs.446691
Gene Chromosome Location
Chr.7: 55681074-55704567 on Build GRCh38
Transcripts
NR_003949.1 NR_027339.1 NR_027340.1 NR_027342.1

SNP Detail

SNP ID Chromosome Location
rs903220192 Chr.7: 55682935-55682935 on Build GRCh38
rs13310841 Chr.7: 55682640-55682640 on Build GRCh38
rs753012275 Chr.7: 55682865-55682865 on Build GRCh38
rs543266022 Chr.7: 55682927-55682927 on Build GRCh38
rs192047867 Chr.7: 55682874-55682874 on Build GRCh38
rs755097915 Chr.7: 55682714-55682714 on Build GRCh38
rs545269063 Chr.7: 55683082-55683082 on Build GRCh38
rs781050015 Chr.7: 55682853-55682854 on Build GRCh38
rs763012792 Chr.7: 55682839-55682839 on Build GRCh38
rs973777205 Chr.7: 55683055-55683055 on Build GRCh38
rs1054831772 Chr.7: 55682969-55682969 on Build GRCh38
rs200141514 Chr.7: 55682808-55682808 on Build GRCh38
rs771364061 Chr.7: 55682724-55682724 on Build GRCh38
rs1045971314 Chr.7: 55682925-55682925 on Build GRCh38
rs1008182783 Chr.7: 55682991-55682991 on Build GRCh38
rs2961433 Chr.7: 55682864-55682864 on Build GRCh38
rs766256740 Chr.7: 55682761-55682761 on Build GRCh38
rs11238374 Chr.7: 55682697-55682697 on Build GRCh38
rs893240584 Chr.7: 55682972-55682972 on Build GRCh38
rs981976102 Chr.7: 55683115-55683115 on Build GRCh38
rs746888691 Chr.7: 55682903-55682903 on Build GRCh38
rs569899646 Chr.7: 55682722-55682722 on Build GRCh38
rs777970785 Chr.7: 55682873-55682873 on Build GRCh38
rs540772948 Chr.7: 55683027-55683027 on Build GRCh38
rs553401906 Chr.7: 55682846-55682846 on Build GRCh38
rs540677502 Chr.7: 55682696-55682696 on Build GRCh38
rs756653191 Chr.7: 55682793-55682793 on Build GRCh38
rs757051009 Chr.7: 55682851-55682852 on Build GRCh38
rs528844579 Chr.7: 55682639-55682639 on Build GRCh38
rs567664958 Chr.7: 55682790-55682790 on Build GRCh38
rs778107384 Chr.7: 55682795-55682795 on Build GRCh38
rs750414512 Chr.7: 55682737-55682737 on Build GRCh38
rs934680419 Chr.7: 55682936-55682936 on Build GRCh38
rs762634790 Chr.7: 55682739-55682739 on Build GRCh38
rs13311251 Chr.7: 55682661-55682661 on Build GRCh38
rs558913861 Chr.7: 55683062-55683062 on Build GRCh38
rs139805707 Chr.7: 55682635-55682635 on Build GRCh38
rs749642106 Chr.7: 55682723-55682723 on Build GRCh38
rs563844261 Chr.7: 55683105-55683105 on Build GRCh38
rs745795382 Chr.7: 55682861-55682861 on Build GRCh38
rs781491264 Chr.7: 55682718-55682718 on Build GRCh38
rs140672300 Chr.7: 55682984-55682984 on Build GRCh38
rs776070633 Chr.7: 55682821-55682821 on Build GRCh38
rs11524166 Chr.7: 55682704-55682704 on Build GRCh38
rs961065111 Chr.7: 55682840-55682840 on Build GRCh38
rs201762383 Chr.7: 55682996-55682997 on Build GRCh38
rs746816023 Chr.7: 55682845-55682846 on Build GRCh38
rs765180834 Chr.7: 55682734-55682734 on Build GRCh38
rs376147722 Chr.7: 55683125-55683125 on Build GRCh38
rs4074683 Chr.7: 55682978-55682978 on Build GRCh38
rs746598849 Chr.7: 55682807-55682807 on Build GRCh38
rs764944511 Chr.7: 55682682-55682682 on Build GRCh38
rs187722813 Chr.7: 55682916-55682916 on Build GRCh38
rs756244269 Chr.7: 55682869-55682869 on Build GRCh38
rs1033093930 Chr.7: 55682758-55682758 on Build GRCh38
rs759780242 Chr.7: 55682862-55682862 on Build GRCh38
rs751380116 Chr.7: 55682857-55682857 on Build GRCh38
rs374488397 Chr.7: 55682863-55682863 on Build GRCh38
rs749846634 Chr.7: 55682803-55682803 on Build GRCh38
rs201294758 Chr.7: 55682833-55682833 on Build GRCh38
rs751728528 Chr.7: 55682767-55682767 on Build GRCh38
rs187579557 Chr.7: 55682835-55682835 on Build GRCh38
rs755259327 Chr.7: 55682777-55682777 on Build GRCh38
rs1028183938 Chr.7: 55682979-55682979 on Build GRCh38
rs148705904 Chr.7: 55682730-55682730 on Build GRCh38
rs7801173 Chr.7: 55683108-55683108 on Build GRCh38
rs761679833 Chr.7: 55682732-55682732 on Build GRCh38
rs761281298 Chr.7: 55682827-55682827 on Build GRCh38
rs183386787 Chr.7: 55682888-55682888 on Build GRCh38
rs779398793 Chr.7: 55682806-55682806 on Build GRCh38
rs757250872 Chr.7: 55682666-55682666 on Build GRCh38
rs142346795 Chr.7: 55682731-55682731 on Build GRCh38
rs774981392 Chr.7: 55682726-55682726 on Build GRCh38
rs146600095 Chr.7: 55683110-55683110 on Build GRCh38
rs771183920 Chr.7: 55682805-55682805 on Build GRCh38
rs760431367 Chr.7: 55682728-55682728 on Build GRCh38
rs777651292 Chr.7: 55682830-55682830 on Build GRCh38
rs193084168 Chr.7: 55683068-55683068 on Build GRCh38
rs772117176 Chr.7: 55682901-55682902 on Build GRCh38
rs182929013 Chr.7: 55682709-55682709 on Build GRCh38
rs748602643 Chr.7: 55682720-55682720 on Build GRCh38
rs565707870 Chr.7: 55683135-55683135 on Build GRCh38

View Full Product Details