Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55698968-55699457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP9P1
Entrez Gene ID
360132
Gene Name
FK506 binding protein 9 pseudogene 1
Gene Aliases
FKBP9L
UniGene
Hs.446691
Gene Chromosome Location
Chr.7: 55681074-55704567 on Build GRCh38
Transcripts
NR_003949.1

SNP Detail

SNP ID Chromosome Location
rs544204861 Chr.7: 55699226-55699226 on Build GRCh38
rs536985125 Chr.7: 55699102-55699102 on Build GRCh38
rs561106928 Chr.7: 55699257-55699257 on Build GRCh38
rs756541546 Chr.7: 55699044-55699044 on Build GRCh38
rs749177545 Chr.7: 55699140-55699140 on Build GRCh38
rs62455757 Chr.7: 55699314-55699314 on Build GRCh38
rs267601546 Chr.7: 55699012-55699012 on Build GRCh38
rs558803286 Chr.7: 55699149-55699149 on Build GRCh38
rs764141014 Chr.7: 55699116-55699116 on Build GRCh38
rs779317454 Chr.7: 55699062-55699062 on Build GRCh38
rs780875099 Chr.7: 55699130-55699130 on Build GRCh38
rs555187696 Chr.7: 55699122-55699122 on Build GRCh38
rs974337687 Chr.7: 55699398-55699398 on Build GRCh38
rs199530907 Chr.7: 55699085-55699085 on Build GRCh38
rs774299276 Chr.7: 55699103-55699103 on Build GRCh38
rs528287191 Chr.7: 55699265-55699265 on Build GRCh38
rs867828639 Chr.7: 55699367-55699367 on Build GRCh38
rs144227170 Chr.7: 55699248-55699248 on Build GRCh38
rs749300544 Chr.7: 55699100-55699100 on Build GRCh38
rs573387914 Chr.7: 55699134-55699134 on Build GRCh38
rs755051584 Chr.7: 55699037-55699038 on Build GRCh38
rs767644285 Chr.7: 55699037-55699037 on Build GRCh38
rs548624636 Chr.7: 55699368-55699368 on Build GRCh38
rs759780295 Chr.7: 55699105-55699105 on Build GRCh38
rs1002267173 Chr.7: 55699219-55699219 on Build GRCh38
rs759840981 Chr.7: 55699035-55699035 on Build GRCh38
rs940232197 Chr.7: 55699387-55699387 on Build GRCh38
rs143693502 Chr.7: 55699323-55699323 on Build GRCh38
rs760025242 Chr.7: 55699337-55699337 on Build GRCh38
rs759906854 Chr.7: 55699365-55699365 on Build GRCh38
rs762312103 Chr.7: 55699121-55699121 on Build GRCh38
rs772712768 Chr.7: 55699055-55699055 on Build GRCh38
rs954520230 Chr.7: 55699339-55699339 on Build GRCh38
rs757606419 Chr.7: 55699057-55699057 on Build GRCh38
rs534273944 Chr.7: 55699148-55699148 on Build GRCh38
rs577128520 Chr.7: 55699215-55699215 on Build GRCh38
rs73350457 Chr.7: 55699000-55699000 on Build GRCh38
rs979226083 Chr.7: 55699032-55699032 on Build GRCh38
rs545252281 Chr.7: 55699386-55699386 on Build GRCh38
rs760790299 Chr.7: 55699113-55699113 on Build GRCh38
rs562591274 Chr.7: 55699235-55699235 on Build GRCh38
rs767814477 Chr.7: 55699112-55699112 on Build GRCh38
rs530397773 Chr.7: 55699359-55699359 on Build GRCh38
rs773062944 Chr.7: 55699031-55699031 on Build GRCh38
rs764635105 Chr.7: 55699050-55699050 on Build GRCh38
rs771524279 Chr.7: 55699096-55699096 on Build GRCh38
rs1055628379 Chr.7: 55699229-55699229 on Build GRCh38
rs769256384 Chr.7: 55699034-55699034 on Build GRCh38
rs186076897 Chr.7: 55699303-55699303 on Build GRCh38
rs752847666 Chr.7: 55699039-55699039 on Build GRCh38
rs780526948 Chr.7: 55699076-55699076 on Build GRCh38
rs540283119 Chr.7: 55699283-55699283 on Build GRCh38
rs753872292 Chr.7: 55698998-55699000 on Build GRCh38
rs555893172 Chr.7: 55699135-55699135 on Build GRCh38
rs568907819 Chr.7: 55699345-55699345 on Build GRCh38
rs149991473 Chr.7: 55699239-55699239 on Build GRCh38
rs758801682 Chr.7: 55699126-55699126 on Build GRCh38
rs920058394 Chr.7: 55699405-55699405 on Build GRCh38
rs762785800 Chr.7: 55699032-55699032 on Build GRCh38
rs754640270 Chr.7: 55699073-55699073 on Build GRCh38
rs550584627 Chr.7: 55699327-55699327 on Build GRCh38
rs752350221 Chr.7: 55699133-55699133 on Build GRCh38
rs769800438 Chr.7: 55699022-55699022 on Build GRCh38
rs766331874 Chr.7: 55699033-55699033 on Build GRCh38
rs746723152 Chr.7: 55698999-55698999 on Build GRCh38
rs754366285 Chr.7: 55699119-55699119 on Build GRCh38
rs746600486 Chr.7: 55699063-55699063 on Build GRCh38
rs776211879 Chr.7: 55699013-55699013 on Build GRCh38
rs953230134 Chr.7: 55699064-55699064 on Build GRCh38

View Full Product Details