Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 65755297-65755846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT6P1
Entrez Gene ID
643253
Gene Name
chaperonin containing TCP1 subunit 6 pseudogene 1
Gene Aliases
CCT6-5P, CCT6AP1
UniGene
Hs.488399
Gene Chromosome Location
Chr.7: 65751105-65763675 on Build GRCh38
Transcripts
NR_003110.2 NR_038378.4 NR_002961.1

Gene Symbol
LOC441242
Entrez Gene ID
441242
Gene Name
uncharacterized LOC441242
Gene Aliases
-
UniGene
Hs.373941
Gene Chromosome Location
Chr.7: 65647010-65770810 on Build GRCh38
Transcripts
NR_003110.2 NR_038378.4 NR_002961.1

Gene Symbol
SNORA22
Entrez Gene ID
677807
Gene Name
small nucleolar RNA, H/ACA box 22
Gene Aliases
ACA22, SNORA22A
UniGene
Hs.675336
Gene Chromosome Location
Chr.7: 65755526-65755659 on Build GRCh38
Transcripts
NR_003110.2 NR_038378.4 NR_002961.1

SNP Detail

SNP ID Chromosome Location
rs782193869 Chr.7: 65755480-65755480 on Build GRCh38
rs782652181 Chr.7: 65755680-65755680 on Build GRCh38
rs781783365 Chr.7: 65755668-65755668 on Build GRCh38
rs180758124 Chr.7: 65755699-65755699 on Build GRCh38
rs540587262 Chr.7: 65755776-65755776 on Build GRCh38
rs782403625 Chr.7: 65755582-65755582 on Build GRCh38
rs538175788 Chr.7: 65755587-65755587 on Build GRCh38
rs782770215 Chr.7: 65755496-65755496 on Build GRCh38
rs561422776 Chr.7: 65755714-65755714 on Build GRCh38
rs541031160 Chr.7: 65755616-65755616 on Build GRCh38
rs34167201 Chr.7: 65755476-65755477 on Build GRCh38
rs533475355 Chr.7: 65755477-65755477 on Build GRCh38
rs184973623 Chr.7: 65755711-65755711 on Build GRCh38
rs376933914 Chr.7: 65755693-65755693 on Build GRCh38
rs782029129 Chr.7: 65755485-65755485 on Build GRCh38
rs4718275 Chr.7: 65755476-65755476 on Build GRCh38
rs535904541 Chr.7: 65755521-65755521 on Build GRCh38
rs370067626 Chr.7: 65755502-65755502 on Build GRCh38
rs782722049 Chr.7: 65755665-65755665 on Build GRCh38
rs782304245 Chr.7: 65755483-65755483 on Build GRCh38
rs782108103 Chr.7: 65755650-65755650 on Build GRCh38
rs782000470 Chr.7: 65755585-65755585 on Build GRCh38
rs375819971 Chr.7: 65755476-65755476 on Build GRCh38
rs782571858 Chr.7: 65755560-65755560 on Build GRCh38
rs782149805 Chr.7: 65755486-65755486 on Build GRCh38
rs372692697 Chr.7: 65755649-65755649 on Build GRCh38
rs782242884 Chr.7: 65755590-65755590 on Build GRCh38
rs782294371 Chr.7: 65755704-65755704 on Build GRCh38
rs782289030 Chr.7: 65755574-65755574 on Build GRCh38
rs550323169 Chr.7: 65755511-65755511 on Build GRCh38
rs782783096 Chr.7: 65755648-65755648 on Build GRCh38
rs753701027 Chr.7: 65755774-65755774 on Build GRCh38
rs782232641 Chr.7: 65755544-65755544 on Build GRCh38
rs782155257 Chr.7: 65755637-65755637 on Build GRCh38
rs781894510 Chr.7: 65755519-65755519 on Build GRCh38
rs782746940 Chr.7: 65755504-65755504 on Build GRCh38
rs760140835 Chr.7: 65755656-65755660 on Build GRCh38
rs555889722 Chr.7: 65755607-65755607 on Build GRCh38
rs782354839 Chr.7: 65755606-65755606 on Build GRCh38
rs369279072 Chr.7: 65755706-65755706 on Build GRCh38
rs781872499 Chr.7: 65755682-65755682 on Build GRCh38
rs782014673 Chr.7: 65755473-65755473 on Build GRCh38
rs188283364 Chr.7: 65755410-65755410 on Build GRCh38
rs566226165 Chr.7: 65755612-65755612 on Build GRCh38
rs4718276 Chr.7: 65755710-65755710 on Build GRCh38
rs782442261 Chr.7: 65755546-65755546 on Build GRCh38
rs577181239 Chr.7: 65755695-65755695 on Build GRCh38
rs782500089 Chr.7: 65755689-65755689 on Build GRCh38
rs570159166 Chr.7: 65755518-65755518 on Build GRCh38
rs782441144 Chr.7: 65755517-65755517 on Build GRCh38
rs527286365 Chr.7: 65755351-65755351 on Build GRCh38
rs527416068 Chr.7: 65755758-65755758 on Build GRCh38
rs782558442 Chr.7: 65755678-65755678 on Build GRCh38
rs563764586 Chr.7: 65755802-65755802 on Build GRCh38
rs782630408 Chr.7: 65755530-65755530 on Build GRCh38

View Full Product Details